Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83747955-83748509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKAP1
Entrez Gene ID
80318
Gene Name
G kinase anchoring protein 1
Gene Aliases
FKSG21, GKAP42
UniGene
Hs.522255
Gene Chromosome Location
Chr.9: 83739421-83817837 on Build GRCh38
Transcripts
NM_001135953.1 NM_025211.3 XM_005252241.2 XM_006717293.1 XM_011519058.2 XM_017015171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671323 c.869delA p.N290fs*5 Chr.9: 83748344-83748344 on Build GRCh38
6434407 c.888G>T p.M296I Chr.9: 83748325-83748325 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs988897117 Chr.9: 83748392-83748392 on Build GRCh38
rs200525497 Chr.9: 83748398-83748398 on Build GRCh38
rs551091746 Chr.9: 83748196-83748196 on Build GRCh38
rs926033681 Chr.9: 83747993-83747994 on Build GRCh38
rs141952900 Chr.9: 83748337-83748337 on Build GRCh38
rs752745834 Chr.9: 83748261-83748263 on Build GRCh38
rs894618323 Chr.9: 83748083-83748083 on Build GRCh38
rs947300127 Chr.9: 83748209-83748209 on Build GRCh38
rs900739492 Chr.9: 83748109-83748109 on Build GRCh38
rs75996055 Chr.9: 83748060-83748060 on Build GRCh38
rs370560418 Chr.9: 83748288-83748288 on Build GRCh38
rs972347321 Chr.9: 83748116-83748116 on Build GRCh38
rs910642055 Chr.9: 83748169-83748169 on Build GRCh38
rs776861182 Chr.9: 83748264-83748264 on Build GRCh38
rs781277893 Chr.9: 83748407-83748407 on Build GRCh38
rs944894650 Chr.9: 83748433-83748433 on Build GRCh38
rs922193277 Chr.9: 83748449-83748449 on Build GRCh38
rs749911516 Chr.9: 83748474-83748474 on Build GRCh38
rs1023578086 Chr.9: 83748289-83748289 on Build GRCh38
rs902893435 Chr.9: 83748054-83748054 on Build GRCh38
rs1049242310 Chr.9: 83748478-83748478 on Build GRCh38
rs1026311080 Chr.9: 83748119-83748119 on Build GRCh38
rs1035289947 Chr.9: 83748202-83748202 on Build GRCh38
rs769026151 Chr.9: 83748263-83748263 on Build GRCh38
rs752093981 Chr.9: 83748408-83748411 on Build GRCh38
rs756853935 Chr.9: 83748363-83748363 on Build GRCh38
rs760140567 Chr.9: 83748336-83748336 on Build GRCh38
rs1039596596 Chr.9: 83747991-83747991 on Build GRCh38
rs780663377 Chr.9: 83748387-83748387 on Build GRCh38
rs541500487 Chr.9: 83748385-83748386 on Build GRCh38
rs752245123 Chr.9: 83748300-83748300 on Build GRCh38
rs944987936 Chr.9: 83748072-83748072 on Build GRCh38
rs145264205 Chr.9: 83748294-83748294 on Build GRCh38
rs532988515 Chr.9: 83748179-83748179 on Build GRCh38
rs934729405 Chr.9: 83748022-83748027 on Build GRCh38
rs530544079 Chr.9: 83748219-83748219 on Build GRCh38
rs930900394 Chr.9: 83748476-83748476 on Build GRCh38
rs534347079 Chr.9: 83747981-83747981 on Build GRCh38
rs757551445 Chr.9: 83748420-83748423 on Build GRCh38
rs759009975 Chr.9: 83748381-83748381 on Build GRCh38
rs183832155 Chr.9: 83748103-83748103 on Build GRCh38
rs559375722 Chr.9: 83748149-83748149 on Build GRCh38
rs893041869 Chr.9: 83748035-83748035 on Build GRCh38
rs376120919 Chr.9: 83748057-83748057 on Build GRCh38
rs1041626961 Chr.9: 83748313-83748313 on Build GRCh38
rs370107727 Chr.9: 83748293-83748293 on Build GRCh38
rs1012060690 Chr.9: 83748223-83748223 on Build GRCh38
rs763073600 Chr.9: 83748263-83748266 on Build GRCh38
rs956341512 Chr.9: 83748382-83748387 on Build GRCh38
rs141221414 Chr.9: 83748036-83748036 on Build GRCh38
rs963603445 Chr.9: 83748113-83748113 on Build GRCh38
rs371496329 Chr.9: 83748394-83748394 on Build GRCh38
rs943497024 Chr.9: 83748189-83748189 on Build GRCh38
rs1041364730 Chr.9: 83748040-83748040 on Build GRCh38
rs974840833 Chr.9: 83748448-83748448 on Build GRCh38
rs149678796 Chr.9: 83748340-83748340 on Build GRCh38
rs144989367 Chr.9: 83748462-83748462 on Build GRCh38
rs763858908 Chr.9: 83748156-83748156 on Build GRCh38
rs779392512 Chr.9: 83748372-83748372 on Build GRCh38
rs1013514141 Chr.9: 83748091-83748091 on Build GRCh38
rs563263318 Chr.9: 83748218-83748218 on Build GRCh38
rs750730303 Chr.9: 83748377-83748377 on Build GRCh38
rs751180308 Chr.9: 83748200-83748200 on Build GRCh38
rs899364207 Chr.9: 83748484-83748484 on Build GRCh38
rs767421016 Chr.9: 83748296-83748296 on Build GRCh38

View Full Product Details