Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55676742-55677193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN1
Entrez Gene ID
29924
Gene Name
epsin 1
Gene Aliases
-
UniGene
Hs.279953
Gene Chromosome Location
Chr.19: 55675195-55695767 on Build GRCh38
Transcripts
NM_001130071.1 NM_001130072.1 NM_013333.3 XM_005258829.2 XM_005258830.2 XM_011526880.1 XM_011526881.1 XM_017026722.1 XM_017026723.1 XM_017026724.1 XM_017026725.1

SNP Detail

SNP ID Chromosome Location
rs960372790 Chr.19: 55676773-55676774 on Build GRCh38
rs150008375 Chr.19: 55676770-55676770 on Build GRCh38
rs558088737 Chr.19: 55676806-55676806 on Build GRCh38
rs374112040 Chr.19: 55676996-55676996 on Build GRCh38
rs752003829 Chr.19: 55677088-55677088 on Build GRCh38
rs529011885 Chr.19: 55677015-55677015 on Build GRCh38
rs767318795 Chr.19: 55677157-55677157 on Build GRCh38
rs902743281 Chr.19: 55676960-55676960 on Build GRCh38
rs889993517 Chr.19: 55676913-55676913 on Build GRCh38
rs552656142 Chr.19: 55677085-55677085 on Build GRCh38
rs930635691 Chr.19: 55677103-55677103 on Build GRCh38
rs939650392 Chr.19: 55677007-55677007 on Build GRCh38
rs999613584 Chr.19: 55676974-55676974 on Build GRCh38
rs530722268 Chr.19: 55677148-55677148 on Build GRCh38
rs1049624314 Chr.19: 55677138-55677138 on Build GRCh38
rs774345918 Chr.19: 55677122-55677122 on Build GRCh38
rs572523099 Chr.19: 55676793-55676793 on Build GRCh38
rs562979644 Chr.19: 55677093-55677093 on Build GRCh38
rs746413304 Chr.19: 55676823-55676823 on Build GRCh38
rs540603254 Chr.19: 55676988-55676988 on Build GRCh38
rs755543612 Chr.19: 55676848-55676851 on Build GRCh38
rs752417436 Chr.19: 55677127-55677127 on Build GRCh38
rs140345053 Chr.19: 55676896-55676896 on Build GRCh38
rs532570399 Chr.19: 55677036-55677036 on Build GRCh38
rs1037005720 Chr.19: 55676852-55676852 on Build GRCh38
rs761976240 Chr.19: 55676897-55676897 on Build GRCh38
rs900631564 Chr.19: 55676889-55676889 on Build GRCh38
rs544522989 Chr.19: 55676905-55676905 on Build GRCh38
rs367545855 Chr.19: 55676860-55676860 on Build GRCh38
rs538320332 Chr.19: 55676794-55676794 on Build GRCh38
rs11667416 Chr.19: 55676815-55676815 on Build GRCh38
rs921942335 Chr.19: 55677095-55677095 on Build GRCh38
rs990470799 Chr.19: 55676788-55676789 on Build GRCh38
rs75998004 Chr.19: 55676788-55676788 on Build GRCh38
rs1011068094 Chr.19: 55676935-55676935 on Build GRCh38
rs766783413 Chr.19: 55677038-55677038 on Build GRCh38
rs960593444 Chr.19: 55676941-55676941 on Build GRCh38
rs1039229301 Chr.19: 55677094-55677094 on Build GRCh38
rs761696135 Chr.19: 55677150-55677150 on Build GRCh38
rs944734383 Chr.19: 55676848-55676848 on Build GRCh38
rs772963550 Chr.19: 55677165-55677165 on Build GRCh38
rs956434330 Chr.19: 55677032-55677032 on Build GRCh38
rs967671639 Chr.19: 55676995-55676995 on Build GRCh38
rs145413177 Chr.19: 55676976-55676976 on Build GRCh38
rs912133927 Chr.19: 55676847-55676847 on Build GRCh38
rs561849699 Chr.19: 55676940-55676940 on Build GRCh38
rs984564838 Chr.19: 55677100-55677100 on Build GRCh38
rs781653460 Chr.19: 55677108-55677108 on Build GRCh38
rs1039009793 Chr.19: 55676888-55676888 on Build GRCh38
rs978986945 Chr.19: 55676803-55676803 on Build GRCh38
rs922686243 Chr.19: 55676881-55676881 on Build GRCh38
rs886287884 Chr.19: 55676927-55676927 on Build GRCh38
rs8106718 Chr.19: 55677166-55677166 on Build GRCh38

View Full Product Details