Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132625525-132626074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX31
Entrez Gene ID
64794
Gene Name
DEAD-box helicase 31
Gene Aliases
PPP1R25
UniGene
Hs.660767
Gene Chromosome Location
Chr.9: 132592997-132670403 on Build GRCh38
Transcripts
NM_001322340.1 NM_001322341.1 NM_001322342.1 NM_001322343.1 NM_022779.8 NR_136309.1 XM_005272207.4 XM_006717236.3 XM_011518921.2 XM_011518922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6229935 c.2026C>T p.Q676* Chr.9: 132625666-132625666 on Build GRCh38
6229934 c.2026C>T p.Q676* Chr.9: 132625666-132625666 on Build GRCh38
4984274 c.2009G>T p.G670V Chr.9: 132625683-132625683 on Build GRCh38
6871423 c.1965G>A p.M655I Chr.9: 132625727-132625727 on Build GRCh38
3664192 c.2008G>A p.G670R Chr.9: 132625684-132625684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939541227 Chr.9: 132625958-132625958 on Build GRCh38
rs57378280 Chr.9: 132625916-132625916 on Build GRCh38
rs1022274839 Chr.9: 132625610-132625610 on Build GRCh38
rs781568454 Chr.9: 132625644-132625644 on Build GRCh38
rs947759246 Chr.9: 132625789-132625789 on Build GRCh38
rs756200268 Chr.9: 132625667-132625667 on Build GRCh38
rs919583247 Chr.9: 132625845-132625845 on Build GRCh38
rs777486731 Chr.9: 132625681-132625681 on Build GRCh38
rs775203733 Chr.9: 132625718-132625718 on Build GRCh38
rs764913539 Chr.9: 132625625-132625625 on Build GRCh38
rs891407164 Chr.9: 132625893-132625893 on Build GRCh38
rs373280508 Chr.9: 132625797-132625797 on Build GRCh38
rs944286398 Chr.9: 132625896-132625896 on Build GRCh38
rs900001292 Chr.9: 132625918-132625918 on Build GRCh38
rs1043747869 Chr.9: 132625986-132625986 on Build GRCh38
rs138460016 Chr.9: 132625745-132625745 on Build GRCh38
rs746829421 Chr.9: 132625687-132625687 on Build GRCh38
rs1031631735 Chr.9: 132626022-132626022 on Build GRCh38
rs763847617 Chr.9: 132625724-132625724 on Build GRCh38
rs748110807 Chr.9: 132625655-132625655 on Build GRCh38
rs929661333 Chr.9: 132625860-132625860 on Build GRCh38
rs769244173 Chr.9: 132625714-132625714 on Build GRCh38
rs369807608 Chr.9: 132625796-132625796 on Build GRCh38
rs764620078 Chr.9: 132625638-132625638 on Build GRCh38
rs998838374 Chr.9: 132626050-132626050 on Build GRCh38
rs1008428129 Chr.9: 132625556-132625556 on Build GRCh38
rs1055288348 Chr.9: 132625962-132625962 on Build GRCh38
rs999517970 Chr.9: 132625999-132625999 on Build GRCh38
rs967444841 Chr.9: 132625688-132625688 on Build GRCh38
rs991928432 Chr.9: 132625763-132625763 on Build GRCh38
rs895231657 Chr.9: 132625964-132625964 on Build GRCh38
rs910316945 Chr.9: 132625804-132625804 on Build GRCh38
rs201238129 Chr.9: 132625698-132625698 on Build GRCh38
rs369976336 Chr.9: 132625744-132625744 on Build GRCh38
rs757777020 Chr.9: 132625642-132625642 on Build GRCh38
rs112872016 Chr.9: 132626046-132626046 on Build GRCh38
rs998731252 Chr.9: 132625650-132625650 on Build GRCh38
rs751920599 Chr.9: 132625733-132625733 on Build GRCh38
rs964464370 Chr.9: 132625812-132625812 on Build GRCh38
rs373900113 Chr.9: 132625766-132625766 on Build GRCh38
rs759635919 Chr.9: 132625596-132625596 on Build GRCh38
rs916428519 Chr.9: 132625770-132625770 on Build GRCh38
rs559476442 Chr.9: 132625778-132625778 on Build GRCh38
rs1043182860 Chr.9: 132626039-132626039 on Build GRCh38
rs545072932 Chr.9: 132625869-132625869 on Build GRCh38
rs73659424 Chr.9: 132625669-132625669 on Build GRCh38
rs756217983 Chr.9: 132625751-132625751 on Build GRCh38
rs376410126 Chr.9: 132625722-132625722 on Build GRCh38
rs377431043 Chr.9: 132625560-132625560 on Build GRCh38
rs902478363 Chr.9: 132625996-132625996 on Build GRCh38
rs759062105 Chr.9: 132625700-132625700 on Build GRCh38
rs929569749 Chr.9: 132625878-132625878 on Build GRCh38
rs763395146 Chr.9: 132625636-132625636 on Build GRCh38
rs762627330 Chr.9: 132625719-132625719 on Build GRCh38
rs866416835 Chr.9: 132625692-132625692 on Build GRCh38
rs549690532 Chr.9: 132625930-132625930 on Build GRCh38
rs185340839 Chr.9: 132625821-132625821 on Build GRCh38
rs1010969851 Chr.9: 132625968-132625968 on Build GRCh38
rs561964571 Chr.9: 132626021-132626021 on Build GRCh38
rs751976063 Chr.9: 132625639-132625639 on Build GRCh38
rs770817452 Chr.9: 132625691-132625691 on Build GRCh38
rs749543596 Chr.9: 132625677-132625677 on Build GRCh38
rs1012073658 Chr.9: 132625983-132625983 on Build GRCh38
rs372472395 Chr.9: 132625629-132625629 on Build GRCh38
rs113758089 Chr.9: 132625568-132625568 on Build GRCh38
rs755144199 Chr.9: 132625786-132625786 on Build GRCh38
rs555960510 Chr.9: 132625678-132625678 on Build GRCh38
rs200774243 Chr.9: 132625665-132625665 on Build GRCh38
rs895215414 Chr.9: 132625557-132625557 on Build GRCh38
rs376054607 Chr.9: 132625870-132625870 on Build GRCh38
rs17149380 Chr.9: 132625557-132625557 on Build GRCh38

View Full Product Details