Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132611818-132612377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX31
Entrez Gene ID
64794
Gene Name
DEAD-box helicase 31
Gene Aliases
PPP1R25
UniGene
Hs.660767
Gene Chromosome Location
Chr.9: 132592997-132670403 on Build GRCh38
Transcripts
NM_001322340.1 NM_001322341.1 NM_001322342.1 NM_001322343.1 NM_022779.8 NR_136309.1 XM_005272207.4 XM_006717236.3 XM_011518921.2 XM_011518922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3905121 c.2225G>A p.R742H Chr.9: 132612229-132612229 on Build GRCh38
4536280 c.2286G>A p.K762K Chr.9: 132612110-132612110 on Build GRCh38
4477477 c.2215C>T p.P739S Chr.9: 132612239-132612239 on Build GRCh38
1203245 c.2194A>G p.I732V Chr.9: 132612202-132612202 on Build GRCh38
3212622 c.2302G>A p.V768M Chr.9: 132612094-132612094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749663144 Chr.9: 132612289-132612289 on Build GRCh38
rs150568440 Chr.9: 132612143-132612143 on Build GRCh38
rs746707235 Chr.9: 132612076-132612076 on Build GRCh38
rs748740427 Chr.9: 132612223-132612224 on Build GRCh38
rs906322213 Chr.9: 132612022-132612022 on Build GRCh38
rs772255493 Chr.9: 132612304-132612304 on Build GRCh38
rs1046173200 Chr.9: 132612014-132612014 on Build GRCh38
rs746543127 Chr.9: 132612158-132612158 on Build GRCh38
rs750060351 Chr.9: 132612237-132612240 on Build GRCh38
rs767806336 Chr.9: 132612128-132612128 on Build GRCh38
rs952223087 Chr.9: 132611886-132611886 on Build GRCh38
rs1013663647 Chr.9: 132612269-132612269 on Build GRCh38
rs369733110 Chr.9: 132612351-132612351 on Build GRCh38
rs368755662 Chr.9: 132612044-132612044 on Build GRCh38
rs887949772 Chr.9: 132612210-132612210 on Build GRCh38
rs767606249 Chr.9: 132612193-132612193 on Build GRCh38
rs769007026 Chr.9: 132612294-132612294 on Build GRCh38
rs758502438 Chr.9: 132612036-132612036 on Build GRCh38
rs750991746 Chr.9: 132612230-132612230 on Build GRCh38
rs777080941 Chr.9: 132612155-132612155 on Build GRCh38
rs754922101 Chr.9: 132612061-132612061 on Build GRCh38
rs550221533 Chr.9: 132612024-132612024 on Build GRCh38
rs572017150 Chr.9: 132612272-132612272 on Build GRCh38
rs962155678 Chr.9: 132612264-132612264 on Build GRCh38
rs774250665 Chr.9: 132612189-132612189 on Build GRCh38
rs751775620 Chr.9: 132612042-132612042 on Build GRCh38
rs548076766 Chr.9: 132611973-132611973 on Build GRCh38
rs140122725 Chr.9: 132612352-132612352 on Build GRCh38
rs957512364 Chr.9: 132612315-132612315 on Build GRCh38
rs945821075 Chr.9: 132612321-132612321 on Build GRCh38
rs1014977010 Chr.9: 132611915-132611915 on Build GRCh38
rs764266174 Chr.9: 132612037-132612037 on Build GRCh38
rs368964816 Chr.9: 132612084-132612084 on Build GRCh38
rs913844563 Chr.9: 132612225-132612225 on Build GRCh38
rs961864621 Chr.9: 132611837-132611837 on Build GRCh38
rs370585772 Chr.9: 132612150-132612150 on Build GRCh38
rs1037283188 Chr.9: 132612328-132612328 on Build GRCh38
rs749938364 Chr.9: 132612355-132612355 on Build GRCh38
rs373679561 Chr.9: 132612295-132612295 on Build GRCh38
rs759393060 Chr.9: 132612209-132612209 on Build GRCh38
rs562246111 Chr.9: 132611912-132611912 on Build GRCh38
rs148432033 Chr.9: 132612094-132612094 on Build GRCh38
rs147506196 Chr.9: 132612095-132612095 on Build GRCh38
rs769152913 Chr.9: 132612185-132612185 on Build GRCh38
rs750709275 Chr.9: 132612194-132612194 on Build GRCh38
rs781610315 Chr.9: 132612198-132612198 on Build GRCh38
rs139557900 Chr.9: 132612178-132612178 on Build GRCh38
rs190226377 Chr.9: 132612068-132612068 on Build GRCh38
rs533131295 Chr.9: 132612028-132612028 on Build GRCh38
rs746198151 Chr.9: 132612177-132612177 on Build GRCh38
rs981948722 Chr.9: 132612238-132612238 on Build GRCh38
rs754863844 Chr.9: 132612148-132612148 on Build GRCh38
rs373400046 Chr.9: 132611851-132611851 on Build GRCh38
rs751797661 Chr.9: 132612111-132612111 on Build GRCh38
rs762870849 Chr.9: 132612322-132612322 on Build GRCh38
rs922953070 Chr.9: 132612327-132612327 on Build GRCh38
rs539392456 Chr.9: 132612308-132612308 on Build GRCh38
rs2165579 Chr.9: 132612110-132612110 on Build GRCh38
rs759967386 Chr.9: 132612087-132612087 on Build GRCh38
rs1009817062 Chr.9: 132611860-132611860 on Build GRCh38
rs545539027 Chr.9: 132611978-132611978 on Build GRCh38
rs985430715 Chr.9: 132611862-132611862 on Build GRCh38
rs989698472 Chr.9: 132611914-132611914 on Build GRCh38
rs749125486 Chr.9: 132612059-132612059 on Build GRCh38
rs763905126 Chr.9: 132612338-132612338 on Build GRCh38
rs752492961 Chr.9: 132612320-132612320 on Build GRCh38
rs770134913 Chr.9: 132612278-132612278 on Build GRCh38
rs758353390 Chr.9: 132612229-132612229 on Build GRCh38
rs1054188737 Chr.9: 132611842-132611842 on Build GRCh38
rs115033964 Chr.9: 132611969-132611969 on Build GRCh38
rs576874553 Chr.9: 132611843-132611843 on Build GRCh38
rs927200455 Chr.9: 132612296-132612296 on Build GRCh38
rs954042669 Chr.9: 132611861-132611861 on Build GRCh38
rs757979307 Chr.9: 132612154-132612154 on Build GRCh38
rs910437059 Chr.9: 132611874-132611874 on Build GRCh38
rs761780932 Chr.9: 132612300-132612300 on Build GRCh38
rs996429609 Chr.9: 132611868-132611868 on Build GRCh38
rs780327707 Chr.9: 132612283-132612283 on Build GRCh38
rs927668065 Chr.9: 132612248-132612248 on Build GRCh38
rs745575013 Chr.9: 132612273-132612273 on Build GRCh38
rs756832277 Chr.9: 132612256-132612256 on Build GRCh38
rs373015847 Chr.9: 132612280-132612280 on Build GRCh38
rs966568781 Chr.9: 132612236-132612236 on Build GRCh38
rs760914075 Chr.9: 132612314-132612314 on Build GRCh38
rs779028067 Chr.9: 132612063-132612063 on Build GRCh38
rs755728832 Chr.9: 132612268-132612269 on Build GRCh38
rs780928341 Chr.9: 132612170-132612170 on Build GRCh38
rs373977389 Chr.9: 132612218-132612218 on Build GRCh38
rs116392521 Chr.9: 132611902-132611902 on Build GRCh38
rs962041837 Chr.9: 132611964-132611964 on Build GRCh38
rs989400739 Chr.9: 132612203-132612203 on Build GRCh38
rs764214695 Chr.9: 132612107-132612107 on Build GRCh38
rs935774950 Chr.9: 132611838-132611838 on Build GRCh38
rs576078478 Chr.9: 132611845-132611845 on Build GRCh38
rs770441677 Chr.9: 132612164-132612164 on Build GRCh38
rs755919516 Chr.9: 132612043-132612043 on Build GRCh38
rs917916915 Chr.9: 132612006-132612006 on Build GRCh38
rs757351589 Chr.9: 132612116-132612116 on Build GRCh38
rs374832274 Chr.9: 132612069-132612069 on Build GRCh38
rs969355965 Chr.9: 132612282-132612282 on Build GRCh38
rs763513866 Chr.9: 132612179-132612179 on Build GRCh38
rs752545960 Chr.9: 132612221-132612221 on Build GRCh38
rs372159165 Chr.9: 132612064-132612064 on Build GRCh38
rs770622225 Chr.9: 132612083-132612083 on Build GRCh38
rs945231879 Chr.9: 132612005-132612005 on Build GRCh38
rs766723368 Chr.9: 132612319-132612319 on Build GRCh38
rs751564129 Chr.9: 132612341-132612341 on Build GRCh38
rs748155369 Chr.9: 132612153-132612153 on Build GRCh38

View Full Product Details