Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15287757-15288229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB11
Entrez Gene ID
140456
Gene Name
ankyrin repeat and SOCS box containing 11
Gene Aliases
-
UniGene
Hs.352183
Gene Chromosome Location
Chr.X: 15281697-15315624 on Build GRCh38
Transcripts
NM_001012428.2 NM_001201583.1 NM_080873.2

SNP Detail

SNP ID Chromosome Location
rs749788594 Chr.X: 15287995-15287995 on Build GRCh38
rs773871027 Chr.X: 15287887-15287887 on Build GRCh38
rs35859007 Chr.X: 15287941-15287941 on Build GRCh38
rs61756318 Chr.X: 15287880-15287880 on Build GRCh38
rs137906487 Chr.X: 15288020-15288020 on Build GRCh38
rs781564627 Chr.X: 15287961-15287961 on Build GRCh38
rs760467867 Chr.X: 15287908-15287908 on Build GRCh38
rs759155699 Chr.X: 15288162-15288162 on Build GRCh38
rs34670756 Chr.X: 15288203-15288204 on Build GRCh38
rs34025595 Chr.X: 15287983-15287983 on Build GRCh38
rs764279966 Chr.X: 15287937-15287937 on Build GRCh38
rs775137827 Chr.X: 15288021-15288021 on Build GRCh38
rs745370216 Chr.X: 15288017-15288017 on Build GRCh38
rs765212160 Chr.X: 15288062-15288062 on Build GRCh38
rs759150211 Chr.X: 15287888-15287888 on Build GRCh38
rs770874074 Chr.X: 15288144-15288144 on Build GRCh38
rs776458541 Chr.X: 15288042-15288042 on Build GRCh38
rs755198846 Chr.X: 15287843-15287843 on Build GRCh38
rs201156950 Chr.X: 15287909-15287909 on Build GRCh38
rs766413011 Chr.X: 15287939-15287939 on Build GRCh38
rs746617167 Chr.X: 15288035-15288035 on Build GRCh38
rs778231280 Chr.X: 15287984-15287984 on Build GRCh38
rs368639487 Chr.X: 15287936-15287936 on Build GRCh38
rs772940413 Chr.X: 15288002-15288003 on Build GRCh38
rs867166879 Chr.X: 15288113-15288113 on Build GRCh38
rs199601166 Chr.X: 15287840-15287840 on Build GRCh38
rs947862737 Chr.X: 15288044-15288044 on Build GRCh38
rs960420120 Chr.X: 15288125-15288125 on Build GRCh38
rs755521735 Chr.X: 15287823-15287823 on Build GRCh38
rs142033679 Chr.X: 15287955-15287955 on Build GRCh38
rs768472092 Chr.X: 15288036-15288036 on Build GRCh38
rs560610195 Chr.X: 15288204-15288204 on Build GRCh38
rs761707523 Chr.X: 15288061-15288061 on Build GRCh38
rs778334178 Chr.X: 15288011-15288011 on Build GRCh38
rs778425250 Chr.X: 15287867-15287867 on Build GRCh38
rs774365256 Chr.X: 15288077-15288077 on Build GRCh38
rs781726059 Chr.X: 15287844-15287844 on Build GRCh38
rs977151570 Chr.X: 15287835-15287835 on Build GRCh38
rs771690353 Chr.X: 15287890-15287890 on Build GRCh38
rs960099666 Chr.X: 15287791-15287791 on Build GRCh38
rs915903725 Chr.X: 15288166-15288166 on Build GRCh38
rs373647947 Chr.X: 15287875-15287875 on Build GRCh38
rs1046473083 Chr.X: 15288055-15288055 on Build GRCh38
rs1029692863 Chr.X: 15287837-15287837 on Build GRCh38
rs771435987 Chr.X: 15288009-15288009 on Build GRCh38
rs752897584 Chr.X: 15287968-15287968 on Build GRCh38
rs188147009 Chr.X: 15287822-15287822 on Build GRCh38
rs776615453 Chr.X: 15288159-15288159 on Build GRCh38
rs1007517047 Chr.X: 15287962-15287962 on Build GRCh38
rs772045096 Chr.X: 15287886-15287886 on Build GRCh38
rs992804778 Chr.X: 15288128-15288128 on Build GRCh38
rs775320558 Chr.X: 15287906-15287906 on Build GRCh38
rs993570040 Chr.X: 15288043-15288043 on Build GRCh38
rs770486086 Chr.X: 15287865-15287865 on Build GRCh38
rs916143657 Chr.X: 15287827-15287827 on Build GRCh38
rs762657163 Chr.X: 15288096-15288096 on Build GRCh38
rs926155917 Chr.X: 15287896-15287896 on Build GRCh38
rs200705921 Chr.X: 15288111-15288111 on Build GRCh38
rs532675354 Chr.X: 15287861-15287861 on Build GRCh38
rs372168670 Chr.X: 15287952-15287952 on Build GRCh38
rs749744149 Chr.X: 15287872-15287872 on Build GRCh38

View Full Product Details