Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111295260-111295849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
UniGene
Hs.34780
Gene Chromosome Location
Chr.X: 111293779-111412232 on Build GRCh38
Transcripts
NM_000555.3 NM_001195553.1 NM_178151.2 NM_178152.2 NM_178153.2 XM_011530878.2 XM_011530879.2 XM_011530880.2 XM_017029312.1

SNP Detail

SNP ID Chromosome Location
rs952749950 Chr.X: 111295445-111295445 on Build GRCh38
rs146012202 Chr.X: 111295680-111295680 on Build GRCh38
rs779509815 Chr.X: 111295788-111295788 on Build GRCh38
rs911192295 Chr.X: 111295546-111295547 on Build GRCh38
rs892244838 Chr.X: 111295581-111295581 on Build GRCh38
rs748712470 Chr.X: 111295439-111295439 on Build GRCh38
rs1025644082 Chr.X: 111295290-111295290 on Build GRCh38
rs930074982 Chr.X: 111295783-111295783 on Build GRCh38
rs777843110 Chr.X: 111295475-111295475 on Build GRCh38
rs974688429 Chr.X: 111295724-111295724 on Build GRCh38
rs180686559 Chr.X: 111295360-111295360 on Build GRCh38
rs41304474 Chr.X: 111295436-111295436 on Build GRCh38
rs1032173060 Chr.X: 111295557-111295557 on Build GRCh38
rs980374215 Chr.X: 111295676-111295676 on Build GRCh38
rs970383800 Chr.X: 111295646-111295646 on Build GRCh38
rs1055870196 Chr.X: 111295744-111295744 on Build GRCh38
rs930076207 Chr.X: 111295405-111295405 on Build GRCh38
rs1039661696 Chr.X: 111295490-111295490 on Build GRCh38
rs935728361 Chr.X: 111295725-111295725 on Build GRCh38
rs748535261 Chr.X: 111295792-111295792 on Build GRCh38
rs899714153 Chr.X: 111295552-111295552 on Build GRCh38
rs142572190 Chr.X: 111295601-111295601 on Build GRCh38
rs920100018 Chr.X: 111295780-111295780 on Build GRCh38
rs1024617935 Chr.X: 111295639-111295639 on Build GRCh38
rs752919312 Chr.X: 111295306-111295306 on Build GRCh38
rs748512811 Chr.X: 111295337-111295338 on Build GRCh38
rs147995300 Chr.X: 111295479-111295479 on Build GRCh38
rs976737697 Chr.X: 111295565-111295565 on Build GRCh38
rs371611737 Chr.X: 111295493-111295493 on Build GRCh38
rs1025757824 Chr.X: 111295442-111295442 on Build GRCh38
rs185356653 Chr.X: 111295549-111295549 on Build GRCh38
rs922517125 Chr.X: 111295629-111295629 on Build GRCh38
rs1001109236 Chr.X: 111295555-111295555 on Build GRCh38
rs945914533 Chr.X: 111295648-111295648 on Build GRCh38
rs958947775 Chr.X: 111295720-111295720 on Build GRCh38
rs775845615 Chr.X: 111295690-111295690 on Build GRCh38
rs1046710605 Chr.X: 111295823-111295823 on Build GRCh38
rs977198827 Chr.X: 111295652-111295652 on Build GRCh38

View Full Product Details