Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119560163-119560705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf56
Entrez Gene ID
63932
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
UniGene
Hs.248572
Gene Chromosome Location
Chr.X: 119538149-119565434 on Build GRCh38
Transcripts
NM_001170569.1 NM_001170570.1 NM_022101.3

SNP Detail

SNP ID Chromosome Location
rs755125676 Chr.X: 119560253-119560253 on Build GRCh38
rs748260882 Chr.X: 119560256-119560261 on Build GRCh38
rs370194566 Chr.X: 119560235-119560235 on Build GRCh38
rs1011220701 Chr.X: 119560609-119560609 on Build GRCh38
rs768182141 Chr.X: 119560397-119560397 on Build GRCh38
rs766481675 Chr.X: 119560421-119560421 on Build GRCh38
rs748977581 Chr.X: 119560379-119560379 on Build GRCh38
rs903548560 Chr.X: 119560591-119560591 on Build GRCh38
rs757230589 Chr.X: 119560339-119560339 on Build GRCh38
rs768386894 Chr.X: 119560384-119560384 on Build GRCh38
rs762856669 Chr.X: 119560669-119560669 on Build GRCh38
rs891439037 Chr.X: 119560501-119560501 on Build GRCh38
rs746281553 Chr.X: 119560452-119560452 on Build GRCh38
rs370307624 Chr.X: 119560392-119560392 on Build GRCh38
rs34481136 Chr.X: 119560316-119560317 on Build GRCh38
rs773767111 Chr.X: 119560401-119560401 on Build GRCh38
rs972181766 Chr.X: 119560438-119560438 on Build GRCh38
rs901754560 Chr.X: 119560440-119560440 on Build GRCh38
rs73637855 Chr.X: 119560224-119560224 on Build GRCh38
rs145119663 Chr.X: 119560340-119560340 on Build GRCh38
rs753817932 Chr.X: 119560433-119560433 on Build GRCh38
rs947970128 Chr.X: 119560478-119560478 on Build GRCh38
rs1038901279 Chr.X: 119560420-119560420 on Build GRCh38
rs761026859 Chr.X: 119560404-119560404 on Build GRCh38
rs187814735 Chr.X: 119560346-119560346 on Build GRCh38
rs148441214 Chr.X: 119560281-119560281 on Build GRCh38
rs774807972 Chr.X: 119560353-119560353 on Build GRCh38
rs56158651 Chr.X: 119560265-119560265 on Build GRCh38
rs768189294 Chr.X: 119560455-119560455 on Build GRCh38
rs201896934 Chr.X: 119560347-119560347 on Build GRCh38
rs929303902 Chr.X: 119560315-119560315 on Build GRCh38
rs985036677 Chr.X: 119560341-119560341 on Build GRCh38
rs772733610 Chr.X: 119560636-119560636 on Build GRCh38
rs1056325914 Chr.X: 119560471-119560471 on Build GRCh38
rs144496881 Chr.X: 119560279-119560279 on Build GRCh38
rs769335743 Chr.X: 119560352-119560352 on Build GRCh38
rs376296913 Chr.X: 119560197-119560197 on Build GRCh38
rs943183688 Chr.X: 119560403-119560403 on Build GRCh38
rs191969993 Chr.X: 119560512-119560512 on Build GRCh38
rs72607659 Chr.X: 119560473-119560473 on Build GRCh38
rs764686674 Chr.X: 119560552-119560552 on Build GRCh38

View Full Product Details