Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186644619-186645137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FETUB
Entrez Gene ID
26998
Gene Name
fetuin B
Gene Aliases
16G2, Gugu, IRL685
UniGene
Hs.81073
Gene Chromosome Location
Chr.3: 186635828-186653008 on Build GRCh38
Transcripts
NM_001308077.1 NM_001308079.1 NM_014375.2 XM_005247351.3 XM_011512681.2 XM_011512682.2 XM_011512683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5995004 c.571A>G p.K191E Chr.3: 186644897-186644897 on Build GRCh38
446147 c.527C>G p.A176G Chr.3: 186644853-186644853 on Build GRCh38
3591002 c.551C>T p.S184F Chr.3: 186644877-186644877 on Build GRCh38
730099 c.525T>C p.L175L Chr.3: 186644851-186644851 on Build GRCh38
6033485 c.578C>T p.T193I Chr.3: 186644904-186644904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902486278 Chr.3: 186644710-186644710 on Build GRCh38
rs375098849 Chr.3: 186644675-186644675 on Build GRCh38
rs761066967 Chr.3: 186644661-186644661 on Build GRCh38
rs747442158 Chr.3: 186644864-186644866 on Build GRCh38
rs145489603 Chr.3: 186644949-186644949 on Build GRCh38
rs748222875 Chr.3: 186644776-186644776 on Build GRCh38
rs267599724 Chr.3: 186644842-186644842 on Build GRCh38
rs540816433 Chr.3: 186644891-186644891 on Build GRCh38
rs564245691 Chr.3: 186644925-186644925 on Build GRCh38
rs756739630 Chr.3: 186645107-186645107 on Build GRCh38
rs766911612 Chr.3: 186644711-186644711 on Build GRCh38
rs867727735 Chr.3: 186644993-186644993 on Build GRCh38
rs760168975 Chr.3: 186644944-186644944 on Build GRCh38
rs914618708 Chr.3: 186644723-186644723 on Build GRCh38
rs918811126 Chr.3: 186645103-186645103 on Build GRCh38
rs918883210 Chr.3: 186644762-186644762 on Build GRCh38
rs752152102 Chr.3: 186644844-186644844 on Build GRCh38
rs750225373 Chr.3: 186644961-186644961 on Build GRCh38
rs775043391 Chr.3: 186644835-186644835 on Build GRCh38
rs985454635 Chr.3: 186644733-186644733 on Build GRCh38
rs267599723 Chr.3: 186644841-186644841 on Build GRCh38
rs1037279428 Chr.3: 186644761-186644761 on Build GRCh38
rs1031546008 Chr.3: 186644901-186644901 on Build GRCh38
rs774885834 Chr.3: 186644913-186644913 on Build GRCh38
rs193281601 Chr.3: 186645096-186645096 on Build GRCh38
rs760171993 Chr.3: 186644845-186644845 on Build GRCh38
rs772356511 Chr.3: 186644754-186644755 on Build GRCh38
rs780384772 Chr.3: 186644859-186644861 on Build GRCh38
rs76189223 Chr.3: 186644981-186644981 on Build GRCh38
rs753484336 Chr.3: 186644850-186644850 on Build GRCh38
rs375848987 Chr.3: 186644951-186644951 on Build GRCh38
rs895175063 Chr.3: 186645098-186645098 on Build GRCh38
rs766288543 Chr.3: 186644971-186644971 on Build GRCh38
rs148507156 Chr.3: 186644869-186644869 on Build GRCh38
rs758157844 Chr.3: 186644969-186644969 on Build GRCh38
rs757032153 Chr.3: 186644760-186644760 on Build GRCh38
rs935942868 Chr.3: 186645017-186645017 on Build GRCh38
rs985936992 Chr.3: 186644974-186644974 on Build GRCh38
rs766718434 Chr.3: 186644794-186644794 on Build GRCh38
rs771387356 Chr.3: 186644833-186644833 on Build GRCh38
rs79613655 Chr.3: 186644813-186644813 on Build GRCh38
rs948491606 Chr.3: 186644818-186644818 on Build GRCh38
rs199634697 Chr.3: 186644843-186644843 on Build GRCh38
rs79103998 Chr.3: 186644829-186644829 on Build GRCh38
rs745773631 Chr.3: 186644769-186644769 on Build GRCh38
rs200463844 Chr.3: 186644720-186644720 on Build GRCh38
rs541290079 Chr.3: 186644775-186644775 on Build GRCh38
rs778177725 Chr.3: 186644876-186644876 on Build GRCh38
rs188313975 Chr.3: 186644771-186644771 on Build GRCh38
rs778889689 Chr.3: 186644764-186644764 on Build GRCh38
rs769915545 Chr.3: 186644780-186644780 on Build GRCh38
rs891716923 Chr.3: 186644928-186644928 on Build GRCh38
rs976557696 Chr.3: 186644929-186644929 on Build GRCh38
rs375505903 Chr.3: 186644716-186644716 on Build GRCh38
rs764832026 Chr.3: 186644952-186644952 on Build GRCh38
rs773429917 Chr.3: 186644802-186644802 on Build GRCh38
rs561960451 Chr.3: 186645025-186645025 on Build GRCh38
rs777245602 Chr.3: 186644897-186644901 on Build GRCh38
rs763841541 Chr.3: 186644975-186644975 on Build GRCh38
rs954278249 Chr.3: 186644724-186644724 on Build GRCh38
rs895228605 Chr.3: 186644736-186644736 on Build GRCh38
rs912535817 Chr.3: 186644734-186644734 on Build GRCh38
rs763473292 Chr.3: 186644822-186644822 on Build GRCh38
rs777415453 Chr.3: 186644732-186644732 on Build GRCh38
rs78581831 Chr.3: 186644854-186644854 on Build GRCh38
rs748130205 Chr.3: 186644962-186644964 on Build GRCh38
rs373411837 Chr.3: 186644938-186644938 on Build GRCh38
rs149523201 Chr.3: 186644853-186644853 on Build GRCh38
rs541856731 Chr.3: 186644865-186644865 on Build GRCh38
rs1053777177 Chr.3: 186645072-186645072 on Build GRCh38
rs972733704 Chr.3: 186645102-186645102 on Build GRCh38
rs748969702 Chr.3: 186644755-186644755 on Build GRCh38
rs749495217 Chr.3: 186644890-186644890 on Build GRCh38
rs772469145 Chr.3: 186644936-186644936 on Build GRCh38
rs944083157 Chr.3: 186644748-186644748 on Build GRCh38
rs753647178 Chr.3: 186644749-186644749 on Build GRCh38
rs757085308 Chr.3: 186644851-186644851 on Build GRCh38
rs113428750 Chr.3: 186644896-186644896 on Build GRCh38
rs769014961 Chr.3: 186644878-186644879 on Build GRCh38
rs759152959 Chr.3: 186644706-186644706 on Build GRCh38

View Full Product Details