Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 15938950-15939537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL7
Entrez Gene ID
23194
Gene Name
F-box and leucine rich repeat protein 7
Gene Aliases
FBL6, FBL7
UniGene
Hs.433057
Gene Chromosome Location
Chr.5: 15500196-15939796 on Build GRCh38
Transcripts
NM_001278317.1 NM_012304.4 XM_011513998.1 XM_017009262.1

SNP Detail

SNP ID Chromosome Location
rs375443554 Chr.5: 15939263-15939263 on Build GRCh38
rs538906307 Chr.5: 15939425-15939425 on Build GRCh38
rs530958389 Chr.5: 15939258-15939258 on Build GRCh38
rs903896542 Chr.5: 15938990-15938990 on Build GRCh38
rs963457250 Chr.5: 15939176-15939176 on Build GRCh38
rs543266379 Chr.5: 15939063-15939063 on Build GRCh38
rs892811630 Chr.5: 15939314-15939314 on Build GRCh38
rs1043277416 Chr.5: 15939365-15939365 on Build GRCh38
rs781627875 Chr.5: 15939238-15939238 on Build GRCh38
rs909387228 Chr.5: 15939171-15939171 on Build GRCh38
rs759986047 Chr.5: 15939031-15939031 on Build GRCh38
rs561324425 Chr.5: 15939351-15939351 on Build GRCh38
rs149205811 Chr.5: 15939495-15939495 on Build GRCh38
rs910432770 Chr.5: 15939356-15939356 on Build GRCh38
rs1013824457 Chr.5: 15939075-15939075 on Build GRCh38
rs904389836 Chr.5: 15939393-15939393 on Build GRCh38
rs559696451 Chr.5: 15939094-15939094 on Build GRCh38
rs546744635 Chr.5: 15939375-15939375 on Build GRCh38
rs191928894 Chr.5: 15938986-15938986 on Build GRCh38
rs952908789 Chr.5: 15939039-15939039 on Build GRCh38
rs943309136 Chr.5: 15939364-15939364 on Build GRCh38
rs1035562863 Chr.5: 15939415-15939415 on Build GRCh38
rs533651593 Chr.5: 15939141-15939141 on Build GRCh38
rs918913340 Chr.5: 15939270-15939270 on Build GRCh38
rs971823833 Chr.5: 15939217-15939217 on Build GRCh38
rs991486562 Chr.5: 15938978-15938978 on Build GRCh38
rs1031000958 Chr.5: 15939049-15939049 on Build GRCh38
rs531933036 Chr.5: 15939481-15939481 on Build GRCh38
rs571367540 Chr.5: 15939424-15939424 on Build GRCh38
rs763987614 Chr.5: 15939405-15939405 on Build GRCh38
rs574140712 Chr.5: 15939062-15939062 on Build GRCh38
rs947283970 Chr.5: 15939317-15939317 on Build GRCh38
rs958802575 Chr.5: 15938976-15938976 on Build GRCh38
rs545393890 Chr.5: 15939229-15939229 on Build GRCh38
rs892570233 Chr.5: 15939426-15939426 on Build GRCh38
rs933011626 Chr.5: 15939280-15939280 on Build GRCh38
rs1025205839 Chr.5: 15939076-15939076 on Build GRCh38
rs965680142 Chr.5: 15939491-15939492 on Build GRCh38
rs931771108 Chr.5: 15939213-15939213 on Build GRCh38
rs985842796 Chr.5: 15939497-15939497 on Build GRCh38
rs1000995995 Chr.5: 15939048-15939048 on Build GRCh38
rs563657942 Chr.5: 15939233-15939233 on Build GRCh38
rs549496663 Chr.5: 15939319-15939319 on Build GRCh38
rs983026662 Chr.5: 15939440-15939440 on Build GRCh38
rs73752391 Chr.5: 15939449-15939449 on Build GRCh38
rs914213208 Chr.5: 15939500-15939500 on Build GRCh38
rs756056779 Chr.5: 15939119-15939119 on Build GRCh38
rs905406149 Chr.5: 15939494-15939494 on Build GRCh38
rs894558553 Chr.5: 15939418-15939418 on Build GRCh38
rs183026159 Chr.5: 15939352-15939352 on Build GRCh38
rs1027024031 Chr.5: 15939011-15939011 on Build GRCh38
rs936725878 Chr.5: 15939402-15939402 on Build GRCh38
rs752704387 Chr.5: 15939116-15939116 on Build GRCh38
rs1042042828 Chr.5: 15939331-15939331 on Build GRCh38
rs952842248 Chr.5: 15939439-15939439 on Build GRCh38
rs1050042077 Chr.5: 15939221-15939221 on Build GRCh38
rs1024981562 Chr.5: 15939492-15939492 on Build GRCh38

View Full Product Details