Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10226412-10226860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM173B
Entrez Gene ID
134145
Gene Name
family with sequence similarity 173 member B
Gene Aliases
JS-2
UniGene
Hs.481569
Gene Chromosome Location
Chr.5: 10225507-10249909 on Build GRCh38
Transcripts
NM_001258388.1 NM_001258389.1 NM_199133.3 NR_047668.1 NR_047669.1 NR_047670.1 XM_011513964.1

SNP Detail

SNP ID Chromosome Location
rs1004777741 Chr.5: 10226526-10226526 on Build GRCh38
rs936663104 Chr.5: 10226459-10226459 on Build GRCh38
rs775854222 Chr.5: 10226515-10226515 on Build GRCh38
rs112112971 Chr.5: 10226575-10226575 on Build GRCh38
rs550066464 Chr.5: 10226513-10226513 on Build GRCh38
rs547740485 Chr.5: 10226690-10226690 on Build GRCh38
rs138090861 Chr.5: 10226724-10226724 on Build GRCh38
rs993001469 Chr.5: 10226593-10226593 on Build GRCh38
rs1038972436 Chr.5: 10226705-10226705 on Build GRCh38
rs914490146 Chr.5: 10226706-10226706 on Build GRCh38
rs191579354 Chr.5: 10226788-10226788 on Build GRCh38
rs936963946 Chr.5: 10226704-10226704 on Build GRCh38
rs764751047 Chr.5: 10226779-10226780 on Build GRCh38
rs897341634 Chr.5: 10226440-10226440 on Build GRCh38
rs183828896 Chr.5: 10226830-10226830 on Build GRCh38
rs558932773 Chr.5: 10226752-10226752 on Build GRCh38
rs35523327 Chr.5: 10226757-10226758 on Build GRCh38
rs925530815 Chr.5: 10226680-10226680 on Build GRCh38
rs978259724 Chr.5: 10226647-10226647 on Build GRCh38
rs967263594 Chr.5: 10226646-10226646 on Build GRCh38
rs1039044283 Chr.5: 10226489-10226489 on Build GRCh38
rs1037570920 Chr.5: 10226814-10226814 on Build GRCh38
rs529977803 Chr.5: 10226455-10226455 on Build GRCh38
rs1045369 Chr.5: 10226727-10226727 on Build GRCh38
rs113872726 Chr.5: 10226736-10226736 on Build GRCh38
rs576698287 Chr.5: 10226732-10226732 on Build GRCh38
rs556793295 Chr.5: 10226729-10226729 on Build GRCh38
rs541548900 Chr.5: 10226794-10226794 on Build GRCh38
rs1034021275 Chr.5: 10226559-10226559 on Build GRCh38
rs150369651 Chr.5: 10226537-10226537 on Build GRCh38
rs888405821 Chr.5: 10226473-10226473 on Build GRCh38
rs184891344 Chr.5: 10226663-10226663 on Build GRCh38
rs900592177 Chr.5: 10226494-10226494 on Build GRCh38
rs140349465 Chr.5: 10226454-10226454 on Build GRCh38
rs1012481241 Chr.5: 10226813-10226813 on Build GRCh38
rs189440139 Chr.5: 10226776-10226776 on Build GRCh38
rs1035907655 Chr.5: 10226577-10226577 on Build GRCh38
rs947320966 Chr.5: 10226708-10226708 on Build GRCh38
rs534413009 Chr.5: 10226655-10226655 on Build GRCh38
rs563312246 Chr.5: 10226514-10226514 on Build GRCh38
rs1016168805 Chr.5: 10226534-10226534 on Build GRCh38
rs752086168 Chr.5: 10226781-10226781 on Build GRCh38
rs565431946 Chr.5: 10226632-10226632 on Build GRCh38
rs963316582 Chr.5: 10226536-10226536 on Build GRCh38
rs1048581158 Chr.5: 10226784-10226784 on Build GRCh38
rs888688110 Chr.5: 10226798-10226798 on Build GRCh38

View Full Product Details