Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33319595-33320181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAXX
Entrez Gene ID
1616
Gene Name
death domain associated protein
Gene Aliases
BING2, DAP6, EAP1
UniGene
Hs.336916
Gene Chromosome Location
Chr.6: 33318558-33323016 on Build GRCh38
Transcripts
NM_001141969.1 NM_001141970.1 NM_001254717.1 NM_001350.4 XM_005248860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5357385 c.1486C>A p.P496T Chr.6: 33319834-33319834 on Build GRCh38
6503464 c.1665_1666insG p.T556fs*13 Chr.6: 33319654-33319655 on Build GRCh38
6945623 c.1362_1364delAGA p.E457delE Chr.6: 33320112-33320114 on Build GRCh38
6921331 c.1595T>A p.L532* Chr.6: 33319725-33319725 on Build GRCh38
7199027 c.1530T>C p.P510P Chr.6: 33319790-33319790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755501402 Chr.6: 33320111-33320111 on Build GRCh38
rs773386948 Chr.6: 33319710-33319710 on Build GRCh38
rs770543662 Chr.6: 33320138-33320138 on Build GRCh38
rs764132702 Chr.6: 33319968-33319968 on Build GRCh38
rs778324215 Chr.6: 33320022-33320022 on Build GRCh38
rs960795168 Chr.6: 33319888-33319888 on Build GRCh38
rs777217124 Chr.6: 33320135-33320135 on Build GRCh38
rs761962503 Chr.6: 33320065-33320065 on Build GRCh38
rs764801966 Chr.6: 33319728-33319733 on Build GRCh38
rs747188410 Chr.6: 33319682-33319682 on Build GRCh38
rs200416162 Chr.6: 33319995-33319995 on Build GRCh38
rs759473289 Chr.6: 33319962-33319962 on Build GRCh38
rs752770741 Chr.6: 33319648-33319648 on Build GRCh38
rs751521776 Chr.6: 33320011-33320013 on Build GRCh38
rs770031980 Chr.6: 33320079-33320081 on Build GRCh38
rs755745890 Chr.6: 33320150-33320152 on Build GRCh38
rs145347312 Chr.6: 33320045-33320045 on Build GRCh38
rs61748592 Chr.6: 33319727-33319727 on Build GRCh38
rs753718788 Chr.6: 33319793-33319793 on Build GRCh38
rs771514387 Chr.6: 33320028-33320028 on Build GRCh38
rs150463674 Chr.6: 33320032-33320032 on Build GRCh38
rs746429898 Chr.6: 33319691-33319691 on Build GRCh38
rs755821596 Chr.6: 33319808-33319808 on Build GRCh38
rs769768112 Chr.6: 33320053-33320053 on Build GRCh38
rs780938451 Chr.6: 33320050-33320058 on Build GRCh38
rs777757425 Chr.6: 33319974-33319975 on Build GRCh38
rs774350850 Chr.6: 33320063-33320063 on Build GRCh38
rs771722185 Chr.6: 33319883-33319883 on Build GRCh38
rs749346133 Chr.6: 33320112-33320114 on Build GRCh38
rs753080247 Chr.6: 33320129-33320129 on Build GRCh38
rs377489315 Chr.6: 33320001-33320001 on Build GRCh38
rs756739133 Chr.6: 33319841-33319841 on Build GRCh38
rs929024196 Chr.6: 33320048-33320048 on Build GRCh38
rs750612170 Chr.6: 33319991-33319991 on Build GRCh38
rs141395832 Chr.6: 33319743-33319743 on Build GRCh38
rs766097275 Chr.6: 33319889-33319889 on Build GRCh38
rs771235091 Chr.6: 33319686-33319686 on Build GRCh38
rs766292072 Chr.6: 33319792-33319792 on Build GRCh38
rs779489722 Chr.6: 33320125-33320125 on Build GRCh38
rs778573009 Chr.6: 33319785-33319785 on Build GRCh38
rs757490566 Chr.6: 33319988-33319988 on Build GRCh38
rs190677625 Chr.6: 33319941-33319941 on Build GRCh38
rs767634147 Chr.6: 33320075-33320075 on Build GRCh38
rs760674231 Chr.6: 33319788-33319788 on Build GRCh38
rs780917649 Chr.6: 33320145-33320145 on Build GRCh38
rs201295312 Chr.6: 33319969-33319969 on Build GRCh38
rs1010089014 Chr.6: 33319899-33319899 on Build GRCh38
rs755994636 Chr.6: 33319631-33319631 on Build GRCh38
rs372876513 Chr.6: 33319750-33319750 on Build GRCh38
rs746717061 Chr.6: 33320137-33320137 on Build GRCh38
rs200567881 Chr.6: 33320104-33320106 on Build GRCh38
rs990280461 Chr.6: 33319984-33319984 on Build GRCh38
rs770739609 Chr.6: 33320036-33320036 on Build GRCh38
rs146868243 Chr.6: 33319809-33319809 on Build GRCh38
rs759434939 Chr.6: 33320051-33320051 on Build GRCh38
rs761486064 Chr.6: 33319755-33319755 on Build GRCh38
rs572793805 Chr.6: 33319956-33319956 on Build GRCh38
rs773862800 Chr.6: 33319871-33319871 on Build GRCh38
rs780522375 Chr.6: 33319998-33319998 on Build GRCh38
rs757482758 Chr.6: 33319823-33319823 on Build GRCh38
rs562744463 Chr.6: 33319923-33319923 on Build GRCh38
rs774915926 Chr.6: 33320121-33320126 on Build GRCh38
rs772433267 Chr.6: 33320133-33320138 on Build GRCh38
rs747802784 Chr.6: 33319874-33319874 on Build GRCh38
rs772967665 Chr.6: 33319770-33319770 on Build GRCh38
rs781720244 Chr.6: 33319830-33319830 on Build GRCh38
rs758413375 Chr.6: 33319802-33319802 on Build GRCh38
rs752227208 Chr.6: 33319871-33319871 on Build GRCh38
rs750701787 Chr.6: 33320087-33320087 on Build GRCh38
rs780716471 Chr.6: 33319863-33319863 on Build GRCh38
rs757006416 Chr.6: 33320020-33320022 on Build GRCh38
rs768082927 Chr.6: 33319868-33319868 on Build GRCh38
rs146304558 Chr.6: 33320019-33320019 on Build GRCh38
rs545922224 Chr.6: 33320141-33320141 on Build GRCh38
rs776581247 Chr.6: 33319894-33319894 on Build GRCh38
rs370615019 Chr.6: 33320033-33320033 on Build GRCh38
rs754198630 Chr.6: 33320098-33320098 on Build GRCh38
rs944317260 Chr.6: 33319637-33319637 on Build GRCh38
rs557099139 Chr.6: 33319893-33319893 on Build GRCh38
rs773879056 Chr.6: 33319744-33319744 on Build GRCh38
rs561526683 Chr.6: 33319990-33319990 on Build GRCh38
rs751755249 Chr.6: 33319821-33319821 on Build GRCh38
rs151039809 Chr.6: 33319663-33319663 on Build GRCh38
rs760369795 Chr.6: 33320123-33320123 on Build GRCh38
rs775204775 Chr.6: 33319728-33319728 on Build GRCh38
rs555992054 Chr.6: 33319689-33319689 on Build GRCh38
rs748790861 Chr.6: 33319735-33319735 on Build GRCh38
rs758706597 Chr.6: 33319659-33319659 on Build GRCh38
rs74986527 Chr.6: 33320042-33320042 on Build GRCh38
rs779012166 Chr.6: 33319643-33319643 on Build GRCh38
rs758034267 Chr.6: 33319900-33319900 on Build GRCh38
rs557828859 Chr.6: 33320154-33320156 on Build GRCh38
rs556533833 Chr.6: 33319838-33319838 on Build GRCh38
rs535673316 Chr.6: 33319692-33319692 on Build GRCh38
rs757935715 Chr.6: 33320131-33320131 on Build GRCh38
rs551346113 Chr.6: 33320092-33320092 on Build GRCh38
rs746227699 Chr.6: 33319835-33319835 on Build GRCh38
rs780858272 Chr.6: 33320040-33320042 on Build GRCh38
rs762240849 Chr.6: 33320120-33320121 on Build GRCh38
rs767236551 Chr.6: 33319763-33319763 on Build GRCh38
rs910218439 Chr.6: 33319626-33319626 on Build GRCh38
rs768675869 Chr.6: 33320121-33320123 on Build GRCh38
rs115501111 Chr.6: 33319886-33319886 on Build GRCh38
rs577927719 Chr.6: 33319866-33319866 on Build GRCh38

View Full Product Details