Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36685527-36686102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN1A
Entrez Gene ID
1026
Gene Name
cyclin dependent kinase inhibitor 1A
Gene Aliases
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
UniGene
Hs.370771
Gene Chromosome Location
Chr.6: 36676460-36687339 on Build GRCh38
Transcripts
NM_000389.4 NM_001220777.1 NM_001220778.1 NM_001291549.1 NM_078467.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3076247 c.466C>T p.R156W Chr.6: 36685771-36685771 on Build GRCh38
6625706 c.565C>T p.R189C Chr.6: 36685768-36685768 on Build GRCh38
6946557 c.474C>G p.I158M Chr.6: 36685779-36685779 on Build GRCh38
6106750 c.588G>T p.R196S Chr.6: 36685791-36685791 on Build GRCh38
3626274 c.576C>T p.I192I Chr.6: 36685779-36685779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763632708 Chr.6: 36685917-36685917 on Build GRCh38
rs1021888639 Chr.6: 36685637-36685637 on Build GRCh38
rs750249677 Chr.6: 36685768-36685768 on Build GRCh38
rs114982296 Chr.6: 36686057-36686057 on Build GRCh38
rs575815855 Chr.6: 36685852-36685852 on Build GRCh38
rs748554253 Chr.6: 36685605-36685605 on Build GRCh38
rs781057608 Chr.6: 36685767-36685767 on Build GRCh38
rs755706954 Chr.6: 36686008-36686008 on Build GRCh38
rs998171288 Chr.6: 36685855-36685855 on Build GRCh38
rs905530808 Chr.6: 36685600-36685600 on Build GRCh38
rs150869821 Chr.6: 36685835-36685835 on Build GRCh38
rs3176356 Chr.6: 36685700-36685700 on Build GRCh38
rs951389423 Chr.6: 36685916-36685916 on Build GRCh38
rs191431994 Chr.6: 36685995-36685995 on Build GRCh38
rs201408395 Chr.6: 36685831-36685831 on Build GRCh38
rs976707693 Chr.6: 36685905-36685910 on Build GRCh38
rs762424928 Chr.6: 36685736-36685736 on Build GRCh38
rs41272170 Chr.6: 36685833-36685833 on Build GRCh38
rs375173291 Chr.6: 36685832-36685832 on Build GRCh38
rs965385710 Chr.6: 36685687-36685687 on Build GRCh38
rs750860362 Chr.6: 36685710-36685712 on Build GRCh38
rs764981917 Chr.6: 36685712-36685712 on Build GRCh38
rs1010097881 Chr.6: 36685636-36685636 on Build GRCh38
rs1034635876 Chr.6: 36685669-36685669 on Build GRCh38
rs564494785 Chr.6: 36685942-36685942 on Build GRCh38
rs762632530 Chr.6: 36685828-36685828 on Build GRCh38
rs779826646 Chr.6: 36685932-36685932 on Build GRCh38
rs778242620 Chr.6: 36686059-36686059 on Build GRCh38
rs529170426 Chr.6: 36686007-36686007 on Build GRCh38
rs914304254 Chr.6: 36685937-36685937 on Build GRCh38
rs1056094900 Chr.6: 36686021-36686021 on Build GRCh38
rs760541872 Chr.6: 36685874-36685874 on Build GRCh38
rs113000891 Chr.6: 36685704-36685704 on Build GRCh38
rs41272172 Chr.6: 36686083-36686083 on Build GRCh38
rs554325867 Chr.6: 36685613-36685613 on Build GRCh38
rs1041396487 Chr.6: 36685944-36685944 on Build GRCh38
rs139304346 Chr.6: 36685803-36685803 on Build GRCh38
rs751490716 Chr.6: 36685752-36685752 on Build GRCh38
rs907923458 Chr.6: 36685868-36685868 on Build GRCh38
rs758444092 Chr.6: 36685638-36685638 on Build GRCh38
rs374049839 Chr.6: 36685701-36685701 on Build GRCh38
rs981464191 Chr.6: 36685784-36685784 on Build GRCh38
rs754443671 Chr.6: 36685773-36685773 on Build GRCh38
rs111926164 Chr.6: 36685851-36685851 on Build GRCh38
rs370565004 Chr.6: 36685706-36685706 on Build GRCh38
rs960959039 Chr.6: 36686025-36686025 on Build GRCh38
rs112675295 Chr.6: 36685943-36685943 on Build GRCh38
rs779063779 Chr.6: 36685772-36685772 on Build GRCh38
rs984194375 Chr.6: 36685705-36685705 on Build GRCh38
rs952159534 Chr.6: 36685858-36685858 on Build GRCh38
rs772087100 Chr.6: 36685779-36685779 on Build GRCh38
rs747014318 Chr.6: 36685804-36685804 on Build GRCh38
rs376862038 Chr.6: 36685546-36685546 on Build GRCh38
rs3176357 Chr.6: 36685838-36685838 on Build GRCh38
rs922225207 Chr.6: 36685935-36685935 on Build GRCh38
rs376945964 Chr.6: 36685703-36685703 on Build GRCh38
rs187742871 Chr.6: 36685936-36685936 on Build GRCh38
rs1028124783 Chr.6: 36685856-36685856 on Build GRCh38
rs763653680 Chr.6: 36685748-36685748 on Build GRCh38
rs775069456 Chr.6: 36685822-36685822 on Build GRCh38
rs546844573 Chr.6: 36685590-36685590 on Build GRCh38
rs752332685 Chr.6: 36685722-36685722 on Build GRCh38
rs910165861 Chr.6: 36685559-36685559 on Build GRCh38
rs770267002 Chr.6: 36685821-36685821 on Build GRCh38
rs945802868 Chr.6: 36685823-36685823 on Build GRCh38
rs537025349 Chr.6: 36685770-36685770 on Build GRCh38
rs1007220462 Chr.6: 36685672-36685672 on Build GRCh38
rs756718995 Chr.6: 36685994-36685994 on Build GRCh38
rs199801759 Chr.6: 36685757-36685757 on Build GRCh38
rs866686724 Chr.6: 36685957-36685957 on Build GRCh38
rs199543282 Chr.6: 36685764-36685764 on Build GRCh38
rs368048005 Chr.6: 36685792-36685792 on Build GRCh38
rs546659289 Chr.6: 36685981-36685981 on Build GRCh38
rs754567385 Chr.6: 36685840-36685840 on Build GRCh38
rs983400213 Chr.6: 36685866-36685866 on Build GRCh38
rs535671586 Chr.6: 36685683-36685683 on Build GRCh38
rs1056940516 Chr.6: 36685612-36685612 on Build GRCh38
rs1059234 Chr.6: 36685820-36685820 on Build GRCh38
rs755968076 Chr.6: 36685843-36685843 on Build GRCh38
rs781689614 Chr.6: 36686067-36686067 on Build GRCh38
rs374154006 Chr.6: 36685771-36685771 on Build GRCh38
rs568174818 Chr.6: 36685642-36685642 on Build GRCh38
rs777207887 Chr.6: 36685811-36685811 on Build GRCh38
rs948319635 Chr.6: 36685566-36685566 on Build GRCh38
rs1045297391 Chr.6: 36685585-36685585 on Build GRCh38
rs771172537 Chr.6: 36685739-36685739 on Build GRCh38
rs543585485 Chr.6: 36686048-36686048 on Build GRCh38

View Full Product Details