Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36354581-36355161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV7
Entrez Gene ID
51513
Gene Name
ETS variant 7
Gene Aliases
TEL-2, TEL2, TELB
UniGene
Hs.272398
Gene Chromosome Location
Chr.6: 36354221-36387800 on Build GRCh38
Transcripts
NM_001207035.1 NM_001207039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5717316 c.913G>A p.G305R Chr.6: 36354683-36354683 on Build GRCh38
5548000 c.909-8delT p.? Chr.6: 36354695-36354695 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182436821 Chr.6: 36354737-36354737 on Build GRCh38
rs142291137 Chr.6: 36354771-36354773 on Build GRCh38
rs935156331 Chr.6: 36354707-36354707 on Build GRCh38
rs535902890 Chr.6: 36355011-36355011 on Build GRCh38
rs575407760 Chr.6: 36354935-36354935 on Build GRCh38
rs765578011 Chr.6: 36355065-36355065 on Build GRCh38
rs1007984174 Chr.6: 36354854-36354854 on Build GRCh38
rs765080878 Chr.6: 36354629-36354629 on Build GRCh38
rs1017583081 Chr.6: 36354857-36354857 on Build GRCh38
rs949136596 Chr.6: 36354647-36354647 on Build GRCh38
rs1005219347 Chr.6: 36354786-36354786 on Build GRCh38
rs768836368 Chr.6: 36354695-36354696 on Build GRCh38
rs916207451 Chr.6: 36354950-36354950 on Build GRCh38
rs1046162516 Chr.6: 36354670-36354670 on Build GRCh38
rs1029557545 Chr.6: 36354841-36354841 on Build GRCh38
rs575696065 Chr.6: 36355073-36355073 on Build GRCh38
rs893721942 Chr.6: 36354758-36354758 on Build GRCh38
rs750092791 Chr.6: 36354850-36354850 on Build GRCh38
rs992979467 Chr.6: 36354985-36354985 on Build GRCh38
rs996124786 Chr.6: 36354791-36354791 on Build GRCh38
rs991777686 Chr.6: 36354944-36354944 on Build GRCh38
rs184576677 Chr.6: 36355030-36355030 on Build GRCh38
rs188359850 Chr.6: 36354814-36354814 on Build GRCh38
rs552578370 Chr.6: 36354796-36354796 on Build GRCh38
rs563469381 Chr.6: 36354695-36354695 on Build GRCh38
rs539871057 Chr.6: 36355078-36355078 on Build GRCh38
rs909515192 Chr.6: 36355033-36355033 on Build GRCh38
rs75688032 Chr.6: 36354695-36354695 on Build GRCh38
rs915102198 Chr.6: 36355125-36355125 on Build GRCh38
rs1009369760 Chr.6: 36354851-36354851 on Build GRCh38
rs371256151 Chr.6: 36354973-36354973 on Build GRCh38
rs990436152 Chr.6: 36354608-36354608 on Build GRCh38
rs192547126 Chr.6: 36354999-36354999 on Build GRCh38
rs762281517 Chr.6: 36354954-36354954 on Build GRCh38
rs962145531 Chr.6: 36354866-36354866 on Build GRCh38
rs994962875 Chr.6: 36354921-36354921 on Build GRCh38
rs749158016 Chr.6: 36354694-36354695 on Build GRCh38
rs528915550 Chr.6: 36354693-36354693 on Build GRCh38
rs567760496 Chr.6: 36354963-36354963 on Build GRCh38
rs888210686 Chr.6: 36354783-36354783 on Build GRCh38
rs758164782 Chr.6: 36354720-36354720 on Build GRCh38
rs770600739 Chr.6: 36354671-36354671 on Build GRCh38
rs916275096 Chr.6: 36354609-36354609 on Build GRCh38
rs532804176 Chr.6: 36354757-36354757 on Build GRCh38
rs970428366 Chr.6: 36354962-36354962 on Build GRCh38
rs1036779950 Chr.6: 36355072-36355072 on Build GRCh38
rs534090558 Chr.6: 36354773-36354775 on Build GRCh38
rs1044456425 Chr.6: 36355024-36355024 on Build GRCh38
rs935104869 Chr.6: 36355075-36355075 on Build GRCh38
rs982231452 Chr.6: 36355031-36355031 on Build GRCh38
rs917506793 Chr.6: 36354633-36354633 on Build GRCh38
rs58528208 Chr.6: 36354924-36354924 on Build GRCh38
rs550266142 Chr.6: 36354724-36354724 on Build GRCh38
rs561750263 Chr.6: 36354632-36354632 on Build GRCh38
rs752630162 Chr.6: 36355083-36355084 on Build GRCh38
rs745359191 Chr.6: 36354698-36354698 on Build GRCh38
rs547134004 Chr.6: 36354995-36354995 on Build GRCh38
rs1015016293 Chr.6: 36354852-36354852 on Build GRCh38
rs1053661719 Chr.6: 36354738-36354738 on Build GRCh38

View Full Product Details