Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53267237-53267825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL5
Entrez Gene ID
60481
Gene Name
ELOVL fatty acid elongase 5
Gene Aliases
HELO1, SCA38, dJ483K16.1
UniGene
Hs.520189
Gene Chromosome Location
Chr.6: 53267398-53349179 on Build GRCh38
Transcripts
NM_001242828.1 NM_001242830.1 NM_001301856.1 NM_021814.4

SNP Detail

SNP ID Chromosome Location
rs748892429 Chr.6: 53267795-53267795 on Build GRCh38
rs966487620 Chr.6: 53267767-53267767 on Build GRCh38
rs554236292 Chr.6: 53267568-53267568 on Build GRCh38
rs963800297 Chr.6: 53267582-53267582 on Build GRCh38
rs928813793 Chr.6: 53267301-53267301 on Build GRCh38
rs576587221 Chr.6: 53267680-53267680 on Build GRCh38
rs531555851 Chr.6: 53267264-53267264 on Build GRCh38
rs916025475 Chr.6: 53267394-53267394 on Build GRCh38
rs3207469 Chr.6: 53267515-53267515 on Build GRCh38
rs556433052 Chr.6: 53267679-53267679 on Build GRCh38
rs947362364 Chr.6: 53267419-53267419 on Build GRCh38
rs995133186 Chr.6: 53267602-53267602 on Build GRCh38
rs951063334 Chr.6: 53267670-53267670 on Build GRCh38
rs1019023620 Chr.6: 53267700-53267700 on Build GRCh38
rs748109188 Chr.6: 53267569-53267569 on Build GRCh38
rs73433434 Chr.6: 53267492-53267492 on Build GRCh38
rs570188014 Chr.6: 53267378-53267378 on Build GRCh38
rs1000373467 Chr.6: 53267345-53267346 on Build GRCh38
rs1023215340 Chr.6: 53267268-53267268 on Build GRCh38
rs2273012 Chr.6: 53267677-53267677 on Build GRCh38
rs781069526 Chr.6: 53267540-53267540 on Build GRCh38
rs983322774 Chr.6: 53267297-53267297 on Build GRCh38
rs534182403 Chr.6: 53267561-53267561 on Build GRCh38
rs992080452 Chr.6: 53267736-53267736 on Build GRCh38
rs896175299 Chr.6: 53267615-53267615 on Build GRCh38
rs535343655 Chr.6: 53267482-53267482 on Build GRCh38
rs532901678 Chr.6: 53267384-53267384 on Build GRCh38
rs938889928 Chr.6: 53267332-53267332 on Build GRCh38
rs183091687 Chr.6: 53267454-53267454 on Build GRCh38
rs769273870 Chr.6: 53267663-53267663 on Build GRCh38
rs890958756 Chr.6: 53267510-53267510 on Build GRCh38
rs754774914 Chr.6: 53267524-53267524 on Build GRCh38
rs988891998 Chr.6: 53267405-53267405 on Build GRCh38
rs906074303 Chr.6: 53267287-53267287 on Build GRCh38
rs139025292 Chr.6: 53267532-53267532 on Build GRCh38
rs74843119 Chr.6: 53267513-53267513 on Build GRCh38
rs577377753 Chr.6: 53267614-53267614 on Build GRCh38
rs146650314 Chr.6: 53267356-53267356 on Build GRCh38
rs190581187 Chr.6: 53267447-53267447 on Build GRCh38
rs1026660770 Chr.6: 53267639-53267639 on Build GRCh38
rs956126215 Chr.6: 53267398-53267398 on Build GRCh38
rs975107502 Chr.6: 53267458-53267458 on Build GRCh38
rs1052815495 Chr.6: 53267494-53267494 on Build GRCh38
rs903740365 Chr.6: 53267489-53267489 on Build GRCh38

View Full Product Details