Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108857905-108858331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
UniGene
Hs.645481
Gene Chromosome Location
Chr.6: 108848410-108990738 on Build GRCh38
Transcripts
NM_001286609.1 NM_032131.5 XM_005267154.3 XM_005267155.2 XM_005267157.3 XM_006715573.2 XM_006715574.2 XM_011536166.1 XM_011536167.1 XM_011536168.2 XM_011536170.2 XM_011536172.2 XM_011536176.2 XM_017011350.1 XM_017011351.1 XM_017011352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1311444 c.286G>C p.E96Q Chr.6: 108858266-108858266 on Build GRCh38
5526958 c.230C>T p.S77F Chr.6: 108858210-108858210 on Build GRCh38
1311443 c.247G>A p.D83N Chr.6: 108858227-108858227 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766709979 Chr.6: 108858263-108858263 on Build GRCh38
rs763385882 Chr.6: 108858259-108858259 on Build GRCh38
rs182708795 Chr.6: 108857939-108857939 on Build GRCh38
rs77521665 Chr.6: 108858022-108858022 on Build GRCh38
rs777242036 Chr.6: 108858180-108858180 on Build GRCh38
rs140140644 Chr.6: 108858200-108858200 on Build GRCh38
rs755879511 Chr.6: 108858156-108858156 on Build GRCh38
rs116681118 Chr.6: 108858195-108858195 on Build GRCh38
rs893412106 Chr.6: 108858291-108858291 on Build GRCh38
rs752221246 Chr.6: 108858155-108858155 on Build GRCh38
rs745873707 Chr.6: 108858205-108858205 on Build GRCh38
rs201510425 Chr.6: 108858221-108858221 on Build GRCh38
rs115448807 Chr.6: 108858030-108858030 on Build GRCh38
rs748180938 Chr.6: 108858239-108858239 on Build GRCh38
rs774849207 Chr.6: 108858268-108858268 on Build GRCh38
rs976075285 Chr.6: 108858027-108858027 on Build GRCh38
rs781052314 Chr.6: 108858103-108858103 on Build GRCh38
rs941219543 Chr.6: 108858124-108858124 on Build GRCh38
rs145671233 Chr.6: 108858226-108858226 on Build GRCh38
rs1008422107 Chr.6: 108858071-108858071 on Build GRCh38
rs768113010 Chr.6: 108858295-108858295 on Build GRCh38
rs552738573 Chr.6: 108858098-108858098 on Build GRCh38
rs771178912 Chr.6: 108858251-108858251 on Build GRCh38
rs961394250 Chr.6: 108858084-108858084 on Build GRCh38
rs946077311 Chr.6: 108858220-108858220 on Build GRCh38
rs994298052 Chr.6: 108858087-108858087 on Build GRCh38
rs749003215 Chr.6: 108858288-108858289 on Build GRCh38
rs752278213 Chr.6: 108858308-108858308 on Build GRCh38
rs528390007 Chr.6: 108858076-108858076 on Build GRCh38
rs960057466 Chr.6: 108858133-108858133 on Build GRCh38
rs545068127 Chr.6: 108858235-108858235 on Build GRCh38
rs756944003 Chr.6: 108858199-108858199 on Build GRCh38
rs9374052 Chr.6: 108858074-108858074 on Build GRCh38
rs1049460762 Chr.6: 108858033-108858033 on Build GRCh38
rs367971410 Chr.6: 108858236-108858236 on Build GRCh38
rs980320335 Chr.6: 108858112-108858113 on Build GRCh38
rs889439052 Chr.6: 108858046-108858046 on Build GRCh38
rs760159922 Chr.6: 108858277-108858277 on Build GRCh38
rs147755566 Chr.6: 108858252-108858252 on Build GRCh38
rs1042223426 Chr.6: 108857970-108857970 on Build GRCh38
rs927311358 Chr.6: 108858116-108858116 on Build GRCh38
rs966129966 Chr.6: 108857961-108857961 on Build GRCh38
rs55695435 Chr.6: 108858148-108858148 on Build GRCh38
rs781690132 Chr.6: 108858258-108858259 on Build GRCh38

View Full Product Details