Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106316016-106316567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG5
Entrez Gene ID
9474
Gene Name
autophagy related 5
Gene Aliases
APG5, APG5-LIKE, APG5L, ASP, hAPG5
UniGene
Hs.486063
Gene Chromosome Location
Chr.6: 106184476-106325820 on Build GRCh38
Transcripts
NM_001286106.1 NM_001286107.1 NM_001286108.1 NM_001286111.1 NM_004849.3 NR_104402.1 NR_104403.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1132070 c.43C>T p.R15* Chr.6: 106316166-106316166 on Build GRCh38
5818600 c.19G>T p.V7L Chr.6: 106316190-106316190 on Build GRCh38
6302604 c.3G>A p.M1I Chr.6: 106316206-106316206 on Build GRCh38
5967897 c.62C>T p.T21M Chr.6: 106316147-106316147 on Build GRCh38
2152807 c.25C>T p.R9* Chr.6: 106316184-106316184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047185301 Chr.6: 106316302-106316302 on Build GRCh38
rs769022755 Chr.6: 106316158-106316158 on Build GRCh38
rs1024434672 Chr.6: 106316460-106316460 on Build GRCh38
rs762640445 Chr.6: 106316404-106316404 on Build GRCh38
rs762501872 Chr.6: 106316229-106316229 on Build GRCh38
rs376583384 Chr.6: 106316490-106316490 on Build GRCh38
rs762253344 Chr.6: 106316061-106316061 on Build GRCh38
rs1040236655 Chr.6: 106316244-106316244 on Build GRCh38
rs761286247 Chr.6: 106316101-106316101 on Build GRCh38
rs376493675 Chr.6: 106316062-106316062 on Build GRCh38
rs2787553 Chr.6: 106316456-106316456 on Build GRCh38
rs758661105 Chr.6: 106316146-106316146 on Build GRCh38
rs765899258 Chr.6: 106316235-106316235 on Build GRCh38
rs762272747 Chr.6: 106316374-106316374 on Build GRCh38
rs772682612 Chr.6: 106316220-106316220 on Build GRCh38
rs772658835 Chr.6: 106316067-106316067 on Build GRCh38
rs770274468 Chr.6: 106316201-106316201 on Build GRCh38
rs930046384 Chr.6: 106316256-106316256 on Build GRCh38
rs747253760 Chr.6: 106316157-106316157 on Build GRCh38
rs973267606 Chr.6: 106316190-106316190 on Build GRCh38
rs188559112 Chr.6: 106316523-106316523 on Build GRCh38
rs900599839 Chr.6: 106316361-106316361 on Build GRCh38
rs569890100 Chr.6: 106316421-106316421 on Build GRCh38
rs779035963 Chr.6: 106316129-106316129 on Build GRCh38
rs767248771 Chr.6: 106316107-106316107 on Build GRCh38
rs80324027 Chr.6: 106316386-106316386 on Build GRCh38
rs145438411 Chr.6: 106316528-106316528 on Build GRCh38
rs370977998 Chr.6: 106316069-106316069 on Build GRCh38
rs140026982 Chr.6: 106316245-106316245 on Build GRCh38
rs181872038 Chr.6: 106316541-106316541 on Build GRCh38
rs923898492 Chr.6: 106316411-106316411 on Build GRCh38
rs777144954 Chr.6: 106316052-106316052 on Build GRCh38
rs533119895 Chr.6: 106316371-106316371 on Build GRCh38
rs776216313 Chr.6: 106316058-106316058 on Build GRCh38
rs75345161 Chr.6: 106316387-106316387 on Build GRCh38
rs543752850 Chr.6: 106316491-106316491 on Build GRCh38
rs944098477 Chr.6: 106316339-106316339 on Build GRCh38
rs370558728 Chr.6: 106316375-106316375 on Build GRCh38
rs766062613 Chr.6: 106316087-106316087 on Build GRCh38
rs756019944 Chr.6: 106316127-106316127 on Build GRCh38
rs555447306 Chr.6: 106316485-106316485 on Build GRCh38
rs180896836 Chr.6: 106316191-106316191 on Build GRCh38
rs185306016 Chr.6: 106316336-106316336 on Build GRCh38
rs751336627 Chr.6: 106316093-106316093 on Build GRCh38
rs373794853 Chr.6: 106316165-106316165 on Build GRCh38
rs911755956 Chr.6: 106316317-106316317 on Build GRCh38
rs374757627 Chr.6: 106316374-106316375 on Build GRCh38
rs752612830 Chr.6: 106316116-106316116 on Build GRCh38
rs558341820 Chr.6: 106316417-106316419 on Build GRCh38
rs752650486 Chr.6: 106316524-106316525 on Build GRCh38
rs780471238 Chr.6: 106316149-106316149 on Build GRCh38
rs772777225 Chr.6: 106316313-106316313 on Build GRCh38
rs976841437 Chr.6: 106316343-106316343 on Build GRCh38
rs7755637 Chr.6: 106316503-106316503 on Build GRCh38
rs967605433 Chr.6: 106316461-106316461 on Build GRCh38
rs965961643 Chr.6: 106316541-106316542 on Build GRCh38
rs7755506 Chr.6: 106316486-106316486 on Build GRCh38
rs1003280752 Chr.6: 106316134-106316134 on Build GRCh38
rs953446326 Chr.6: 106316303-106316303 on Build GRCh38
rs114109860 Chr.6: 106316143-106316143 on Build GRCh38
rs542567440 Chr.6: 106316076-106316076 on Build GRCh38
rs961688404 Chr.6: 106316161-106316161 on Build GRCh38
rs906702550 Chr.6: 106316511-106316511 on Build GRCh38
rs957097995 Chr.6: 106316045-106316045 on Build GRCh38
rs11541477 Chr.6: 106316239-106316239 on Build GRCh38

View Full Product Details