Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23689316-23689769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM221A
Entrez Gene ID
340277
Gene Name
family with sequence similarity 221 member A
Gene Aliases
C7orf46
UniGene
Hs.592178
Gene Chromosome Location
Chr.7: 23680114-23703249 on Build GRCh38
Transcripts
NM_001127364.2 NM_001127365.2 NM_001300932.1 NM_199136.4 XM_011515369.2 XM_011515370.2 XM_011515371.2 XM_011515372.2 XM_017012134.1 XM_017012135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3026246 c.344C>T p.P115L Chr.7: 23689373-23689373 on Build GRCh38
4919833 c.388G>A p.A130T Chr.7: 23689417-23689417 on Build GRCh38
3995514 c.344C>G p.P115R Chr.7: 23689373-23689373 on Build GRCh38
3880291 c.392A>T p.D131V Chr.7: 23689421-23689421 on Build GRCh38
6109693 c.403G>A p.A135T Chr.7: 23689432-23689432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768438024 Chr.7: 23689374-23689374 on Build GRCh38
rs754006444 Chr.7: 23689462-23689462 on Build GRCh38
rs751870008 Chr.7: 23689436-23689436 on Build GRCh38
rs756193289 Chr.7: 23689458-23689458 on Build GRCh38
rs776283408 Chr.7: 23689402-23689402 on Build GRCh38
rs768409590 Chr.7: 23689415-23689416 on Build GRCh38
rs891304829 Chr.7: 23689524-23689524 on Build GRCh38
rs895277808 Chr.7: 23689623-23689623 on Build GRCh38
rs1043788491 Chr.7: 23689744-23689744 on Build GRCh38
rs780009666 Chr.7: 23689461-23689461 on Build GRCh38
rs151159653 Chr.7: 23689437-23689437 on Build GRCh38
rs756281093 Chr.7: 23689353-23689353 on Build GRCh38
rs201198425 Chr.7: 23689456-23689456 on Build GRCh38
rs536299854 Chr.7: 23689523-23689523 on Build GRCh38
rs979493556 Chr.7: 23689729-23689729 on Build GRCh38
rs749473997 Chr.7: 23689371-23689371 on Build GRCh38
rs748912573 Chr.7: 23689364-23689364 on Build GRCh38
rs757828068 Chr.7: 23689344-23689344 on Build GRCh38
rs186669590 Chr.7: 23689513-23689513 on Build GRCh38
rs774003324 Chr.7: 23689386-23689386 on Build GRCh38
rs948229014 Chr.7: 23689640-23689640 on Build GRCh38
rs745915879 Chr.7: 23689349-23689349 on Build GRCh38
rs964812176 Chr.7: 23689440-23689440 on Build GRCh38
rs142130918 Chr.7: 23689401-23689401 on Build GRCh38
rs201569286 Chr.7: 23689492-23689492 on Build GRCh38
rs575599254 Chr.7: 23689676-23689676 on Build GRCh38
rs764999905 Chr.7: 23689418-23689418 on Build GRCh38
rs146437347 Chr.7: 23689358-23689358 on Build GRCh38
rs138938244 Chr.7: 23689397-23689397 on Build GRCh38
rs768084256 Chr.7: 23689452-23689452 on Build GRCh38
rs1008419429 Chr.7: 23689543-23689543 on Build GRCh38
rs766035754 Chr.7: 23689593-23689593 on Build GRCh38
rs749948035 Chr.7: 23689581-23689581 on Build GRCh38
rs755148945 Chr.7: 23689491-23689491 on Build GRCh38
rs781782705 Chr.7: 23689347-23689347 on Build GRCh38
rs573664797 Chr.7: 23689515-23689515 on Build GRCh38
rs752050425 Chr.7: 23689342-23689342 on Build GRCh38
rs370705342 Chr.7: 23689592-23689592 on Build GRCh38
rs17855786 Chr.7: 23689412-23689412 on Build GRCh38
rs748002654 Chr.7: 23689388-23689388 on Build GRCh38
rs747916372 Chr.7: 23689496-23689496 on Build GRCh38
rs554186164 Chr.7: 23689522-23689522 on Build GRCh38
rs955963281 Chr.7: 23689425-23689425 on Build GRCh38
rs771966204 Chr.7: 23689396-23689396 on Build GRCh38
rs1022805256 Chr.7: 23689391-23689391 on Build GRCh38
rs368463433 Chr.7: 23689509-23689509 on Build GRCh38

View Full Product Details