Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100168425-100169024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST4
Entrez Gene ID
79690
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
GAL3ST-4
UniGene
Hs.44856
Gene Chromosome Location
Chr.7: 100159242-100168750 on Build GRCh38
Transcripts
NM_024637.4

SNP Detail

SNP ID Chromosome Location
rs972584232 Chr.7: 100168477-100168477 on Build GRCh38
rs936981620 Chr.7: 100168512-100168512 on Build GRCh38
rs891765302 Chr.7: 100168929-100168929 on Build GRCh38
rs532908543 Chr.7: 100168899-100168899 on Build GRCh38
rs779968787 Chr.7: 100168884-100168885 on Build GRCh38
rs200789574 Chr.7: 100168953-100168956 on Build GRCh38
rs562009062 Chr.7: 100168606-100168606 on Build GRCh38
rs971185009 Chr.7: 100168843-100168843 on Build GRCh38
rs188415752 Chr.7: 100168714-100168714 on Build GRCh38
rs946317740 Chr.7: 100168584-100168584 on Build GRCh38
rs967886576 Chr.7: 100168670-100168670 on Build GRCh38
rs562471004 Chr.7: 100168884-100168884 on Build GRCh38
rs551482790 Chr.7: 100168959-100168959 on Build GRCh38
rs914820104 Chr.7: 100168579-100168579 on Build GRCh38
rs927036319 Chr.7: 100168506-100168506 on Build GRCh38
rs764343165 Chr.7: 100168918-100168918 on Build GRCh38
rs889803984 Chr.7: 100168470-100168470 on Build GRCh38
rs540418926 Chr.7: 100168602-100168602 on Build GRCh38
rs898678121 Chr.7: 100168822-100168822 on Build GRCh38
rs1021211772 Chr.7: 100168667-100168667 on Build GRCh38
rs550801301 Chr.7: 100168765-100168765 on Build GRCh38
rs962990005 Chr.7: 100168467-100168467 on Build GRCh38
rs181180175 Chr.7: 100168734-100168734 on Build GRCh38
rs556093215 Chr.7: 100168457-100168457 on Build GRCh38
rs931469156 Chr.7: 100168847-100168847 on Build GRCh38
rs1008259424 Chr.7: 100168549-100168549 on Build GRCh38
rs1050436904 Chr.7: 100168699-100168699 on Build GRCh38
rs1019607142 Chr.7: 100168559-100168559 on Build GRCh38
rs979605685 Chr.7: 100168682-100168682 on Build GRCh38
rs1006643250 Chr.7: 100168453-100168453 on Build GRCh38
rs866390481 Chr.7: 100168792-100168792 on Build GRCh38
rs1009919089 Chr.7: 100168930-100168930 on Build GRCh38
rs937745634 Chr.7: 100168717-100168717 on Build GRCh38
rs990260456 Chr.7: 100168530-100168530 on Build GRCh38
rs764928119 Chr.7: 100168503-100168503 on Build GRCh38
rs1037245846 Chr.7: 100168821-100168821 on Build GRCh38
rs1000731078 Chr.7: 100168954-100168954 on Build GRCh38
rs889204334 Chr.7: 100168704-100168704 on Build GRCh38
rs569135172 Chr.7: 100168881-100168884 on Build GRCh38
rs150226909 Chr.7: 100168459-100168459 on Build GRCh38
rs989817509 Chr.7: 100168935-100168935 on Build GRCh38
rs200433042 Chr.7: 100168504-100168504 on Build GRCh38
rs750911287 Chr.7: 100168793-100168793 on Build GRCh38
rs914248445 Chr.7: 100168974-100168974 on Build GRCh38
rs917574562 Chr.7: 100168748-100168748 on Build GRCh38
rs1025507258 Chr.7: 100168498-100168498 on Build GRCh38
rs991836946 Chr.7: 100168993-100168993 on Build GRCh38
rs994467263 Chr.7: 100168766-100168766 on Build GRCh38
rs945753512 Chr.7: 100168985-100168985 on Build GRCh38
rs532321267 Chr.7: 100168754-100168754 on Build GRCh38
rs578044056 Chr.7: 100168458-100168458 on Build GRCh38
rs1050335530 Chr.7: 100168880-100168881 on Build GRCh38
rs991827112 Chr.7: 100168735-100168735 on Build GRCh38
rs879612892 Chr.7: 100168500-100168500 on Build GRCh38
rs553950210 Chr.7: 100168472-100168472 on Build GRCh38
rs968358532 Chr.7: 100168937-100168937 on Build GRCh38
rs1034096136 Chr.7: 100168892-100168892 on Build GRCh38
rs933505869 Chr.7: 100168649-100168649 on Build GRCh38
rs35289232 Chr.7: 100168539-100168540 on Build GRCh38
rs113196307 Chr.7: 100168545-100168545 on Build GRCh38
rs369479533 Chr.7: 100168891-100168894 on Build GRCh38
rs981274358 Chr.7: 100168845-100168845 on Build GRCh38

View Full Product Details