Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121378550-121379061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3C
Entrez Gene ID
10447
Gene Name
family with sequence similarity 3 member C
Gene Aliases
GS3786, ILEI
UniGene
Hs.434053
Gene Chromosome Location
Chr.7: 121348851-121396369 on Build GRCh38
Transcripts
NM_001040020.1 NM_014888.2 XM_011515736.2 XM_011515737.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
452217 c.117T>G p.F39L Chr.7: 121378911-121378911 on Build GRCh38
1084591 c.88A>C p.K30Q Chr.7: 121378940-121378940 on Build GRCh38
5881310 c.73C>A p.Q25K Chr.7: 121378955-121378955 on Build GRCh38
7102988 c.28G>A p.V10M Chr.7: 121379000-121379000 on Build GRCh38
5629618 c.91A>G p.M31V Chr.7: 121378937-121378937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117771746 Chr.7: 121378786-121378786 on Build GRCh38
rs923673597 Chr.7: 121378890-121378890 on Build GRCh38
rs200231128 Chr.7: 121378715-121378716 on Build GRCh38
rs910584373 Chr.7: 121378745-121378746 on Build GRCh38
rs768266059 Chr.7: 121378914-121378914 on Build GRCh38
rs905102059 Chr.7: 121378666-121378666 on Build GRCh38
rs954393404 Chr.7: 121378815-121378815 on Build GRCh38
rs1044790810 Chr.7: 121378664-121378664 on Build GRCh38
rs543740456 Chr.7: 121378816-121378816 on Build GRCh38
rs1026324545 Chr.7: 121378713-121378713 on Build GRCh38
rs893426756 Chr.7: 121378639-121378639 on Build GRCh38
rs773826328 Chr.7: 121378985-121378985 on Build GRCh38
rs397756259 Chr.7: 121378717-121378718 on Build GRCh38
rs79125101 Chr.7: 121378605-121378605 on Build GRCh38
rs1027743653 Chr.7: 121378732-121378732 on Build GRCh38
rs576142741 Chr.7: 121378618-121378618 on Build GRCh38
rs748584682 Chr.7: 121378972-121378972 on Build GRCh38
rs1044739712 Chr.7: 121379035-121379035 on Build GRCh38
rs764004885 Chr.7: 121378910-121378910 on Build GRCh38
rs961922151 Chr.7: 121378721-121378721 on Build GRCh38
rs181249571 Chr.7: 121378677-121378677 on Build GRCh38
rs753642906 Chr.7: 121378633-121378633 on Build GRCh38
rs1034832459 Chr.7: 121378708-121378708 on Build GRCh38
rs878867994 Chr.7: 121378688-121378688 on Build GRCh38
rs756974338 Chr.7: 121378913-121378913 on Build GRCh38
rs751362515 Chr.7: 121378911-121378911 on Build GRCh38
rs772300135 Chr.7: 121378975-121378975 on Build GRCh38
rs756287917 Chr.7: 121378927-121378927 on Build GRCh38
rs761389458 Chr.7: 121378641-121378641 on Build GRCh38
rs936434467 Chr.7: 121378899-121378899 on Build GRCh38
rs914992663 Chr.7: 121378926-121378926 on Build GRCh38
rs1003489909 Chr.7: 121378699-121378699 on Build GRCh38
rs3068114 Chr.7: 121378713-121378714 on Build GRCh38
rs577773549 Chr.7: 121378833-121378833 on Build GRCh38
rs1016523913 Chr.7: 121378781-121378781 on Build GRCh38
rs151288890 Chr.7: 121378934-121378934 on Build GRCh38
rs749522040 Chr.7: 121378933-121378933 on Build GRCh38
rs948973252 Chr.7: 121379000-121379000 on Build GRCh38
rs752403094 Chr.7: 121378885-121378885 on Build GRCh38
rs72382715 Chr.7: 121378714-121378715 on Build GRCh38
rs747465982 Chr.7: 121379012-121379012 on Build GRCh38
rs555121287 Chr.7: 121378702-121378702 on Build GRCh38
rs769442838 Chr.7: 121379025-121379025 on Build GRCh38
rs1055033542 Chr.7: 121378594-121378594 on Build GRCh38
rs1013596258 Chr.7: 121378614-121378614 on Build GRCh38
rs754894167 Chr.7: 121378677-121378678 on Build GRCh38
rs552994541 Chr.7: 121378742-121378742 on Build GRCh38
rs372222980 Chr.7: 121378882-121378882 on Build GRCh38
rs763940855 Chr.7: 121378631-121378631 on Build GRCh38
rs764883473 Chr.7: 121378644-121378644 on Build GRCh38
rs750318600 Chr.7: 121378917-121378917 on Build GRCh38
rs567820522 Chr.7: 121378724-121378724 on Build GRCh38
rs745907404 Chr.7: 121378930-121378930 on Build GRCh38
rs759133488 Chr.7: 121378870-121378870 on Build GRCh38
rs564048822 Chr.7: 121378935-121378935 on Build GRCh38
rs917726 Chr.7: 121378803-121378803 on Build GRCh38
rs376952658 Chr.7: 121378936-121378936 on Build GRCh38
rs780037177 Chr.7: 121378931-121378931 on Build GRCh38
rs79379594 Chr.7: 121378810-121378810 on Build GRCh38
rs762637223 Chr.7: 121378904-121378904 on Build GRCh38
rs1054599048 Chr.7: 121378932-121378932 on Build GRCh38
rs775092417 Chr.7: 121379031-121379031 on Build GRCh38
rs536939777 Chr.7: 121378737-121378737 on Build GRCh38

View Full Product Details