Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150742515-150743086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP1-GIMAP5
Entrez Gene ID
100527949
Gene Name
GIMAP1-GIMAP5 readthrough
Gene Aliases
-
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150716557-150743649 on Build GRCh38
Transcripts
NM_001199577.1 NM_001303630.1 NM_018384.4

Gene Symbol
GIMAP5
Entrez Gene ID
55340
Gene Name
GTPase, IMAP family member 5
Gene Aliases
HIMAP3, IAN-5, IAN4, IAN4L1, IAN5, IMAP3, IROD
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150737348-150743649 on Build GRCh38
Transcripts
NM_001199577.1 NM_001303630.1 NM_018384.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3411792 c.884T>A p.L295H Chr.7: 150743023-150743023 on Build GRCh38
5406949 c.609C>T p.L203L Chr.7: 150742748-150742748 on Build GRCh38
7101739 c.803C>T p.A268V Chr.7: 150742942-150742942 on Build GRCh38
3636123 c.790G>A p.E264K Chr.7: 150742929-150742929 on Build GRCh38
334766 c.881T>C p.I294T Chr.7: 150743020-150743020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756881232 Chr.7: 150742571-150742571 on Build GRCh38
rs772939317 Chr.7: 150743018-150743018 on Build GRCh38
rs763872372 Chr.7: 150742941-150742941 on Build GRCh38
rs61751036 Chr.7: 150742736-150742736 on Build GRCh38
rs771317324 Chr.7: 150742923-150742923 on Build GRCh38
rs774162620 Chr.7: 150743023-150743023 on Build GRCh38
rs552913796 Chr.7: 150742753-150742753 on Build GRCh38
rs762863387 Chr.7: 150742982-150742983 on Build GRCh38
rs866287630 Chr.7: 150742789-150742789 on Build GRCh38
rs1003396391 Chr.7: 150743059-150743059 on Build GRCh38
rs199764342 Chr.7: 150742897-150742897 on Build GRCh38
rs772407527 Chr.7: 150742907-150742907 on Build GRCh38
rs749165559 Chr.7: 150742591-150742591 on Build GRCh38
rs747068784 Chr.7: 150742981-150742982 on Build GRCh38
rs751642629 Chr.7: 150743034-150743034 on Build GRCh38
rs747550555 Chr.7: 150742971-150742971 on Build GRCh38
rs376880387 Chr.7: 150742770-150742770 on Build GRCh38
rs752925970 Chr.7: 150743029-150743029 on Build GRCh38
rs778817495 Chr.7: 150742608-150742608 on Build GRCh38
rs759323787 Chr.7: 150742814-150742814 on Build GRCh38
rs754045016 Chr.7: 150742886-150742886 on Build GRCh38
rs756243315 Chr.7: 150742775-150742775 on Build GRCh38
rs773185282 Chr.7: 150742979-150742980 on Build GRCh38
rs201290574 Chr.7: 150742867-150742867 on Build GRCh38
rs768199039 Chr.7: 150742998-150742998 on Build GRCh38
rs1051756770 Chr.7: 150742549-150742549 on Build GRCh38
rs746075907 Chr.7: 150742694-150742694 on Build GRCh38
rs772318869 Chr.7: 150742845-150742845 on Build GRCh38
rs770097013 Chr.7: 150742711-150742711 on Build GRCh38
rs375326276 Chr.7: 150742554-150742554 on Build GRCh38
rs918681913 Chr.7: 150742767-150742767 on Build GRCh38
rs76946650 Chr.7: 150742989-150742989 on Build GRCh38
rs779419627 Chr.7: 150742835-150742836 on Build GRCh38
rs1051255215 Chr.7: 150742869-150742869 on Build GRCh38
rs749909322 Chr.7: 150742577-150742577 on Build GRCh38
rs180974779 Chr.7: 150742786-150742786 on Build GRCh38
rs752298821 Chr.7: 150742852-150742852 on Build GRCh38
rs753161728 Chr.7: 150742559-150742559 on Build GRCh38
rs765713700 Chr.7: 150742558-150742558 on Build GRCh38
rs769533038 Chr.7: 150742985-150742985 on Build GRCh38
rs746191943 Chr.7: 150742759-150742759 on Build GRCh38
rs765212870 Chr.7: 150742760-150742760 on Build GRCh38
rs765904238 Chr.7: 150742799-150742799 on Build GRCh38
rs747265690 Chr.7: 150742917-150742917 on Build GRCh38
rs72650695 Chr.7: 150742750-150742750 on Build GRCh38
rs746872238 Chr.7: 150742773-150742773 on Build GRCh38
rs905926485 Chr.7: 150743040-150743040 on Build GRCh38
rs199678719 Chr.7: 150742741-150742741 on Build GRCh38
rs764838871 Chr.7: 150742842-150742842 on Build GRCh38
rs762584358 Chr.7: 150742935-150742935 on Build GRCh38
rs773424993 Chr.7: 150742787-150742787 on Build GRCh38
rs200640496 Chr.7: 150742927-150742927 on Build GRCh38
rs771386178 Chr.7: 150742976-150742976 on Build GRCh38
rs757736944 Chr.7: 150743044-150743044 on Build GRCh38
rs761120317 Chr.7: 150742798-150742798 on Build GRCh38
rs776346628 Chr.7: 150742801-150742801 on Build GRCh38
rs752671075 Chr.7: 150742769-150742769 on Build GRCh38
rs13229806 Chr.7: 150743010-150743010 on Build GRCh38
rs368122062 Chr.7: 150743019-150743019 on Build GRCh38
rs777685680 Chr.7: 150742693-150742693 on Build GRCh38
rs1046355 Chr.7: 150742955-150742955 on Build GRCh38
rs186198396 Chr.7: 150742855-150742855 on Build GRCh38
rs968780914 Chr.7: 150743043-150743043 on Build GRCh38
rs778930695 Chr.7: 150742900-150742900 on Build GRCh38
rs763339553 Chr.7: 150742643-150742643 on Build GRCh38
rs1021223904 Chr.7: 150742746-150742746 on Build GRCh38
rs779012038 Chr.7: 150742678-150742678 on Build GRCh38
rs768736186 Chr.7: 150742780-150742780 on Build GRCh38
rs778371662 Chr.7: 150742914-150742914 on Build GRCh38
rs200235816 Chr.7: 150742537-150742537 on Build GRCh38
rs72650696 Chr.7: 150742856-150742856 on Build GRCh38
rs893104714 Chr.7: 150742925-150742925 on Build GRCh38
rs934922713 Chr.7: 150742896-150742896 on Build GRCh38
rs780845085 Chr.7: 150742574-150742574 on Build GRCh38
rs367554935 Chr.7: 150742782-150742782 on Build GRCh38
rs543877528 Chr.7: 150742562-150742562 on Build GRCh38
rs149571417 Chr.7: 150742566-150742566 on Build GRCh38
rs368965016 Chr.7: 150742548-150742548 on Build GRCh38
rs183707481 Chr.7: 150742618-150742618 on Build GRCh38
rs761419370 Chr.7: 150743065-150743066 on Build GRCh38
rs773691352 Chr.7: 150742745-150742745 on Build GRCh38
rs113666617 Chr.7: 150742948-150742948 on Build GRCh38
rs751272459 Chr.7: 150742860-150742860 on Build GRCh38
rs776825281 Chr.7: 150742553-150742553 on Build GRCh38
rs758285735 Chr.7: 150742686-150742686 on Build GRCh38
rs778080681 Chr.7: 150742970-150742970 on Build GRCh38
rs751872887 Chr.7: 150742654-150742654 on Build GRCh38
rs767490478 Chr.7: 150743006-150743006 on Build GRCh38
rs562399062 Chr.7: 150742612-150742612 on Build GRCh38
rs147182394 Chr.7: 150742672-150742672 on Build GRCh38
rs569763678 Chr.7: 150742712-150742712 on Build GRCh38
rs74162072 Chr.7: 150742880-150742880 on Build GRCh38
rs781674820 Chr.7: 150742980-150742980 on Build GRCh38
rs753235405 Chr.7: 150742756-150742756 on Build GRCh38
rs761778169 Chr.7: 150743003-150743003 on Build GRCh38
rs764207608 Chr.7: 150743031-150743031 on Build GRCh38
rs140160080 Chr.7: 150743002-150743002 on Build GRCh38
rs773799372 Chr.7: 150743001-150743001 on Build GRCh38
rs13229808 Chr.7: 150743013-150743013 on Build GRCh38
rs138178760 Chr.7: 150742611-150742611 on Build GRCh38
rs775709068 Chr.7: 150742642-150742642 on Build GRCh38
rs199689906 Chr.7: 150742981-150742981 on Build GRCh38
rs747907400 Chr.7: 150742683-150742683 on Build GRCh38
rs267601411 Chr.7: 150743030-150743030 on Build GRCh38
rs980646452 Chr.7: 150742633-150742633 on Build GRCh38
rs769112260 Chr.7: 150742732-150742732 on Build GRCh38
rs61751028 Chr.7: 150742649-150742649 on Build GRCh38
rs756199537 Chr.7: 150742928-150742928 on Build GRCh38
rs532533117 Chr.7: 150742841-150742841 on Build GRCh38
rs533934192 Chr.7: 150742951-150742951 on Build GRCh38
rs372355155 Chr.7: 150742671-150742671 on Build GRCh38
rs751529208 Chr.7: 150742945-150742945 on Build GRCh38
rs769752170 Chr.7: 150742638-150742638 on Build GRCh38
rs1034851496 Chr.7: 150743061-150743061 on Build GRCh38
rs61750977 Chr.7: 150742581-150742581 on Build GRCh38
rs760969277 Chr.7: 150742546-150742546 on Build GRCh38
rs574994840 Chr.7: 150742809-150742809 on Build GRCh38
rs755852343 Chr.7: 150742666-150742666 on Build GRCh38
rs768703802 Chr.7: 150742600-150742600 on Build GRCh38
rs758505999 Chr.7: 150742960-150742960 on Build GRCh38
rs752731508 Chr.7: 150742957-150742957 on Build GRCh38
rs375450342 Chr.7: 150742757-150742757 on Build GRCh38
rs769621407 Chr.7: 150742929-150742929 on Build GRCh38
rs765372741 Chr.7: 150743023-150743023 on Build GRCh38
rs748105968 Chr.7: 150742824-150742824 on Build GRCh38
rs763942721 Chr.7: 150742984-150742984 on Build GRCh38
rs750055689 Chr.7: 150742665-150742665 on Build GRCh38
rs61751050 Chr.7: 150742952-150742952 on Build GRCh38
rs775287134 Chr.7: 150742988-150742988 on Build GRCh38
rs749408342 Chr.7: 150742714-150742714 on Build GRCh38
rs765800153 Chr.7: 150742670-150742670 on Build GRCh38
rs780355836 Chr.7: 150742779-150742779 on Build GRCh38
rs866576893 Chr.7: 150742875-150742875 on Build GRCh38
rs769723813 Chr.7: 150742980-150742981 on Build GRCh38
rs368627191 Chr.7: 150742569-150742569 on Build GRCh38
rs775092732 Chr.7: 150742934-150742934 on Build GRCh38
rs748274505 Chr.7: 150742904-150742904 on Build GRCh38

View Full Product Details