Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134993452-134994042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
UniGene
Hs.648616
Gene Chromosome Location
Chr.7: 134985169-135135782 on Build GRCh38
Transcripts
NM_178563.3 XM_011516122.2 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4824986 c.263G>A p.R88Q Chr.7: 134993631-134993631 on Build GRCh38
5139807 c.72delC p.R26fs*7 Chr.7: 134993518-134993518 on Build GRCh38
4417512 c.56T>A p.F19Y Chr.7: 134993502-134993502 on Build GRCh38
5006282 c.269C>A p.P90Q Chr.7: 134993637-134993637 on Build GRCh38
4442225 c.209G>A p.R70H Chr.7: 134993577-134993577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941415156 Chr.7: 134993557-134993557 on Build GRCh38
rs190765263 Chr.7: 134993856-134993856 on Build GRCh38
rs541084151 Chr.7: 134993631-134993631 on Build GRCh38
rs750194969 Chr.7: 134993507-134993507 on Build GRCh38
rs755004222 Chr.7: 134993500-134993501 on Build GRCh38
rs989595729 Chr.7: 134993693-134993693 on Build GRCh38
rs909852873 Chr.7: 134993843-134993843 on Build GRCh38
rs764409851 Chr.7: 134993503-134993503 on Build GRCh38
rs183157366 Chr.7: 134993919-134993919 on Build GRCh38
rs572171708 Chr.7: 134993624-134993624 on Build GRCh38
rs544122297 Chr.7: 134993758-134993758 on Build GRCh38
rs775394843 Chr.7: 134993676-134993676 on Build GRCh38
rs2252826 Chr.7: 134993772-134993772 on Build GRCh38
rs117732894 Chr.7: 134993576-134993576 on Build GRCh38
rs1051763779 Chr.7: 134994012-134994012 on Build GRCh38
rs769888283 Chr.7: 134993658-134993658 on Build GRCh38
rs962553813 Chr.7: 134993765-134993765 on Build GRCh38
rs200515296 Chr.7: 134993556-134993556 on Build GRCh38
rs759611158 Chr.7: 134993481-134993481 on Build GRCh38
rs747721890 Chr.7: 134993577-134993577 on Build GRCh38
rs770641709 Chr.7: 134993602-134993602 on Build GRCh38
rs945007297 Chr.7: 134993917-134993917 on Build GRCh38
rs1040398992 Chr.7: 134993613-134993613 on Build GRCh38
rs998465917 Chr.7: 134994011-134994011 on Build GRCh38
rs906437343 Chr.7: 134993977-134993977 on Build GRCh38
rs561505613 Chr.7: 134993673-134993673 on Build GRCh38
rs747311359 Chr.7: 134993592-134993592 on Build GRCh38
rs766413838 Chr.7: 134993542-134993542 on Build GRCh38
rs771356904 Chr.7: 134993618-134993618 on Build GRCh38
rs369701730 Chr.7: 134993583-134993583 on Build GRCh38
rs759137090 Chr.7: 134993651-134993651 on Build GRCh38
rs371774667 Chr.7: 134993505-134993505 on Build GRCh38
rs187933290 Chr.7: 134993998-134993998 on Build GRCh38
rs867397083 Chr.7: 134993972-134993972 on Build GRCh38
rs777491552 Chr.7: 134993588-134993588 on Build GRCh38
rs1004276178 Chr.7: 134993738-134993738 on Build GRCh38
rs1046369815 Chr.7: 134993924-134993924 on Build GRCh38
rs960664630 Chr.7: 134993795-134993795 on Build GRCh38
rs2718161 Chr.7: 134993879-134993879 on Build GRCh38
rs548269817 Chr.7: 134994006-134994006 on Build GRCh38
rs1031197830 Chr.7: 134993646-134993646 on Build GRCh38
rs12540686 Chr.7: 134993712-134993712 on Build GRCh38
rs926554354 Chr.7: 134993523-134993523 on Build GRCh38
rs186048202 Chr.7: 134993522-134993522 on Build GRCh38
rs964172517 Chr.7: 134993754-134993754 on Build GRCh38
rs754513342 Chr.7: 134993571-134993571 on Build GRCh38
rs764919284 Chr.7: 134993688-134993688 on Build GRCh38
rs998810279 Chr.7: 134993657-134993657 on Build GRCh38
rs1000357047 Chr.7: 134993612-134993612 on Build GRCh38
rs916453344 Chr.7: 134993902-134993902 on Build GRCh38
rs900084341 Chr.7: 134993623-134993623 on Build GRCh38
rs745930842 Chr.7: 134993517-134993518 on Build GRCh38
rs1056137967 Chr.7: 134993564-134993564 on Build GRCh38
rs926410744 Chr.7: 134993788-134993788 on Build GRCh38
rs148391971 Chr.7: 134993499-134993499 on Build GRCh38
rs765293678 Chr.7: 134993489-134993489 on Build GRCh38
rs999182262 Chr.7: 134993638-134993638 on Build GRCh38
rs929994013 Chr.7: 134993790-134993790 on Build GRCh38
rs2348049 Chr.7: 134993502-134993502 on Build GRCh38
rs559071638 Chr.7: 134993630-134993630 on Build GRCh38
rs748506722 Chr.7: 134993947-134993947 on Build GRCh38

View Full Product Details