Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 132885534-132886109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD3
Entrez Gene ID
54927
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
MINOS3, Mic19, PPP1R22
UniGene
Hs.655010
Gene Chromosome Location
Chr.7: 132784862-133082158 on Build GRCh38
Transcripts
NM_001317177.1 NM_017812.3 NR_133671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5357892 c.370-3delT p.? Chr.7: 132885748-132885748 on Build GRCh38
6209602 c.427G>C p.E143Q Chr.7: 132885688-132885688 on Build GRCh38
744477 c.450G>T p.E150D Chr.7: 132885665-132885665 on Build GRCh38
4587129 c.417A>C p.A139A Chr.7: 132885698-132885698 on Build GRCh38
2770917 c.391G>A p.D131N Chr.7: 132885724-132885724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044811611 Chr.7: 132885557-132885557 on Build GRCh38
rs373917058 Chr.7: 132885739-132885739 on Build GRCh38
rs951779566 Chr.7: 132885699-132885699 on Build GRCh38
rs902621105 Chr.7: 132886067-132886067 on Build GRCh38
rs768799044 Chr.7: 132885711-132885711 on Build GRCh38
rs961955358 Chr.7: 132886074-132886074 on Build GRCh38
rs774561008 Chr.7: 132885717-132885717 on Build GRCh38
rs201352617 Chr.7: 132885634-132885634 on Build GRCh38
rs917728101 Chr.7: 132885927-132885927 on Build GRCh38
rs986956161 Chr.7: 132885581-132885581 on Build GRCh38
rs773327023 Chr.7: 132885727-132885727 on Build GRCh38
rs980241499 Chr.7: 132885935-132885936 on Build GRCh38
rs933748300 Chr.7: 132885575-132885575 on Build GRCh38
rs763603359 Chr.7: 132885643-132885643 on Build GRCh38
rs940244069 Chr.7: 132885603-132885603 on Build GRCh38
rs946255523 Chr.7: 132885978-132885978 on Build GRCh38
rs1055015346 Chr.7: 132886011-132886011 on Build GRCh38
rs780751926 Chr.7: 132885653-132885653 on Build GRCh38
rs908323132 Chr.7: 132885946-132885946 on Build GRCh38
rs377651271 Chr.7: 132885651-132885651 on Build GRCh38
rs1014515218 Chr.7: 132886059-132886059 on Build GRCh38
rs762725233 Chr.7: 132885607-132885607 on Build GRCh38
rs756576483 Chr.7: 132885800-132885800 on Build GRCh38
rs1010415357 Chr.7: 132885610-132885610 on Build GRCh38
rs534516884 Chr.7: 132885698-132885698 on Build GRCh38
rs756435027 Chr.7: 132885792-132885792 on Build GRCh38
rs187402681 Chr.7: 132885876-132885876 on Build GRCh38
rs192550713 Chr.7: 132885931-132885931 on Build GRCh38
rs767924939 Chr.7: 132885680-132885680 on Build GRCh38
rs575500976 Chr.7: 132885991-132885991 on Build GRCh38
rs12707059 Chr.7: 132885598-132885598 on Build GRCh38
rs957293572 Chr.7: 132885859-132885859 on Build GRCh38
rs530579905 Chr.7: 132885936-132885936 on Build GRCh38
rs756460103 Chr.7: 132885672-132885672 on Build GRCh38
rs937992929 Chr.7: 132885647-132885647 on Build GRCh38
rs200873719 Chr.7: 132885720-132885720 on Build GRCh38
rs940172689 Chr.7: 132885973-132885973 on Build GRCh38
rs896480978 Chr.7: 132886019-132886019 on Build GRCh38
rs571410928 Chr.7: 132885805-132885805 on Build GRCh38
rs986955731 Chr.7: 132885933-132885933 on Build GRCh38
rs546322725 Chr.7: 132885810-132885810 on Build GRCh38
rs764943413 Chr.7: 132885729-132885729 on Build GRCh38
rs991967783 Chr.7: 132885916-132885918 on Build GRCh38
rs762860961 Chr.7: 132885743-132885743 on Build GRCh38
rs535287020 Chr.7: 132885919-132885919 on Build GRCh38
rs765171300 Chr.7: 132885628-132885628 on Build GRCh38
rs759298066 Chr.7: 132885626-132885626 on Build GRCh38
rs759334307 Chr.7: 132885728-132885728 on Build GRCh38
rs780572296 Chr.7: 132885688-132885688 on Build GRCh38
rs750077465 Chr.7: 132885784-132885784 on Build GRCh38
rs555233844 Chr.7: 132885990-132885990 on Build GRCh38
rs199500967 Chr.7: 132885776-132885776 on Build GRCh38
rs756903257 Chr.7: 132885652-132885652 on Build GRCh38
rs974234365 Chr.7: 132885975-132885975 on Build GRCh38
rs1028000654 Chr.7: 132885891-132885891 on Build GRCh38
rs376803690 Chr.7: 132885697-132885697 on Build GRCh38
rs746224115 Chr.7: 132885667-132885667 on Build GRCh38
rs938276375 Chr.7: 132886010-132886010 on Build GRCh38
rs80199813 Chr.7: 132886068-132886068 on Build GRCh38
rs761365223 Chr.7: 132885768-132885768 on Build GRCh38
rs996739700 Chr.7: 132886025-132886025 on Build GRCh38
rs927341398 Chr.7: 132885571-132885571 on Build GRCh38
rs1042856143 Chr.7: 132886048-132886048 on Build GRCh38
rs949451905 Chr.7: 132886026-132886026 on Build GRCh38
rs772401857 Chr.7: 132885722-132885722 on Build GRCh38
rs936016929 Chr.7: 132885950-132885950 on Build GRCh38
rs1054828497 Chr.7: 132885970-132885970 on Build GRCh38
rs887485445 Chr.7: 132886034-132886034 on Build GRCh38
rs752605987 Chr.7: 132885631-132885631 on Build GRCh38
rs145011493 Chr.7: 132885794-132885794 on Build GRCh38
rs1005894126 Chr.7: 132886035-132886036 on Build GRCh38
rs751042163 Chr.7: 132885765-132885765 on Build GRCh38
rs764081479 Chr.7: 132885745-132885745 on Build GRCh38

View Full Product Details