Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27740330-27740906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6113205 c.577G>T p.E193* Chr.8: 27740492-27740492 on Build GRCh38
4777079 c.577G>A p.E193K Chr.8: 27740492-27740492 on Build GRCh38
7262050 c.552-9delT p.? Chr.8: 27740526-27740526 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749559089 Chr.8: 27740747-27740747 on Build GRCh38
rs537106181 Chr.8: 27740723-27740723 on Build GRCh38
rs1025412342 Chr.8: 27740615-27740615 on Build GRCh38
rs774453872 Chr.8: 27740434-27740434 on Build GRCh38
rs371242916 Chr.8: 27740771-27740771 on Build GRCh38
rs905449575 Chr.8: 27740372-27740372 on Build GRCh38
rs996302993 Chr.8: 27740837-27740837 on Build GRCh38
rs983806741 Chr.8: 27740624-27740624 on Build GRCh38
rs765959748 Chr.8: 27740531-27740531 on Build GRCh38
rs936629081 Chr.8: 27740582-27740582 on Build GRCh38
rs375006724 Chr.8: 27740532-27740532 on Build GRCh38
rs1037733842 Chr.8: 27740815-27740816 on Build GRCh38
rs7825085 Chr.8: 27740783-27740783 on Build GRCh38
rs1045063719 Chr.8: 27740844-27740844 on Build GRCh38
rs761515211 Chr.8: 27740504-27740504 on Build GRCh38
rs149714300 Chr.8: 27740869-27740869 on Build GRCh38
rs935196902 Chr.8: 27740760-27740760 on Build GRCh38
rs928777790 Chr.8: 27740662-27740662 on Build GRCh38
rs754647801 Chr.8: 27740535-27740535 on Build GRCh38
rs781005505 Chr.8: 27740584-27740584 on Build GRCh38
rs780776270 Chr.8: 27740537-27740537 on Build GRCh38
rs765204564 Chr.8: 27740505-27740505 on Build GRCh38
rs535281384 Chr.8: 27740818-27740818 on Build GRCh38
rs28587875 Chr.8: 27740654-27740654 on Build GRCh38
rs760652516 Chr.8: 27740531-27740532 on Build GRCh38
rs563832920 Chr.8: 27740405-27740405 on Build GRCh38
rs772238756 Chr.8: 27740460-27740460 on Build GRCh38
rs762515414 Chr.8: 27740521-27740521 on Build GRCh38
rs938870515 Chr.8: 27740671-27740671 on Build GRCh38
rs756484869 Chr.8: 27740554-27740554 on Build GRCh38
rs191001037 Chr.8: 27740635-27740635 on Build GRCh38
rs905229568 Chr.8: 27740865-27740865 on Build GRCh38
rs1030105499 Chr.8: 27740840-27740840 on Build GRCh38
rs17057721 Chr.8: 27740619-27740619 on Build GRCh38
rs7842672 Chr.8: 27740655-27740655 on Build GRCh38
rs753039774 Chr.8: 27740553-27740553 on Build GRCh38
rs999344626 Chr.8: 27740856-27740856 on Build GRCh38
rs1004234938 Chr.8: 27740413-27740413 on Build GRCh38
rs368152128 Chr.8: 27740421-27740421 on Build GRCh38
rs371690676 Chr.8: 27740485-27740485 on Build GRCh38
rs771475514 Chr.8: 27740563-27740563 on Build GRCh38
rs34636629 Chr.8: 27740526-27740526 on Build GRCh38
rs386723887 Chr.8: 27740654-27740655 on Build GRCh38
rs750411921 Chr.8: 27740510-27740510 on Build GRCh38
rs974173037 Chr.8: 27740369-27740369 on Build GRCh38
rs946671349 Chr.8: 27740819-27740819 on Build GRCh38
rs749763409 Chr.8: 27740558-27740558 on Build GRCh38
rs1006288834 Chr.8: 27740457-27740457 on Build GRCh38
rs964814378 Chr.8: 27740486-27740486 on Build GRCh38
rs182945924 Chr.8: 27740759-27740759 on Build GRCh38
rs544046517 Chr.8: 27740403-27740403 on Build GRCh38
rs113421120 Chr.8: 27740579-27740579 on Build GRCh38
rs773289794 Chr.8: 27740532-27740533 on Build GRCh38
rs58973971 Chr.8: 27740722-27740722 on Build GRCh38
rs780149566 Chr.8: 27740700-27740700 on Build GRCh38

View Full Product Details