Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28452052-28452471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO16
Entrez Gene ID
157574
Gene Name
F-box protein 16
Gene Aliases
FBX16
UniGene
Hs.741522
Gene Chromosome Location
Chr.8: 28428408-28490318 on Build GRCh38
Transcripts
NM_001258211.1 NM_172366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751762 c.599G>A p.R200Q Chr.8: 28452385-28452385 on Build GRCh38
6180730 c.549A>G p.L183L Chr.8: 28452435-28452435 on Build GRCh38
3648006 c.519G>A p.K173K Chr.8: 28452465-28452465 on Build GRCh38
2788602 c.598C>T p.R200W Chr.8: 28452386-28452386 on Build GRCh38
4589857 c.522_523insA p.G175fs*6 Chr.8: 28452461-28452462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918796640 Chr.8: 28452144-28452144 on Build GRCh38
rs750060657 Chr.8: 28452198-28452198 on Build GRCh38
rs75701777 Chr.8: 28452339-28452339 on Build GRCh38
rs200400597 Chr.8: 28452271-28452271 on Build GRCh38
rs766852393 Chr.8: 28452292-28452292 on Build GRCh38
rs775951435 Chr.8: 28452122-28452122 on Build GRCh38
rs376070050 Chr.8: 28452252-28452252 on Build GRCh38
rs891261192 Chr.8: 28452153-28452153 on Build GRCh38
rs762809282 Chr.8: 28452444-28452444 on Build GRCh38
rs112031839 Chr.8: 28452121-28452121 on Build GRCh38
rs773801245 Chr.8: 28452446-28452446 on Build GRCh38
rs751165226 Chr.8: 28452351-28452351 on Build GRCh38
rs200232057 Chr.8: 28452212-28452212 on Build GRCh38
rs375104603 Chr.8: 28452431-28452431 on Build GRCh38
rs369379563 Chr.8: 28452400-28452400 on Build GRCh38
rs765203306 Chr.8: 28452338-28452338 on Build GRCh38
rs756753083 Chr.8: 28452308-28452308 on Build GRCh38
rs931470730 Chr.8: 28452171-28452171 on Build GRCh38
rs755301038 Chr.8: 28452296-28452296 on Build GRCh38
rs372049768 Chr.8: 28452325-28452325 on Build GRCh38
rs753153262 Chr.8: 28452378-28452378 on Build GRCh38
rs769607803 Chr.8: 28452419-28452419 on Build GRCh38
rs753564982 Chr.8: 28452383-28452383 on Build GRCh38
rs780246751 Chr.8: 28452205-28452205 on Build GRCh38
rs372114412 Chr.8: 28452386-28452386 on Build GRCh38
rs1001845925 Chr.8: 28452307-28452307 on Build GRCh38
rs777968638 Chr.8: 28452312-28452312 on Build GRCh38
rs371862747 Chr.8: 28452279-28452279 on Build GRCh38
rs754646979 Chr.8: 28452358-28452358 on Build GRCh38
rs768483018 Chr.8: 28452326-28452326 on Build GRCh38
rs776475025 Chr.8: 28452334-28452334 on Build GRCh38
rs938442597 Chr.8: 28452127-28452127 on Build GRCh38
rs112864205 Chr.8: 28452218-28452218 on Build GRCh38
rs952410726 Chr.8: 28452411-28452411 on Build GRCh38
rs760254665 Chr.8: 28452272-28452272 on Build GRCh38
rs375008847 Chr.8: 28452337-28452337 on Build GRCh38
rs769939024 Chr.8: 28452231-28452231 on Build GRCh38
rs756667453 Chr.8: 28452385-28452385 on Build GRCh38
rs776490725 Chr.8: 28452258-28452258 on Build GRCh38
rs900186156 Chr.8: 28452081-28452081 on Build GRCh38
rs747889711 Chr.8: 28452414-28452414 on Build GRCh38
rs748668037 Chr.8: 28452228-28452228 on Build GRCh38
rs970471492 Chr.8: 28452235-28452235 on Build GRCh38
rs753376225 Chr.8: 28452406-28452406 on Build GRCh38
rs748653752 Chr.8: 28452298-28452298 on Build GRCh38
rs370470041 Chr.8: 28452238-28452238 on Build GRCh38
rs370261817 Chr.8: 28452344-28452344 on Build GRCh38
rs765810228 Chr.8: 28452347-28452347 on Build GRCh38
rs781425326 Chr.8: 28452297-28452297 on Build GRCh38
rs377367222 Chr.8: 28452233-28452233 on Build GRCh38
rs984321769 Chr.8: 28452424-28452424 on Build GRCh38
rs770462983 Chr.8: 28452445-28452445 on Build GRCh38
rs774596636 Chr.8: 28452319-28452319 on Build GRCh38
rs773485702 Chr.8: 28452232-28452232 on Build GRCh38
rs767991916 Chr.8: 28452359-28452359 on Build GRCh38
rs149428353 Chr.8: 28452405-28452405 on Build GRCh38
rs138522609 Chr.8: 28452402-28452402 on Build GRCh38
rs181297266 Chr.8: 28452179-28452179 on Build GRCh38
rs758109456 Chr.8: 28452200-28452200 on Build GRCh38
rs928271772 Chr.8: 28452124-28452124 on Build GRCh38
rs754748031 Chr.8: 28452414-28452415 on Build GRCh38
rs763829467 Chr.8: 28452249-28452249 on Build GRCh38
rs540550522 Chr.8: 28452138-28452138 on Build GRCh38
rs367952507 Chr.8: 28452293-28452293 on Build GRCh38
rs376343896 Chr.8: 28452211-28452211 on Build GRCh38
rs113779780 Chr.8: 28452121-28452121 on Build GRCh38
rs1030095756 Chr.8: 28452137-28452137 on Build GRCh38
rs770942534 Chr.8: 28452317-28452317 on Build GRCh38
rs778914888 Chr.8: 28452401-28452401 on Build GRCh38
rs773065842 Chr.8: 28452437-28452437 on Build GRCh38
rs368871325 Chr.8: 28452274-28452274 on Build GRCh38

View Full Product Details