Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119106322-119106910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLEC10
Entrez Gene ID
10584
Gene Name
collectin subfamily member 10
Gene Aliases
CL-34, CLL1
UniGene
Hs.176615
Gene Chromosome Location
Chr.8: 118952167-119108455 on Build GRCh38
Transcripts
NM_001324095.1 NM_006438.4 XM_005250756.3

SNP Detail

SNP ID Chromosome Location
rs377205780 Chr.8: 119106396-119106396 on Build GRCh38
rs774339997 Chr.8: 119106504-119106504 on Build GRCh38
rs1040229459 Chr.8: 119106758-119106758 on Build GRCh38
rs766266060 Chr.8: 119106410-119106410 on Build GRCh38
rs952974530 Chr.8: 119106692-119106692 on Build GRCh38
rs947313500 Chr.8: 119106783-119106783 on Build GRCh38
rs1008028866 Chr.8: 119106875-119106875 on Build GRCh38
rs757329759 Chr.8: 119106716-119106716 on Build GRCh38
rs764258815 Chr.8: 119106581-119106581 on Build GRCh38
rs957204396 Chr.8: 119106346-119106346 on Build GRCh38
rs887452440 Chr.8: 119106878-119106878 on Build GRCh38
rs925291283 Chr.8: 119106690-119106690 on Build GRCh38
rs899915342 Chr.8: 119106884-119106884 on Build GRCh38
rs76175910 Chr.8: 119106495-119106495 on Build GRCh38
rs527430078 Chr.8: 119106774-119106774 on Build GRCh38
rs1064557 Chr.8: 119106672-119106672 on Build GRCh38
rs1064558 Chr.8: 119106755-119106755 on Build GRCh38
rs1064556 Chr.8: 119106378-119106378 on Build GRCh38
rs949561006 Chr.8: 119106734-119106734 on Build GRCh38
rs777869023 Chr.8: 119106568-119106568 on Build GRCh38
rs544267449 Chr.8: 119106516-119106516 on Build GRCh38
rs750921151 Chr.8: 119106826-119106826 on Build GRCh38
rs182630573 Chr.8: 119106850-119106850 on Build GRCh38
rs780333712 Chr.8: 119106879-119106879 on Build GRCh38
rs908721747 Chr.8: 119106733-119106733 on Build GRCh38
rs951692504 Chr.8: 119106474-119106474 on Build GRCh38
rs937042541 Chr.8: 119106763-119106763 on Build GRCh38
rs912556561 Chr.8: 119106388-119106388 on Build GRCh38
rs1041998784 Chr.8: 119106806-119106806 on Build GRCh38
rs981160714 Chr.8: 119106737-119106737 on Build GRCh38
rs542351790 Chr.8: 119106840-119106840 on Build GRCh38
rs752971357 Chr.8: 119106536-119106536 on Build GRCh38
rs761111129 Chr.8: 119106427-119106428 on Build GRCh38
rs556525815 Chr.8: 119106525-119106525 on Build GRCh38
rs759458302 Chr.8: 119106509-119106509 on Build GRCh38
rs906897100 Chr.8: 119106502-119106502 on Build GRCh38
rs907639066 Chr.8: 119106466-119106466 on Build GRCh38
rs1007357488 Chr.8: 119106368-119106368 on Build GRCh38
rs983383188 Chr.8: 119106699-119106699 on Build GRCh38
rs544897227 Chr.8: 119106445-119106445 on Build GRCh38
rs931507336 Chr.8: 119106420-119106420 on Build GRCh38
rs758212293 Chr.8: 119106558-119106560 on Build GRCh38
rs1011374986 Chr.8: 119106363-119106363 on Build GRCh38
rs1055387663 Chr.8: 119106767-119106767 on Build GRCh38
rs772560276 Chr.8: 119106383-119106383 on Build GRCh38
rs564451937 Chr.8: 119106754-119106754 on Build GRCh38
rs936615139 Chr.8: 119106722-119106722 on Build GRCh38
rs903017857 Chr.8: 119106844-119106844 on Build GRCh38
rs929244435 Chr.8: 119106764-119106765 on Build GRCh38
rs987230528 Chr.8: 119106387-119106387 on Build GRCh38
rs940438293 Chr.8: 119106475-119106475 on Build GRCh38
rs1019669454 Chr.8: 119106629-119106629 on Build GRCh38
rs535589887 Chr.8: 119106397-119106397 on Build GRCh38
rs1045429353 Chr.8: 119106498-119106498 on Build GRCh38
rs548407754 Chr.8: 119106756-119106756 on Build GRCh38
rs556531189 Chr.8: 119106482-119106482 on Build GRCh38
rs545652463 Chr.8: 119106687-119106687 on Build GRCh38
rs375182911 Chr.8: 119106696-119106696 on Build GRCh38
rs993356031 Chr.8: 119106398-119106398 on Build GRCh38
rs964045282 Chr.8: 119106632-119106632 on Build GRCh38
rs758779378 Chr.8: 119106876-119106876 on Build GRCh38
rs1027279428 Chr.8: 119106689-119106689 on Build GRCh38
rs903297180 Chr.8: 119106539-119106539 on Build GRCh38

View Full Product Details