Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143048962-143049422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf31
Entrez Gene ID
286122
Gene Name
chromosome 8 open reading frame 31
Gene Aliases
-
UniGene
Hs.660382
Gene Chromosome Location
Chr.8: 143038983-143054303 on Build GRCh38
Transcripts
NM_173687.3 NR_126398.1 NR_126399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6857313 c.372C>T p.H124H Chr.8: 143049225-143049225 on Build GRCh38
300363 c.364C>A p.R122R Chr.8: 143049217-143049217 on Build GRCh38
4668029 c.365G>A p.R122Q Chr.8: 143049218-143049218 on Build GRCh38
7178182 c.385C>T p.L129F Chr.8: 143049238-143049238 on Build GRCh38
1096900 c.385C>A p.L129I Chr.8: 143049238-143049238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959296550 Chr.8: 143049112-143049112 on Build GRCh38
rs777798342 Chr.8: 143049283-143049283 on Build GRCh38
rs758079925 Chr.8: 143049281-143049281 on Build GRCh38
rs113115637 Chr.8: 143049048-143049048 on Build GRCh38
rs769817525 Chr.8: 143049098-143049098 on Build GRCh38
rs181426288 Chr.8: 143049075-143049075 on Build GRCh38
rs780250066 Chr.8: 143049268-143049268 on Build GRCh38
rs746961115 Chr.8: 143049288-143049288 on Build GRCh38
rs1054096185 Chr.8: 143048981-143048981 on Build GRCh38
rs776252376 Chr.8: 143049159-143049159 on Build GRCh38
rs568264255 Chr.8: 143049319-143049319 on Build GRCh38
rs1020544567 Chr.8: 143049334-143049337 on Build GRCh38
rs1001722083 Chr.8: 143049012-143049012 on Build GRCh38
rs370543040 Chr.8: 143049292-143049292 on Build GRCh38
rs942753587 Chr.8: 143049068-143049068 on Build GRCh38
rs144856660 Chr.8: 143049098-143049098 on Build GRCh38
rs199938354 Chr.8: 143049203-143049203 on Build GRCh38
rs759313442 Chr.8: 143049187-143049187 on Build GRCh38
rs993105721 Chr.8: 143049033-143049033 on Build GRCh38
rs963998394 Chr.8: 143049379-143049379 on Build GRCh38
rs982256233 Chr.8: 143049097-143049097 on Build GRCh38
rs967993030 Chr.8: 143049343-143049343 on Build GRCh38
rs543650035 Chr.8: 143049053-143049053 on Build GRCh38
rs532702098 Chr.8: 143049370-143049370 on Build GRCh38
rs1014424543 Chr.8: 143049311-143049311 on Build GRCh38
rs1024457632 Chr.8: 143049335-143049335 on Build GRCh38
rs750478291 Chr.8: 143049253-143049253 on Build GRCh38
rs756384895 Chr.8: 143049258-143049258 on Build GRCh38
rs767623017 Chr.8: 143049234-143049234 on Build GRCh38
rs148501467 Chr.8: 143049401-143049401 on Build GRCh38
rs765886348 Chr.8: 143049279-143049289 on Build GRCh38
rs755579846 Chr.8: 143048992-143048992 on Build GRCh38
rs915171541 Chr.8: 143049131-143049131 on Build GRCh38
rs201472584 Chr.8: 143049225-143049225 on Build GRCh38
rs1024405055 Chr.8: 143049084-143049084 on Build GRCh38
rs78140118 Chr.8: 143049226-143049226 on Build GRCh38
rs1005840131 Chr.8: 143049052-143049052 on Build GRCh38
rs975492668 Chr.8: 143049400-143049400 on Build GRCh38
rs959121467 Chr.8: 143049398-143049398 on Build GRCh38
rs764671402 Chr.8: 143049218-143049218 on Build GRCh38
rs550010481 Chr.8: 143049344-143049344 on Build GRCh38
rs373633502 Chr.8: 143049293-143049293 on Build GRCh38
rs12677840 Chr.8: 143049013-143049013 on Build GRCh38
rs753216673 Chr.8: 143049281-143049281 on Build GRCh38
rs1042256197 Chr.8: 143049150-143049150 on Build GRCh38
rs74334668 Chr.8: 143049093-143049093 on Build GRCh38
rs778186056 Chr.8: 143049310-143049310 on Build GRCh38
rs745370723 Chr.8: 143049299-143049299 on Build GRCh38
rs776068003 Chr.8: 143049198-143049198 on Build GRCh38
rs747949959 Chr.8: 143049279-143049279 on Build GRCh38
rs761677097 Chr.8: 143049189-143049189 on Build GRCh38
rs989434284 Chr.8: 143049128-143049128 on Build GRCh38
rs565656305 Chr.8: 143048993-143048993 on Build GRCh38
rs576007127 Chr.8: 143049049-143049049 on Build GRCh38
rs994299321 Chr.8: 143049070-143049070 on Build GRCh38
rs558773482 Chr.8: 143049067-143049067 on Build GRCh38
rs202123049 Chr.8: 143049214-143049214 on Build GRCh38
rs774281830 Chr.8: 143049260-143049260 on Build GRCh38
rs901274309 Chr.8: 143049017-143049017 on Build GRCh38
rs767514261 Chr.8: 143049030-143049030 on Build GRCh38
rs945185320 Chr.8: 143048998-143048998 on Build GRCh38
rs571486089 Chr.8: 143049368-143049368 on Build GRCh38
rs778245286 Chr.8: 143049217-143049217 on Build GRCh38
rs541307934 Chr.8: 143049085-143049085 on Build GRCh38
rs759922660 Chr.8: 143049202-143049203 on Build GRCh38
rs12677862 Chr.8: 143049133-143049133 on Build GRCh38
rs149427784 Chr.8: 143048991-143048991 on Build GRCh38
rs1050268149 Chr.8: 143049238-143049238 on Build GRCh38
rs754099552 Chr.8: 143049360-143049360 on Build GRCh38
rs764972621 Chr.8: 143049196-143049196 on Build GRCh38
rs879552033 Chr.8: 143049390-143049390 on Build GRCh38

View Full Product Details