Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175010132-175010717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
UniGene
Hs.508524
Gene Chromosome Location
Chr.1: 174999435-175012027 on Build GRCh38
Transcripts
NM_001007214.1 NM_014412.2 XM_017001046.1

SNP Detail

SNP ID Chromosome Location
rs10912866 Chr.1: 175010590-175010590 on Build GRCh38
rs1018022320 Chr.1: 175010542-175010542 on Build GRCh38
rs768529349 Chr.1: 175010592-175010592 on Build GRCh38
rs41266072 Chr.1: 175010499-175010499 on Build GRCh38
rs995651675 Chr.1: 175010215-175010215 on Build GRCh38
rs75288526 Chr.1: 175010631-175010631 on Build GRCh38
rs897403253 Chr.1: 175010374-175010374 on Build GRCh38
rs867847388 Chr.1: 175010663-175010663 on Build GRCh38
rs564673192 Chr.1: 175010655-175010655 on Build GRCh38
rs956259247 Chr.1: 175010598-175010598 on Build GRCh38
rs898137252 Chr.1: 175010486-175010486 on Build GRCh38
rs182017817 Chr.1: 175010648-175010648 on Build GRCh38
rs760271446 Chr.1: 175010560-175010561 on Build GRCh38
rs1050890604 Chr.1: 175010370-175010370 on Build GRCh38
rs1003845342 Chr.1: 175010171-175010171 on Build GRCh38
rs41266074 Chr.1: 175010612-175010612 on Build GRCh38
rs368519076 Chr.1: 175010677-175010678 on Build GRCh38
rs868841445 Chr.1: 175010318-175010318 on Build GRCh38
rs192630577 Chr.1: 175010255-175010255 on Build GRCh38
rs371438564 Chr.1: 175010694-175010697 on Build GRCh38
rs775628491 Chr.1: 175010523-175010523 on Build GRCh38
rs563817162 Chr.1: 175010247-175010247 on Build GRCh38
rs745376769 Chr.1: 175010333-175010333 on Build GRCh38
rs918550846 Chr.1: 175010637-175010637 on Build GRCh38
rs990893886 Chr.1: 175010621-175010622 on Build GRCh38
rs928832809 Chr.1: 175010291-175010291 on Build GRCh38
rs1021894822 Chr.1: 175010615-175010615 on Build GRCh38
rs1039504193 Chr.1: 175010354-175010354 on Build GRCh38
rs866301179 Chr.1: 175010210-175010210 on Build GRCh38
rs12271 Chr.1: 175010622-175010622 on Build GRCh38
rs578111557 Chr.1: 175010689-175010689 on Build GRCh38
rs10912865 Chr.1: 175010428-175010428 on Build GRCh38
rs537675471 Chr.1: 175010582-175010582 on Build GRCh38
rs866709557 Chr.1: 175010245-175010245 on Build GRCh38
rs922035554 Chr.1: 175010365-175010365 on Build GRCh38
rs376303356 Chr.1: 175010492-175010492 on Build GRCh38
rs1022745746 Chr.1: 175010224-175010224 on Build GRCh38
rs184477070 Chr.1: 175010583-175010583 on Build GRCh38
rs552179957 Chr.1: 175010334-175010334 on Build GRCh38
rs568514178 Chr.1: 175010565-175010565 on Build GRCh38
rs1015675680 Chr.1: 175010187-175010187 on Build GRCh38
rs767634873 Chr.1: 175010656-175010656 on Build GRCh38
rs962441133 Chr.1: 175010208-175010208 on Build GRCh38
rs914542752 Chr.1: 175010316-175010316 on Build GRCh38
rs747506390 Chr.1: 175010676-175010677 on Build GRCh38
rs981613633 Chr.1: 175010284-175010284 on Build GRCh38
rs939925018 Chr.1: 175010399-175010399 on Build GRCh38
rs930172449 Chr.1: 175010376-175010376 on Build GRCh38
rs989108637 Chr.1: 175010298-175010298 on Build GRCh38
rs150725735 Chr.1: 175010369-175010369 on Build GRCh38
rs941398562 Chr.1: 175010630-175010633 on Build GRCh38
rs956273355 Chr.1: 175010294-175010294 on Build GRCh38
rs1051275678 Chr.1: 175010377-175010377 on Build GRCh38
rs772967282 Chr.1: 175010664-175010664 on Build GRCh38
rs892815061 Chr.1: 175010568-175010568 on Build GRCh38
rs891000020 Chr.1: 175010398-175010398 on Build GRCh38

View Full Product Details