Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231223558-231224098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf131
Entrez Gene ID
128061
Gene Name
chromosome 1 open reading frame 131
Gene Aliases
-
UniGene
Hs.743464
Gene Chromosome Location
Chr.1: 231223763-231241187 on Build GRCh38
Transcripts
NM_001300830.1 NM_152379.3

SNP Detail

SNP ID Chromosome Location
rs113465621 Chr.1: 231224031-231224031 on Build GRCh38
rs918558356 Chr.1: 231223989-231223989 on Build GRCh38
rs545624558 Chr.1: 231223743-231223743 on Build GRCh38
rs1050998475 Chr.1: 231223698-231223698 on Build GRCh38
rs572172002 Chr.1: 231223685-231223685 on Build GRCh38
rs943219850 Chr.1: 231223614-231223614 on Build GRCh38
rs1017076221 Chr.1: 231223939-231223939 on Build GRCh38
rs957364326 Chr.1: 231223789-231223789 on Build GRCh38
rs142020572 Chr.1: 231223763-231223763 on Build GRCh38
rs973237571 Chr.1: 231223987-231223987 on Build GRCh38
rs950977799 Chr.1: 231223964-231223964 on Build GRCh38
rs759567598 Chr.1: 231223746-231223746 on Build GRCh38
rs1038781791 Chr.1: 231223639-231223639 on Build GRCh38
rs188032468 Chr.1: 231223687-231223687 on Build GRCh38
rs962740756 Chr.1: 231223960-231223960 on Build GRCh38
rs16854004 Chr.1: 231223673-231223673 on Build GRCh38
rs150609944 Chr.1: 231223858-231223858 on Build GRCh38
rs890293292 Chr.1: 231223610-231223610 on Build GRCh38
rs778243556 Chr.1: 231223909-231223912 on Build GRCh38
rs911843694 Chr.1: 231224053-231224053 on Build GRCh38
rs191777258 Chr.1: 231224036-231224036 on Build GRCh38
rs139711958 Chr.1: 231223941-231223941 on Build GRCh38
rs984043291 Chr.1: 231224005-231224005 on Build GRCh38
rs541470324 Chr.1: 231223654-231223654 on Build GRCh38
rs891030934 Chr.1: 231223705-231223705 on Build GRCh38
rs116260611 Chr.1: 231223915-231223915 on Build GRCh38
rs1035269353 Chr.1: 231223861-231223861 on Build GRCh38
rs769869959 Chr.1: 231223765-231223765 on Build GRCh38
rs1039459104 Chr.1: 231223658-231223658 on Build GRCh38
rs527396043 Chr.1: 231224008-231224008 on Build GRCh38
rs955600140 Chr.1: 231224009-231224009 on Build GRCh38
rs763133655 Chr.1: 231224061-231224061 on Build GRCh38
rs959517873 Chr.1: 231223875-231223875 on Build GRCh38
rs542546873 Chr.1: 231223747-231223747 on Build GRCh38
rs997470388 Chr.1: 231223682-231223682 on Build GRCh38
rs981094265 Chr.1: 231223953-231223953 on Build GRCh38
rs928898476 Chr.1: 231223954-231223954 on Build GRCh38

View Full Product Details