Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 52314737-52315137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK1
Entrez Gene ID
22943
Gene Name
dickkopf WNT signaling pathway inhibitor 1
Gene Aliases
DKK-1, SK
UniGene
Hs.40499
Gene Chromosome Location
Chr.10: 52314281-52317657 on Build GRCh38
Transcripts
NM_012242.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3747196 c.386C>T p.P129L Chr.10: 52315065-52315065 on Build GRCh38
4981467 c.265G>A p.E89K Chr.10: 52314944-52314944 on Build GRCh38
3438687 c.390G>A p.G130G Chr.10: 52315069-52315069 on Build GRCh38
918859 c.352C>T p.R118* Chr.10: 52315031-52315031 on Build GRCh38
1284503 c.300C>A p.P100P Chr.10: 52314979-52314979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576773139 Chr.10: 52314986-52314986 on Build GRCh38
rs755249211 Chr.10: 52314789-52314789 on Build GRCh38
rs770954476 Chr.10: 52314952-52314952 on Build GRCh38
rs41281546 Chr.10: 52314900-52314900 on Build GRCh38
rs939796051 Chr.10: 52314828-52314828 on Build GRCh38
rs779008070 Chr.10: 52315114-52315114 on Build GRCh38
rs753117217 Chr.10: 52315075-52315075 on Build GRCh38
rs1037540371 Chr.10: 52314883-52314883 on Build GRCh38
rs113404907 Chr.10: 52314826-52314826 on Build GRCh38
rs988182645 Chr.10: 52314782-52314782 on Build GRCh38
rs754564931 Chr.10: 52314923-52314923 on Build GRCh38
rs921532650 Chr.10: 52314765-52314765 on Build GRCh38
rs765280492 Chr.10: 52315000-52315000 on Build GRCh38
rs1019730155 Chr.10: 52314935-52314935 on Build GRCh38
rs576196926 Chr.10: 52315001-52315001 on Build GRCh38
rs770445280 Chr.10: 52315052-52315052 on Build GRCh38
rs996936785 Chr.10: 52314963-52314963 on Build GRCh38
rs778192698 Chr.10: 52314944-52314944 on Build GRCh38
rs149268042 Chr.10: 52315038-52315038 on Build GRCh38
rs1008681185 Chr.10: 52314927-52314927 on Build GRCh38
rs112183005 Chr.10: 52314917-52314917 on Build GRCh38
rs145409518 Chr.10: 52314919-52314919 on Build GRCh38
rs201617558 Chr.10: 52314930-52314930 on Build GRCh38
rs145325880 Chr.10: 52314866-52314866 on Build GRCh38
rs548841178 Chr.10: 52314861-52314861 on Build GRCh38
rs773217525 Chr.10: 52315015-52315015 on Build GRCh38
rs746154609 Chr.10: 52314971-52314971 on Build GRCh38
rs749370700 Chr.10: 52314948-52314948 on Build GRCh38
rs112701910 Chr.10: 52314842-52314842 on Build GRCh38
rs757923992 Chr.10: 52314907-52314907 on Build GRCh38
rs780678889 Chr.10: 52314991-52314991 on Build GRCh38
rs965416520 Chr.10: 52314956-52314956 on Build GRCh38
rs1032486630 Chr.10: 52314975-52314975 on Build GRCh38
rs138915253 Chr.10: 52315037-52315037 on Build GRCh38
rs375857052 Chr.10: 52315093-52315093 on Build GRCh38
rs571172078 Chr.10: 52315103-52315103 on Build GRCh38
rs372651276 Chr.10: 52315016-52315016 on Build GRCh38
rs146946503 Chr.10: 52315040-52315040 on Build GRCh38
rs533327602 Chr.10: 52314837-52314837 on Build GRCh38
rs756399342 Chr.10: 52315082-52315082 on Build GRCh38
rs765263687 Chr.10: 52314901-52314901 on Build GRCh38
rs766034631 Chr.10: 52315022-52315022 on Build GRCh38
rs553998785 Chr.10: 52314970-52314970 on Build GRCh38
rs533863517 Chr.10: 52314924-52314924 on Build GRCh38
rs772156603 Chr.10: 52314994-52314994 on Build GRCh38
rs377469990 Chr.10: 52314983-52314983 on Build GRCh38
rs926559519 Chr.10: 52314862-52314862 on Build GRCh38
rs375574588 Chr.10: 52315083-52315083 on Build GRCh38
rs913585030 Chr.10: 52314787-52314787 on Build GRCh38
rs779857735 Chr.10: 52314908-52314908 on Build GRCh38
rs987275148 Chr.10: 52314934-52314934 on Build GRCh38
rs138015066 Chr.10: 52315046-52315046 on Build GRCh38
rs141115379 Chr.10: 52314995-52314995 on Build GRCh38
rs201074100 Chr.10: 52315051-52315051 on Build GRCh38
rs931525177 Chr.10: 52314772-52314772 on Build GRCh38
rs760422268 Chr.10: 52314890-52314890 on Build GRCh38
rs753555582 Chr.10: 52314895-52314895 on Build GRCh38
rs887269895 Chr.10: 52314802-52314802 on Build GRCh38
rs750013290 Chr.10: 52314904-52314904 on Build GRCh38
rs537688792 Chr.10: 52314892-52314892 on Build GRCh38
rs2241529 Chr.10: 52314997-52314997 on Build GRCh38
rs749750815 Chr.10: 52315084-52315084 on Build GRCh38

View Full Product Details