Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84371594-84372135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCSER2
Entrez Gene ID
54462
Gene Name
coiled-coil serine rich protein 2
Gene Aliases
FAM190B, Gcap14, KIAA1128, bA486O22.1
UniGene
Hs.461988
Gene Chromosome Location
Chr.10: 84328558-84518521 on Build GRCh38
Transcripts
NM_001284240.1 NM_001284241.1 NM_018999.3 XM_005269905.3 XM_005269906.3 XM_011539870.2 XM_011539871.2 XM_011539872.2 XM_017016340.1 XM_017016341.1 XM_017016342.1 XM_017016343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3867973 c.729C>T p.S243S Chr.10: 84371781-84371781 on Build GRCh38
5520615 c.897G>A p.K299K Chr.10: 84371949-84371949 on Build GRCh38
7209151 c.1079C>T p.A360V Chr.10: 84372131-84372131 on Build GRCh38
226233 c.859G>T p.G287W Chr.10: 84371911-84371911 on Build GRCh38
108901 c.1017T>A p.N339K Chr.10: 84372069-84372069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143121348 Chr.10: 84372078-84372078 on Build GRCh38
rs982056577 Chr.10: 84372013-84372013 on Build GRCh38
rs754027105 Chr.10: 84371764-84371764 on Build GRCh38
rs759648396 Chr.10: 84372079-84372079 on Build GRCh38
rs146103957 Chr.10: 84371811-84371811 on Build GRCh38
rs769241896 Chr.10: 84371849-84371849 on Build GRCh38
rs761258134 Chr.10: 84371906-84371906 on Build GRCh38
rs759286087 Chr.10: 84371762-84371762 on Build GRCh38
rs145873640 Chr.10: 84372069-84372069 on Build GRCh38
rs966374558 Chr.10: 84371978-84371978 on Build GRCh38
rs759771564 Chr.10: 84371826-84371826 on Build GRCh38
rs757348605 Chr.10: 84371766-84371766 on Build GRCh38
rs773546405 Chr.10: 84372051-84372051 on Build GRCh38
rs747623189 Chr.10: 84371709-84371709 on Build GRCh38
rs184963489 Chr.10: 84371859-84371859 on Build GRCh38
rs779293524 Chr.10: 84371646-84371646 on Build GRCh38
rs935566156 Chr.10: 84372018-84372018 on Build GRCh38
rs752165991 Chr.10: 84371981-84371981 on Build GRCh38
rs755759478 Chr.10: 84371903-84371903 on Build GRCh38
rs756974159 Chr.10: 84371924-84371924 on Build GRCh38
rs922894847 Chr.10: 84371630-84371630 on Build GRCh38
rs753186549 Chr.10: 84371876-84371876 on Build GRCh38
rs757305301 Chr.10: 84371645-84371645 on Build GRCh38
rs936205005 Chr.10: 84371717-84371717 on Build GRCh38
rs375626713 Chr.10: 84371672-84371672 on Build GRCh38
rs899559084 Chr.10: 84372091-84372091 on Build GRCh38
rs769278323 Chr.10: 84371712-84371712 on Build GRCh38
rs755457843 Chr.10: 84371894-84371894 on Build GRCh38
rs770080746 Chr.10: 84371940-84371940 on Build GRCh38
rs374232381 Chr.10: 84371895-84371895 on Build GRCh38
rs769080829 Chr.10: 84371934-84371934 on Build GRCh38
rs774464580 Chr.10: 84372090-84372091 on Build GRCh38
rs150621117 Chr.10: 84371915-84371915 on Build GRCh38
rs371027225 Chr.10: 84371614-84371614 on Build GRCh38
rs867346786 Chr.10: 84371929-84371929 on Build GRCh38
rs752043393 Chr.10: 84371890-84371890 on Build GRCh38
rs749223409 Chr.10: 84371856-84371856 on Build GRCh38
rs61745697 Chr.10: 84371622-84371622 on Build GRCh38
rs574318262 Chr.10: 84371987-84371987 on Build GRCh38
rs763753140 Chr.10: 84371914-84371914 on Build GRCh38
rs924849888 Chr.10: 84371715-84371715 on Build GRCh38
rs758173787 Chr.10: 84372070-84372070 on Build GRCh38
rs534278497 Chr.10: 84371887-84371887 on Build GRCh38
rs201862257 Chr.10: 84371621-84371621 on Build GRCh38
rs559024343 Chr.10: 84371695-84371695 on Build GRCh38
rs750941263 Chr.10: 84371789-84371789 on Build GRCh38
rs746383313 Chr.10: 84371649-84371649 on Build GRCh38
rs773252798 Chr.10: 84371944-84371944 on Build GRCh38
rs546947601 Chr.10: 84371685-84371685 on Build GRCh38
rs922979517 Chr.10: 84371973-84371973 on Build GRCh38
rs376657376 Chr.10: 84371867-84371869 on Build GRCh38
rs552393075 Chr.10: 84371760-84371760 on Build GRCh38
rs766559986 Chr.10: 84371965-84371965 on Build GRCh38
rs371702914 Chr.10: 84371862-84371862 on Build GRCh38
rs139772636 Chr.10: 84371875-84371875 on Build GRCh38
rs745793810 Chr.10: 84371917-84371917 on Build GRCh38
rs373732128 Chr.10: 84371850-84371850 on Build GRCh38
rs758506881 Chr.10: 84371889-84371889 on Build GRCh38
rs145634898 Chr.10: 84371986-84371986 on Build GRCh38
rs773087258 Chr.10: 84371732-84371732 on Build GRCh38
rs991306210 Chr.10: 84371972-84371972 on Build GRCh38
rs760094623 Chr.10: 84371983-84371983 on Build GRCh38
rs372276917 Chr.10: 84372027-84372027 on Build GRCh38
rs771179017 Chr.10: 84372068-84372068 on Build GRCh38
rs200588575 Chr.10: 84372052-84372052 on Build GRCh38
rs1055026573 Chr.10: 84371738-84371738 on Build GRCh38
rs756819853 Chr.10: 84372022-84372022 on Build GRCh38
rs556728657 Chr.10: 84372031-84372031 on Build GRCh38
rs879247925 Chr.10: 84371718-84371718 on Build GRCh38
rs768090857 Chr.10: 84372084-84372084 on Build GRCh38
rs76747767 Chr.10: 84371990-84371990 on Build GRCh38
rs907272809 Chr.10: 84372108-84372108 on Build GRCh38
rs747402875 Chr.10: 84371842-84371842 on Build GRCh38
rs752338113 Chr.10: 84371898-84371898 on Build GRCh38
rs951289872 Chr.10: 84371886-84371886 on Build GRCh38
rs376135346 Chr.10: 84371756-84371756 on Build GRCh38
rs367890058 Chr.10: 84371861-84371861 on Build GRCh38
rs375883422 Chr.10: 84371848-84371848 on Build GRCh38
rs753086124 Chr.10: 84372090-84372090 on Build GRCh38
rs762931084 Chr.10: 84371734-84371734 on Build GRCh38
rs144671442 Chr.10: 84371936-84371936 on Build GRCh38
rs142860601 Chr.10: 84371768-84371768 on Build GRCh38
rs770884931 Chr.10: 84371857-84371857 on Build GRCh38
rs750600804 Chr.10: 84371615-84371615 on Build GRCh38
rs777493764 Chr.10: 84371998-84371998 on Build GRCh38
rs146005806 Chr.10: 84371689-84371689 on Build GRCh38
rs780548735 Chr.10: 84371676-84371676 on Build GRCh38
rs750550458 Chr.10: 84371880-84371880 on Build GRCh38
rs753486357 Chr.10: 84372014-84372014 on Build GRCh38
rs997937112 Chr.10: 84372107-84372107 on Build GRCh38
rs750250954 Chr.10: 84372032-84372032 on Build GRCh38
rs778372736 Chr.10: 84372024-84372024 on Build GRCh38
rs148590499 Chr.10: 84371947-84371947 on Build GRCh38
rs746837144 Chr.10: 84372034-84372034 on Build GRCh38
rs760350677 Chr.10: 84371938-84371938 on Build GRCh38
rs755333378 Chr.10: 84371844-84371844 on Build GRCh38
rs976208006 Chr.10: 84371943-84371943 on Build GRCh38
rs376847515 Chr.10: 84372048-84372048 on Build GRCh38
rs753893589 Chr.10: 84371625-84371625 on Build GRCh38
rs780671431 Chr.10: 84371827-84371827 on Build GRCh38
rs768458114 Chr.10: 84371699-84371699 on Build GRCh38
rs373202985 Chr.10: 84371772-84371772 on Build GRCh38
rs138631845 Chr.10: 84371705-84371705 on Build GRCh38
rs766154480 Chr.10: 84371620-84371620 on Build GRCh38
rs748816526 Chr.10: 84371908-84371908 on Build GRCh38

View Full Product Details