Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 77792131-77792675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG5
Entrez Gene ID
9231
Gene Name
discs large MAGUK scaffold protein 5
Gene Aliases
LP-DLG, P-DLG5, PDLG
UniGene
Hs.652690
Gene Chromosome Location
Chr.10: 77790787-77926590 on Build GRCh38
Transcripts
NM_004747.3 XM_005270276.4 XM_006718056.3 XM_011540341.2 XM_011540342.1 XM_011540344.2 XM_011540345.1 XM_011540347.2 XM_017016913.1 XM_017016914.1 XM_017016915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4834267 c.5712C>T p.V1904V Chr.10: 77792488-77792488 on Build GRCh38
4655508 c.5679G>A p.L1893L Chr.10: 77792521-77792521 on Build GRCh38
685268 c.5690G>T p.C1897F Chr.10: 77792510-77792510 on Build GRCh38
1725147 c.5728A>G p.K1910E Chr.10: 77792472-77792472 on Build GRCh38
5495688 c.5750G>T p.C1917F Chr.10: 77792450-77792450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141127758 Chr.10: 77792448-77792448 on Build GRCh38
rs140994758 Chr.10: 77792490-77792490 on Build GRCh38
rs371607023 Chr.10: 77792567-77792567 on Build GRCh38
rs1055711823 Chr.10: 77792371-77792371 on Build GRCh38
rs775833643 Chr.10: 77792512-77792512 on Build GRCh38
rs549332883 Chr.10: 77792237-77792237 on Build GRCh38
rs750273596 Chr.10: 77792543-77792543 on Build GRCh38
rs547469332 Chr.10: 77792228-77792228 on Build GRCh38
rs756383324 Chr.10: 77792566-77792566 on Build GRCh38
rs779992597 Chr.10: 77792552-77792552 on Build GRCh38
rs547130830 Chr.10: 77792277-77792277 on Build GRCh38
rs376367695 Chr.10: 77792410-77792410 on Build GRCh38
rs372910799 Chr.10: 77792242-77792242 on Build GRCh38
rs557294958 Chr.10: 77792616-77792616 on Build GRCh38
rs774679331 Chr.10: 77792491-77792491 on Build GRCh38
rs928401594 Chr.10: 77792629-77792629 on Build GRCh38
rs1011441825 Chr.10: 77792193-77792193 on Build GRCh38
rs957350577 Chr.10: 77792281-77792281 on Build GRCh38
rs990189299 Chr.10: 77792292-77792292 on Build GRCh38
rs760312523 Chr.10: 77792514-77792514 on Build GRCh38
rs919655234 Chr.10: 77792322-77792322 on Build GRCh38
rs780105288 Chr.10: 77792344-77792344 on Build GRCh38
rs746334522 Chr.10: 77792621-77792621 on Build GRCh38
rs144148304 Chr.10: 77792493-77792493 on Build GRCh38
rs374537076 Chr.10: 77792405-77792405 on Build GRCh38
rs937176200 Chr.10: 77792370-77792370 on Build GRCh38
rs529006149 Chr.10: 77792185-77792185 on Build GRCh38
rs573928320 Chr.10: 77792638-77792638 on Build GRCh38
rs778759674 Chr.10: 77792447-77792447 on Build GRCh38
rs1038559485 Chr.10: 77792313-77792313 on Build GRCh38
rs968515147 Chr.10: 77792417-77792417 on Build GRCh38
rs942870934 Chr.10: 77792278-77792278 on Build GRCh38
rs755775088 Chr.10: 77792426-77792426 on Build GRCh38
rs374768246 Chr.10: 77792229-77792229 on Build GRCh38
rs139909783 Chr.10: 77792542-77792542 on Build GRCh38
rs1034145039 Chr.10: 77792180-77792180 on Build GRCh38
rs964353952 Chr.10: 77792515-77792515 on Build GRCh38
rs1058202 Chr.10: 77792393-77792393 on Build GRCh38
rs779062355 Chr.10: 77792582-77792582 on Build GRCh38
rs184189472 Chr.10: 77792554-77792554 on Build GRCh38
rs763791040 Chr.10: 77792525-77792525 on Build GRCh38
rs987023642 Chr.10: 77792272-77792272 on Build GRCh38
rs149868054 Chr.10: 77792194-77792194 on Build GRCh38
rs543852917 Chr.10: 77792605-77792605 on Build GRCh38
rs2165046 Chr.10: 77792334-77792334 on Build GRCh38
rs148200124 Chr.10: 77792446-77792446 on Build GRCh38
rs1036059687 Chr.10: 77792171-77792171 on Build GRCh38
rs150800568 Chr.10: 77792469-77792469 on Build GRCh38
rs749429394 Chr.10: 77792486-77792486 on Build GRCh38
rs1058204 Chr.10: 77792253-77792253 on Build GRCh38
rs955369286 Chr.10: 77792466-77792466 on Build GRCh38
rs991319010 Chr.10: 77792626-77792626 on Build GRCh38
rs748316545 Chr.10: 77792456-77792456 on Build GRCh38
rs968190124 Chr.10: 77792214-77792214 on Build GRCh38
rs758362626 Chr.10: 77792550-77792550 on Build GRCh38
rs938325053 Chr.10: 77792647-77792647 on Build GRCh38
rs184653394 Chr.10: 77792406-77792406 on Build GRCh38
rs955740397 Chr.10: 77792238-77792238 on Build GRCh38
rs780010962 Chr.10: 77792453-77792453 on Build GRCh38
rs56318751 Chr.10: 77792394-77792394 on Build GRCh38
rs1058203 Chr.10: 77792349-77792349 on Build GRCh38
rs958556184 Chr.10: 77792619-77792619 on Build GRCh38
rs146009016 Chr.10: 77792496-77792496 on Build GRCh38
rs753319002 Chr.10: 77792541-77792541 on Build GRCh38
rs772265186 Chr.10: 77792498-77792498 on Build GRCh38
rs770866878 Chr.10: 77792571-77792571 on Build GRCh38
rs17516354 Chr.10: 77792268-77792268 on Build GRCh38
rs906868747 Chr.10: 77792335-77792335 on Build GRCh38
rs929753635 Chr.10: 77792328-77792328 on Build GRCh38
rs572242682 Chr.10: 77792476-77792476 on Build GRCh38
rs1022798544 Chr.10: 77792202-77792202 on Build GRCh38

View Full Product Details