Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97677674-97678240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPI1
Entrez Gene ID
60370
Gene Name
arginine vasopressin induced 1
Gene Aliases
PP5395, VIP32, VIT32
UniGene
Hs.23918
Gene Chromosome Location
Chr.10: 97677422-97687432 on Build GRCh38
Transcripts
NM_021732.2 XM_017016494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1321962 c.388C>T p.R130C Chr.10: 97677925-97677925 on Build GRCh38
3720936 c.312G>A p.L104L Chr.10: 97678001-97678001 on Build GRCh38
1184270 c.389G>A p.R130H Chr.10: 97677924-97677924 on Build GRCh38
7132133 c.320C>A p.P107Q Chr.10: 97677993-97677993 on Build GRCh38
7095506 c.305C>T p.S102F Chr.10: 97678008-97678008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745481980 Chr.10: 97677941-97677941 on Build GRCh38
rs1036984423 Chr.10: 97677800-97677800 on Build GRCh38
rs139124316 Chr.10: 97677896-97677896 on Build GRCh38
rs142341567 Chr.10: 97677897-97677897 on Build GRCh38
rs373092990 Chr.10: 97678095-97678095 on Build GRCh38
rs770928307 Chr.10: 97678070-97678070 on Build GRCh38
rs992495267 Chr.10: 97677717-97677717 on Build GRCh38
rs901848784 Chr.10: 97678100-97678100 on Build GRCh38
rs997638187 Chr.10: 97677824-97677824 on Build GRCh38
rs941243490 Chr.10: 97677984-97677984 on Build GRCh38
rs904050965 Chr.10: 97677890-97677890 on Build GRCh38
rs537218547 Chr.10: 97677922-97677922 on Build GRCh38
rs757001542 Chr.10: 97677939-97677939 on Build GRCh38
rs146380012 Chr.10: 97678094-97678094 on Build GRCh38
rs777533659 Chr.10: 97677844-97677844 on Build GRCh38
rs148513568 Chr.10: 97678118-97678118 on Build GRCh38
rs758002015 Chr.10: 97678060-97678060 on Build GRCh38
rs146189926 Chr.10: 97677895-97677895 on Build GRCh38
rs553651349 Chr.10: 97678008-97678008 on Build GRCh38
rs564595101 Chr.10: 97678122-97678122 on Build GRCh38
rs868250897 Chr.10: 97678156-97678156 on Build GRCh38
rs114167759 Chr.10: 97678129-97678129 on Build GRCh38
rs78694894 Chr.10: 97677894-97677894 on Build GRCh38
rs749718877 Chr.10: 97677979-97677979 on Build GRCh38
rs745652153 Chr.10: 97677963-97677983 on Build GRCh38
rs1017644036 Chr.10: 97677773-97677773 on Build GRCh38
rs760702581 Chr.10: 97678022-97678022 on Build GRCh38
rs367737358 Chr.10: 97677858-97677858 on Build GRCh38
rs777232895 Chr.10: 97677867-97677867 on Build GRCh38
rs749806834 Chr.10: 97678069-97678069 on Build GRCh38
rs375917724 Chr.10: 97677857-97677857 on Build GRCh38
rs754514820 Chr.10: 97678066-97678066 on Build GRCh38
rs991071978 Chr.10: 97678154-97678154 on Build GRCh38
rs948462563 Chr.10: 97677726-97677726 on Build GRCh38
rs916966163 Chr.10: 97677721-97677721 on Build GRCh38
rs546978561 Chr.10: 97677802-97677802 on Build GRCh38
rs200939072 Chr.10: 97677969-97677969 on Build GRCh38
rs955727583 Chr.10: 97677701-97677701 on Build GRCh38
rs539175341 Chr.10: 97677961-97677961 on Build GRCh38
rs970378832 Chr.10: 97678178-97678178 on Build GRCh38
rs374234934 Chr.10: 97677848-97677848 on Build GRCh38
rs959608891 Chr.10: 97678140-97678140 on Build GRCh38
rs756158266 Chr.10: 97678038-97678048 on Build GRCh38
rs374864315 Chr.10: 97678115-97678116 on Build GRCh38
rs535226445 Chr.10: 97677757-97677757 on Build GRCh38
rs774414600 Chr.10: 97677914-97677914 on Build GRCh38
rs563973464 Chr.10: 97678125-97678125 on Build GRCh38
rs759661969 Chr.10: 97678005-97678005 on Build GRCh38
rs909739228 Chr.10: 97677759-97677759 on Build GRCh38
rs764939476 Chr.10: 97678043-97678043 on Build GRCh38
rs373727438 Chr.10: 97677827-97677827 on Build GRCh38
rs775549165 Chr.10: 97678018-97678018 on Build GRCh38
rs3750565 Chr.10: 97678174-97678174 on Build GRCh38
rs753659845 Chr.10: 97677928-97677928 on Build GRCh38
rs778989445 Chr.10: 97677949-97677949 on Build GRCh38
rs367683380 Chr.10: 97677924-97677924 on Build GRCh38
rs753720850 Chr.10: 97677909-97677909 on Build GRCh38
rs889810034 Chr.10: 97678107-97678107 on Build GRCh38
rs139597558 Chr.10: 97677987-97677987 on Build GRCh38
rs942768343 Chr.10: 97678109-97678110 on Build GRCh38
rs200877398 Chr.10: 97677763-97677763 on Build GRCh38
rs191123799 Chr.10: 97678181-97678181 on Build GRCh38
rs568121112 Chr.10: 97677710-97677710 on Build GRCh38
rs146044000 Chr.10: 97677925-97677925 on Build GRCh38
rs771457766 Chr.10: 97677862-97677862 on Build GRCh38
rs565382622 Chr.10: 97677836-97677836 on Build GRCh38
rs746461379 Chr.10: 97677962-97677962 on Build GRCh38
rs776707429 Chr.10: 97678104-97678104 on Build GRCh38
rs186329018 Chr.10: 97678139-97678139 on Build GRCh38
rs902658741 Chr.10: 97678127-97678127 on Build GRCh38
rs372531998 Chr.10: 97678061-97678061 on Build GRCh38
rs748658146 Chr.10: 97677873-97677873 on Build GRCh38
rs767718318 Chr.10: 97678010-97678010 on Build GRCh38
rs901919010 Chr.10: 97677793-97677793 on Build GRCh38
rs376013843 Chr.10: 97677940-97677940 on Build GRCh38
rs757996345 Chr.10: 97677954-97677954 on Build GRCh38
rs112410295 Chr.10: 97677964-97677964 on Build GRCh38
rs774700831 Chr.10: 97677989-97677989 on Build GRCh38
rs752546906 Chr.10: 97677826-97677826 on Build GRCh38
rs757119007 Chr.10: 97678036-97678036 on Build GRCh38
rs376653134 Chr.10: 97678048-97678048 on Build GRCh38
rs753757621 Chr.10: 97678029-97678029 on Build GRCh38
rs3750564 Chr.10: 97678065-97678065 on Build GRCh38
rs778464017 Chr.10: 97678067-97678067 on Build GRCh38
rs374997584 Chr.10: 97677921-97677921 on Build GRCh38
rs781241141 Chr.10: 97677822-97677822 on Build GRCh38
rs770430421 Chr.10: 97677966-97677966 on Build GRCh38
rs764044933 Chr.10: 97678027-97678027 on Build GRCh38

View Full Product Details