Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22622184-22622723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCF
Entrez Gene ID
2188
Gene Name
Fanconi anemia complementation group F
Gene Aliases
FAF
UniGene
Hs.632151
Gene Chromosome Location
Chr.11: 22622533-22625841 on Build GRCh38
Transcripts
NM_022725.3

SNP Detail

SNP ID Chromosome Location
rs886048148 Chr.11: 22622615-22622615 on Build GRCh38
rs990289797 Chr.11: 22622320-22622320 on Build GRCh38
rs1036378563 Chr.11: 22622624-22622624 on Build GRCh38
rs1037558716 Chr.11: 22622252-22622252 on Build GRCh38
rs539449637 Chr.11: 22622532-22622532 on Build GRCh38
rs1040732375 Chr.11: 22622497-22622497 on Build GRCh38
rs1010180070 Chr.11: 22622446-22622446 on Build GRCh38
rs45625034 Chr.11: 22622563-22622563 on Build GRCh38
rs1007846974 Chr.11: 22622290-22622290 on Build GRCh38
rs755109538 Chr.11: 22622400-22622400 on Build GRCh38
rs1029652651 Chr.11: 22622452-22622452 on Build GRCh38
rs551248573 Chr.11: 22622516-22622516 on Build GRCh38
rs961639506 Chr.11: 22622634-22622634 on Build GRCh38
rs947817832 Chr.11: 22622353-22622353 on Build GRCh38
rs1052801519 Chr.11: 22622436-22622436 on Build GRCh38
rs891209892 Chr.11: 22622470-22622470 on Build GRCh38
rs955485230 Chr.11: 22622215-22622215 on Build GRCh38
rs1023066091 Chr.11: 22622339-22622339 on Build GRCh38
rs973159278 Chr.11: 22622639-22622639 on Build GRCh38
rs748374811 Chr.11: 22622482-22622482 on Build GRCh38
rs45615036 Chr.11: 22622302-22622302 on Build GRCh38
rs985639349 Chr.11: 22622261-22622261 on Build GRCh38
rs919884005 Chr.11: 22622656-22622656 on Build GRCh38
rs558888429 Chr.11: 22622484-22622484 on Build GRCh38
rs974739124 Chr.11: 22622291-22622291 on Build GRCh38
rs532935494 Chr.11: 22622260-22622260 on Build GRCh38
rs749668168 Chr.11: 22622535-22622535 on Build GRCh38
rs180967906 Chr.11: 22622531-22622531 on Build GRCh38
rs45617139 Chr.11: 22622287-22622287 on Build GRCh38
rs1056164057 Chr.11: 22622319-22622319 on Build GRCh38
rs1031860809 Chr.11: 22622303-22622306 on Build GRCh38
rs957105083 Chr.11: 22622313-22622313 on Build GRCh38
rs1048925607 Chr.11: 22622465-22622466 on Build GRCh38
rs576419372 Chr.11: 22622612-22622612 on Build GRCh38
rs780054478 Chr.11: 22622231-22622231 on Build GRCh38
rs757239828 Chr.11: 22622216-22622216 on Build GRCh38
rs940127885 Chr.11: 22622245-22622245 on Build GRCh38
rs974326707 Chr.11: 22622558-22622558 on Build GRCh38
rs1003042380 Chr.11: 22622596-22622596 on Build GRCh38
rs144842627 Chr.11: 22622310-22622310 on Build GRCh38
rs534260150 Chr.11: 22622219-22622219 on Build GRCh38
rs1018411375 Chr.11: 22622488-22622488 on Build GRCh38
rs952777952 Chr.11: 22622669-22622669 on Build GRCh38
rs892906124 Chr.11: 22622537-22622540 on Build GRCh38
rs754552177 Chr.11: 22622328-22622328 on Build GRCh38
rs970288555 Chr.11: 22622370-22622370 on Build GRCh38
rs372028443 Chr.11: 22622545-22622545 on Build GRCh38
rs781192833 Chr.11: 22622445-22622445 on Build GRCh38
rs374459407 Chr.11: 22622314-22622314 on Build GRCh38
rs951636127 Chr.11: 22622593-22622593 on Build GRCh38
rs528990880 Chr.11: 22622304-22622304 on Build GRCh38
rs917658418 Chr.11: 22622348-22622348 on Build GRCh38
rs444879 Chr.11: 22622381-22622381 on Build GRCh38
rs901538646 Chr.11: 22622300-22622300 on Build GRCh38
rs189311515 Chr.11: 22622380-22622380 on Build GRCh38
rs917627689 Chr.11: 22622666-22622666 on Build GRCh38
rs930489007 Chr.11: 22622274-22622275 on Build GRCh38
rs911420757 Chr.11: 22622275-22622275 on Build GRCh38
rs961717338 Chr.11: 22622407-22622407 on Build GRCh38
rs898580155 Chr.11: 22622257-22622259 on Build GRCh38
rs970666127 Chr.11: 22622567-22622567 on Build GRCh38
rs972742871 Chr.11: 22622422-22622422 on Build GRCh38
rs920038739 Chr.11: 22622435-22622435 on Build GRCh38

View Full Product Details