Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36272885-36273445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD9
Entrez Gene ID
29099
Gene Name
COMM domain containing 9
Gene Aliases
HSPC166
UniGene
Hs.279836
Gene Chromosome Location
Chr.11: 36272292-36289449 on Build GRCh38
Transcripts
NM_001101653.1 NM_001307932.1 NM_001307937.1 NM_014186.3 XM_017017625.1

SNP Detail

SNP ID Chromosome Location
rs72946151 Chr.11: 36273195-36273195 on Build GRCh38
rs762001223 Chr.11: 36273064-36273064 on Build GRCh38
rs910849018 Chr.11: 36273194-36273194 on Build GRCh38
rs961232091 Chr.11: 36273120-36273120 on Build GRCh38
rs1029521647 Chr.11: 36273143-36273143 on Build GRCh38
rs989169797 Chr.11: 36273145-36273145 on Build GRCh38
rs905962168 Chr.11: 36272961-36272961 on Build GRCh38
rs1014254140 Chr.11: 36272939-36272939 on Build GRCh38
rs926153724 Chr.11: 36273283-36273283 on Build GRCh38
rs899155592 Chr.11: 36273337-36273337 on Build GRCh38
rs751091315 Chr.11: 36273251-36273251 on Build GRCh38
rs1023794734 Chr.11: 36273075-36273075 on Build GRCh38
rs942666618 Chr.11: 36273149-36273149 on Build GRCh38
rs570613467 Chr.11: 36272973-36272973 on Build GRCh38
rs985109322 Chr.11: 36273179-36273179 on Build GRCh38
rs772934471 Chr.11: 36273160-36273160 on Build GRCh38
rs74469264 Chr.11: 36273404-36273404 on Build GRCh38
rs886498390 Chr.11: 36272985-36272985 on Build GRCh38
rs1017212425 Chr.11: 36273069-36273069 on Build GRCh38
rs1044795503 Chr.11: 36273417-36273417 on Build GRCh38
rs1004897069 Chr.11: 36273009-36273009 on Build GRCh38
rs1005335098 Chr.11: 36273333-36273333 on Build GRCh38
rs1053363145 Chr.11: 36272913-36272913 on Build GRCh38
rs981851966 Chr.11: 36273089-36273089 on Build GRCh38
rs530422088 Chr.11: 36273132-36273132 on Build GRCh38
rs1016643045 Chr.11: 36273018-36273018 on Build GRCh38
rs970857798 Chr.11: 36273081-36273081 on Build GRCh38
rs75341995 Chr.11: 36273378-36273378 on Build GRCh38
rs149056996 Chr.11: 36273203-36273203 on Build GRCh38
rs767880800 Chr.11: 36273125-36273125 on Build GRCh38
rs934935841 Chr.11: 36272912-36272912 on Build GRCh38
rs191633793 Chr.11: 36273141-36273141 on Build GRCh38
rs764406037 Chr.11: 36272921-36272921 on Build GRCh38
rs963663080 Chr.11: 36273024-36273024 on Build GRCh38
rs767671327 Chr.11: 36273192-36273192 on Build GRCh38
rs183631034 Chr.11: 36272977-36272977 on Build GRCh38
rs1046900147 Chr.11: 36273252-36273252 on Build GRCh38
rs934798458 Chr.11: 36273334-36273334 on Build GRCh38
rs979197934 Chr.11: 36273274-36273274 on Build GRCh38
rs933829755 Chr.11: 36273230-36273230 on Build GRCh38
rs1012913011 Chr.11: 36273056-36273056 on Build GRCh38
rs564691787 Chr.11: 36272947-36272947 on Build GRCh38
rs10836524 Chr.11: 36273368-36273368 on Build GRCh38
rs1012531300 Chr.11: 36273362-36273362 on Build GRCh38
rs914853299 Chr.11: 36273148-36273148 on Build GRCh38
rs1022945533 Chr.11: 36272942-36272942 on Build GRCh38
rs886529656 Chr.11: 36273268-36273268 on Build GRCh38
rs188019481 Chr.11: 36272983-36272983 on Build GRCh38

View Full Product Details