Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26957351-26957751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1OP2
Entrez Gene ID
26127
Gene Name
FGFR1 oncogene partner 2
Gene Aliases
HSPC123-like, WIT3.0
UniGene
Hs.591162
Gene Chromosome Location
Chr.12: 26938372-26966648 on Build GRCh38
Transcripts
NM_001171887.1 NM_001171888.1 NM_015633.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4792752 c.388C>T p.H130Y Chr.12: 26957735-26957735 on Build GRCh38
7430048 c.312C>T p.Y104Y Chr.12: 26957659-26957659 on Build GRCh38
6485121 c.364A>T p.I122L Chr.12: 26957711-26957711 on Build GRCh38
6445758 c.344G>A p.S115N Chr.12: 26957691-26957691 on Build GRCh38
301107 c.360G>A p.P120P Chr.12: 26957707-26957707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745673091 Chr.12: 26957711-26957711 on Build GRCh38
rs1040403602 Chr.12: 26957476-26957476 on Build GRCh38
rs558327588 Chr.12: 26957383-26957383 on Build GRCh38
rs771168889 Chr.12: 26957565-26957569 on Build GRCh38
rs367585257 Chr.12: 26957685-26957685 on Build GRCh38
rs910574938 Chr.12: 26957457-26957457 on Build GRCh38
rs750948608 Chr.12: 26957586-26957586 on Build GRCh38
rs369150262 Chr.12: 26957706-26957706 on Build GRCh38
rs990153243 Chr.12: 26957444-26957444 on Build GRCh38
rs906257869 Chr.12: 26957466-26957466 on Build GRCh38
rs1047351063 Chr.12: 26957518-26957518 on Build GRCh38
rs768092079 Chr.12: 26957585-26957585 on Build GRCh38
rs546641291 Chr.12: 26957668-26957668 on Build GRCh38
rs1010796900 Chr.12: 26957523-26957523 on Build GRCh38
rs138526575 Chr.12: 26957628-26957628 on Build GRCh38
rs776450713 Chr.12: 26957635-26957635 on Build GRCh38
rs775003548 Chr.12: 26957655-26957655 on Build GRCh38
rs761611590 Chr.12: 26957676-26957676 on Build GRCh38
rs749386177 Chr.12: 26957608-26957608 on Build GRCh38
rs766810471 Chr.12: 26957692-26957692 on Build GRCh38
rs534253233 Chr.12: 26957474-26957474 on Build GRCh38
rs1031076951 Chr.12: 26957509-26957509 on Build GRCh38
rs1005907656 Chr.12: 26957526-26957526 on Build GRCh38
rs528184573 Chr.12: 26957620-26957620 on Build GRCh38
rs770688810 Chr.12: 26957632-26957632 on Build GRCh38
rs774571715 Chr.12: 26957615-26957615 on Build GRCh38
rs200300827 Chr.12: 26957691-26957691 on Build GRCh38
rs574928146 Chr.12: 26957534-26957534 on Build GRCh38
rs529488831 Chr.12: 26957709-26957709 on Build GRCh38
rs761839225 Chr.12: 26957646-26957646 on Build GRCh38
rs762371523 Chr.12: 26957667-26957667 on Build GRCh38
rs552899009 Chr.12: 26957475-26957475 on Build GRCh38
rs372612021 Chr.12: 26957557-26957557 on Build GRCh38
rs148293272 Chr.12: 26957496-26957496 on Build GRCh38
rs767967536 Chr.12: 26957679-26957679 on Build GRCh38
rs918874898 Chr.12: 26957422-26957422 on Build GRCh38
rs936262933 Chr.12: 26957427-26957427 on Build GRCh38
rs78308909 Chr.12: 26957625-26957625 on Build GRCh38
rs541740829 Chr.12: 26957519-26957519 on Build GRCh38
rs755034016 Chr.12: 26957698-26957698 on Build GRCh38
rs901939759 Chr.12: 26957487-26957487 on Build GRCh38
rs774172300 Chr.12: 26957671-26957671 on Build GRCh38
rs977993693 Chr.12: 26957384-26957384 on Build GRCh38
rs911281909 Chr.12: 26957371-26957371 on Build GRCh38
rs533480644 Chr.12: 26957707-26957707 on Build GRCh38
rs933802325 Chr.12: 26957489-26957489 on Build GRCh38
rs1041439035 Chr.12: 26957403-26957403 on Build GRCh38
rs373660890 Chr.12: 26957569-26957569 on Build GRCh38
rs780178229 Chr.12: 26957606-26957606 on Build GRCh38
rs368072554 Chr.12: 26957611-26957611 on Build GRCh38
rs1012802243 Chr.12: 26957438-26957438 on Build GRCh38
rs1048641255 Chr.12: 26957431-26957432 on Build GRCh38
rs751881356 Chr.12: 26957561-26957561 on Build GRCh38
rs753891063 Chr.12: 26957695-26957695 on Build GRCh38
rs371634012 Chr.12: 26957432-26957432 on Build GRCh38
rs112325643 Chr.12: 26957424-26957424 on Build GRCh38
rs773511881 Chr.12: 26957624-26957624 on Build GRCh38
rs369675830 Chr.12: 26957597-26957597 on Build GRCh38
rs562167202 Chr.12: 26957703-26957703 on Build GRCh38
rs563217092 Chr.12: 26957530-26957530 on Build GRCh38

View Full Product Details