Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51363735-51364326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT6
Entrez Gene ID
11226
Gene Name
polypeptide N-acetylgalactosaminyltransferase 6
Gene Aliases
GALNAC-T6, GalNAcT6
UniGene
Hs.505575
Gene Chromosome Location
Chr.12: 51351870-51391955 on Build GRCh38
Transcripts
NM_007210.3 XM_005268607.1 XM_006719214.2 XM_011537819.2 XM_011537822.2 XM_017018744.1 XM_017018745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7075557 c.919G>C p.E307Q Chr.12: 51364251-51364251 on Build GRCh38
281309 c.894C>T p.I298I Chr.12: 51364276-51364276 on Build GRCh38
431292 c.933C>T p.P311P Chr.12: 51364237-51364237 on Build GRCh38
2009332 c.895G>A p.V299I Chr.12: 51364275-51364275 on Build GRCh38
7335832 c.936_937CC>TT p.Q313* Chr.12: 51364233-51364234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534102589 Chr.12: 51364293-51364293 on Build GRCh38
rs751682648 Chr.12: 51364237-51364237 on Build GRCh38
rs746668719 Chr.12: 51364134-51364134 on Build GRCh38
rs539279424 Chr.12: 51364221-51364221 on Build GRCh38
rs138580901 Chr.12: 51364211-51364211 on Build GRCh38
rs35487700 Chr.12: 51363820-51363821 on Build GRCh38
rs762477181 Chr.12: 51364095-51364095 on Build GRCh38
rs900107801 Chr.12: 51363775-51363775 on Build GRCh38
rs931598576 Chr.12: 51364180-51364180 on Build GRCh38
rs760691190 Chr.12: 51364281-51364281 on Build GRCh38
rs996145921 Chr.12: 51363809-51363809 on Build GRCh38
rs1034076947 Chr.12: 51363951-51363951 on Build GRCh38
rs771230449 Chr.12: 51364070-51364070 on Build GRCh38
rs746281668 Chr.12: 51364163-51364163 on Build GRCh38
rs192500670 Chr.12: 51364236-51364236 on Build GRCh38
rs139003973 Chr.12: 51364269-51364269 on Build GRCh38
rs962780453 Chr.12: 51363947-51363947 on Build GRCh38
rs746406631 Chr.12: 51364253-51364253 on Build GRCh38
rs745715312 Chr.12: 51363834-51363834 on Build GRCh38
rs368818933 Chr.12: 51364111-51364111 on Build GRCh38
rs913753655 Chr.12: 51364042-51364042 on Build GRCh38
rs775170955 Chr.12: 51364182-51364182 on Build GRCh38
rs759188377 Chr.12: 51364086-51364086 on Build GRCh38
rs1004417534 Chr.12: 51364195-51364195 on Build GRCh38
rs1004304548 Chr.12: 51363898-51363898 on Build GRCh38
rs568290859 Chr.12: 51364275-51364275 on Build GRCh38
rs761836894 Chr.12: 51364202-51364202 on Build GRCh38
rs149115702 Chr.12: 51364112-51364112 on Build GRCh38
rs747504266 Chr.12: 51364148-51364148 on Build GRCh38
rs914355606 Chr.12: 51364088-51364088 on Build GRCh38
rs752908979 Chr.12: 51364223-51364223 on Build GRCh38
rs751445455 Chr.12: 51364106-51364106 on Build GRCh38
rs991358991 Chr.12: 51363950-51363950 on Build GRCh38
rs201895050 Chr.12: 51363933-51363933 on Build GRCh38
rs374088917 Chr.12: 51364291-51364291 on Build GRCh38
rs201882458 Chr.12: 51364081-51364081 on Build GRCh38
rs375165419 Chr.12: 51364132-51364132 on Build GRCh38
rs757273714 Chr.12: 51364107-51364107 on Build GRCh38
rs1021259236 Chr.12: 51364276-51364276 on Build GRCh38
rs771498287 Chr.12: 51364160-51364160 on Build GRCh38
rs144722673 Chr.12: 51364131-51364131 on Build GRCh38
rs569288079 Chr.12: 51364084-51364084 on Build GRCh38
rs141181030 Chr.12: 51364245-51364245 on Build GRCh38
rs898796037 Chr.12: 51364151-51364151 on Build GRCh38
rs369520587 Chr.12: 51364145-51364145 on Build GRCh38
rs569315651 Chr.12: 51364113-51364113 on Build GRCh38
rs748338361 Chr.12: 51364140-51364140 on Build GRCh38
rs989776190 Chr.12: 51364015-51364015 on Build GRCh38
rs775155085 Chr.12: 51364130-51364130 on Build GRCh38
rs1025029351 Chr.12: 51363887-51363887 on Build GRCh38
rs201100145 Chr.12: 51364227-51364227 on Build GRCh38
rs966593710 Chr.12: 51364284-51364284 on Build GRCh38
rs764909977 Chr.12: 51364087-51364087 on Build GRCh38
rs138169040 Chr.12: 51364053-51364053 on Build GRCh38
rs146996389 Chr.12: 51363790-51363790 on Build GRCh38
rs760495034 Chr.12: 51364212-51364212 on Build GRCh38
rs773790866 Chr.12: 51364196-51364196 on Build GRCh38
rs776949526 Chr.12: 51364161-51364161 on Build GRCh38
rs1027165591 Chr.12: 51363828-51363828 on Build GRCh38
rs773242274 Chr.12: 51364279-51364279 on Build GRCh38
rs370899778 Chr.12: 51363787-51363787 on Build GRCh38
rs777730506 Chr.12: 51364146-51364146 on Build GRCh38
rs958010625 Chr.12: 51364010-51364010 on Build GRCh38
rs773877136 Chr.12: 51364273-51364273 on Build GRCh38
rs770131667 Chr.12: 51364266-51364266 on Build GRCh38
rs774842425 Chr.12: 51364092-51364092 on Build GRCh38
rs372949504 Chr.12: 51364192-51364192 on Build GRCh38
rs570627140 Chr.12: 51364054-51364054 on Build GRCh38
rs771728741 Chr.12: 51364277-51364277 on Build GRCh38
rs542387310 Chr.12: 51363825-51363825 on Build GRCh38
rs150719035 Chr.12: 51364267-51364267 on Build GRCh38
rs186940282 Chr.12: 51364074-51364074 on Build GRCh38
rs766185241 Chr.12: 51364289-51364289 on Build GRCh38
rs768256768 Chr.12: 51364270-51364270 on Build GRCh38
rs182326946 Chr.12: 51363927-51363927 on Build GRCh38
rs547648001 Chr.12: 51363937-51363937 on Build GRCh38
rs1004299703 Chr.12: 51363869-51363869 on Build GRCh38
rs768772261 Chr.12: 51364102-51364102 on Build GRCh38
rs149435032 Chr.12: 51364306-51364306 on Build GRCh38
rs370958932 Chr.12: 51364246-51364246 on Build GRCh38
rs141325780 Chr.12: 51363763-51363763 on Build GRCh38
rs1030089515 Chr.12: 51363853-51363853 on Build GRCh38
rs770167567 Chr.12: 51364177-51364177 on Build GRCh38
rs377635461 Chr.12: 51364222-51364222 on Build GRCh38

View Full Product Details