Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24842066-24842646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC4
Entrez Gene ID
25932
Gene Name
chloride intracellular channel 4
Gene Aliases
CLIC4L, H1, MTCLIC, huH1, p64H1
UniGene
Hs.440544
Gene Chromosome Location
Chr.1: 24745269-24844324 on Build GRCh38
Transcripts
NM_013943.2

SNP Detail

SNP ID Chromosome Location
rs914636955 Chr.1: 24842172-24842172 on Build GRCh38
rs183897018 Chr.1: 24842276-24842276 on Build GRCh38
rs975977081 Chr.1: 24842415-24842415 on Build GRCh38
rs139150152 Chr.1: 24842422-24842422 on Build GRCh38
rs547620528 Chr.1: 24842198-24842198 on Build GRCh38
rs367587210 Chr.1: 24842487-24842487 on Build GRCh38
rs573541373 Chr.1: 24842460-24842460 on Build GRCh38
rs188432629 Chr.1: 24842538-24842538 on Build GRCh38
rs542461970 Chr.1: 24842396-24842396 on Build GRCh38
rs189128687 Chr.1: 24842223-24842223 on Build GRCh38
rs964475246 Chr.1: 24842395-24842395 on Build GRCh38
rs749264261 Chr.1: 24842537-24842537 on Build GRCh38
rs41292301 Chr.1: 24842200-24842200 on Build GRCh38
rs878924267 Chr.1: 24842491-24842491 on Build GRCh38
rs947487564 Chr.1: 24842570-24842570 on Build GRCh38
rs980718730 Chr.1: 24842599-24842601 on Build GRCh38
rs1057363206 Chr.1: 24842231-24842231 on Build GRCh38
rs775712900 Chr.1: 24842225-24842225 on Build GRCh38
rs914671802 Chr.1: 24842554-24842554 on Build GRCh38
rs1030175150 Chr.1: 24842515-24842515 on Build GRCh38
rs201015135 Chr.1: 24842452-24842452 on Build GRCh38
rs887520414 Chr.1: 24842352-24842352 on Build GRCh38
rs796804231 Chr.1: 24842480-24842480 on Build GRCh38
rs1053623016 Chr.1: 24842132-24842132 on Build GRCh38
rs947563982 Chr.1: 24842182-24842182 on Build GRCh38
rs927451135 Chr.1: 24842622-24842622 on Build GRCh38
rs764242343 Chr.1: 24842490-24842490 on Build GRCh38
rs923277071 Chr.1: 24842099-24842099 on Build GRCh38
rs976092875 Chr.1: 24842091-24842091 on Build GRCh38
rs185865052 Chr.1: 24842103-24842103 on Build GRCh38
rs993135158 Chr.1: 24842298-24842298 on Build GRCh38
rs757845217 Chr.1: 24842594-24842594 on Build GRCh38
rs1017434621 Chr.1: 24842383-24842383 on Build GRCh38
rs1026502856 Chr.1: 24842301-24842301 on Build GRCh38
rs755818518 Chr.1: 24842312-24842326 on Build GRCh38
rs557696059 Chr.1: 24842234-24842234 on Build GRCh38
rs751589829 Chr.1: 24842571-24842571 on Build GRCh38
rs181024620 Chr.1: 24842241-24842241 on Build GRCh38
rs1006500037 Chr.1: 24842360-24842360 on Build GRCh38
rs906150879 Chr.1: 24842219-24842219 on Build GRCh38

View Full Product Details