Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109929524-109930020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT2
Entrez Gene ID
9815
Gene Name
GIT ArfGAP 2
Gene Aliases
CAT-2, CAT2, PKL
UniGene
Hs.434996
Gene Chromosome Location
Chr.12: 109929792-109996389 on Build GRCh38
Transcripts
NM_001135213.1 NM_001135214.1 NM_014776.3 NM_057169.3 NM_057170.3 XM_005253997.3 XM_005254000.3 XM_006719707.3 XM_006719708.3 XM_006719709.3 XM_006719712.3 XM_017020258.1 XM_017020259.1

SNP Detail

SNP ID Chromosome Location
rs994270835 Chr.12: 109929678-109929678 on Build GRCh38
rs966941364 Chr.12: 109929577-109929577 on Build GRCh38
rs555189501 Chr.12: 109929803-109929803 on Build GRCh38
rs558152159 Chr.12: 109929997-109929997 on Build GRCh38
rs377229016 Chr.12: 109929549-109929549 on Build GRCh38
rs913422351 Chr.12: 109929795-109929795 on Build GRCh38
rs936213205 Chr.12: 109929547-109929547 on Build GRCh38
rs116511151 Chr.12: 109929629-109929629 on Build GRCh38
rs957605360 Chr.12: 109929771-109929774 on Build GRCh38
rs938995131 Chr.12: 109929602-109929606 on Build GRCh38
rs1037540993 Chr.12: 109929781-109929781 on Build GRCh38
rs780904242 Chr.12: 109929772-109929772 on Build GRCh38
rs1047541109 Chr.12: 109929699-109929699 on Build GRCh38
rs945143876 Chr.12: 109929594-109929601 on Build GRCh38
rs1051656553 Chr.12: 109929989-109929989 on Build GRCh38
rs1037594089 Chr.12: 109929995-109929995 on Build GRCh38
rs565741929 Chr.12: 109929720-109929720 on Build GRCh38
rs933410369 Chr.12: 109929942-109929942 on Build GRCh38
rs979463869 Chr.12: 109929601-109929601 on Build GRCh38
rs906195555 Chr.12: 109929602-109929602 on Build GRCh38
rs754776252 Chr.12: 109929756-109929756 on Build GRCh38
rs990335903 Chr.12: 109929774-109929774 on Build GRCh38
rs778815913 Chr.12: 109929792-109929792 on Build GRCh38
rs965090598 Chr.12: 109929897-109929897 on Build GRCh38
rs773886782 Chr.12: 109929994-109929994 on Build GRCh38
rs148171384 Chr.12: 109929823-109929823 on Build GRCh38
rs979684837 Chr.12: 109929866-109929866 on Build GRCh38
rs915022744 Chr.12: 109929568-109929568 on Build GRCh38
rs113333945 Chr.12: 109929566-109929566 on Build GRCh38
rs868213740 Chr.12: 109929975-109929975 on Build GRCh38
rs191026169 Chr.12: 109929930-109929930 on Build GRCh38
rs933171191 Chr.12: 109929672-109929672 on Build GRCh38
rs186545021 Chr.12: 109929861-109929861 on Build GRCh38
rs1029707291 Chr.12: 109929681-109929681 on Build GRCh38
rs772675973 Chr.12: 109929944-109929944 on Build GRCh38
rs955593747 Chr.12: 109929690-109929690 on Build GRCh38
rs974751248 Chr.12: 109929913-109929913 on Build GRCh38
rs563554311 Chr.12: 109929828-109929828 on Build GRCh38
rs367730657 Chr.12: 109929719-109929719 on Build GRCh38
rs536462801 Chr.12: 109929765-109929765 on Build GRCh38
rs941949156 Chr.12: 109929754-109929754 on Build GRCh38
rs985477184 Chr.12: 109929757-109929757 on Build GRCh38
rs141083675 Chr.12: 109929876-109929876 on Build GRCh38
rs916529673 Chr.12: 109929916-109929916 on Build GRCh38
rs572097244 Chr.12: 109929609-109929609 on Build GRCh38
rs1054778619 Chr.12: 109929840-109929840 on Build GRCh38
rs143648557 Chr.12: 109929818-109929818 on Build GRCh38
rs943313196 Chr.12: 109929609-109929609 on Build GRCh38
rs895057011 Chr.12: 109929620-109929620 on Build GRCh38
rs966993296 Chr.12: 109929863-109929863 on Build GRCh38
rs574840246 Chr.12: 109929925-109929925 on Build GRCh38
rs570904505 Chr.12: 109929567-109929567 on Build GRCh38
rs925273320 Chr.12: 109929604-109929604 on Build GRCh38
rs61940901 Chr.12: 109929620-109929620 on Build GRCh38
rs779061945 Chr.12: 109929703-109929704 on Build GRCh38
rs886391532 Chr.12: 109929745-109929745 on Build GRCh38
rs1038269030 Chr.12: 109929932-109929932 on Build GRCh38
rs182277378 Chr.12: 109929831-109929831 on Build GRCh38
rs1018966014 Chr.12: 109929764-109929764 on Build GRCh38
rs1032562465 Chr.12: 109929883-109929883 on Build GRCh38
rs905543193 Chr.12: 109929791-109929791 on Build GRCh38
rs775122038 Chr.12: 109929853-109929853 on Build GRCh38

View Full Product Details