Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43164375-43164943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBNA1BP2
Entrez Gene ID
10969
Gene Name
EBNA1 binding protein 2
Gene Aliases
EBP2, NOBP, P40
UniGene
Hs.346868
Gene Chromosome Location
Chr.1: 43164174-43172570 on Build GRCh38
Transcripts
NM_001159936.1 NM_006824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5793514 c.911G>C p.G304A Chr.1: 43164767-43164767 on Build GRCh38
3790036 c.1023G>C p.K341N Chr.1: 43164655-43164655 on Build GRCh38
5846723 c.928A>T p.K310* Chr.1: 43164750-43164750 on Build GRCh38
909714 c.722G>A p.R241Q Chr.1: 43164791-43164791 on Build GRCh38
2170233 c.814C>T p.R272W Chr.1: 43164699-43164699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970016796 Chr.1: 43164710-43164710 on Build GRCh38
rs758943200 Chr.1: 43164622-43164622 on Build GRCh38
rs374106621 Chr.1: 43164505-43164505 on Build GRCh38
rs201884003 Chr.1: 43164520-43164520 on Build GRCh38
rs779385087 Chr.1: 43164877-43164877 on Build GRCh38
rs984043527 Chr.1: 43164409-43164409 on Build GRCh38
rs773905782 Chr.1: 43164543-43164543 on Build GRCh38
rs74484344 Chr.1: 43164610-43164610 on Build GRCh38
rs373757787 Chr.1: 43164636-43164636 on Build GRCh38
rs749250062 Chr.1: 43164651-43164651 on Build GRCh38
rs1020250345 Chr.1: 43164530-43164530 on Build GRCh38
rs148849429 Chr.1: 43164914-43164914 on Build GRCh38
rs377120011 Chr.1: 43164451-43164451 on Build GRCh38
rs770867555 Chr.1: 43164657-43164657 on Build GRCh38
rs144683125 Chr.1: 43164649-43164649 on Build GRCh38
rs770578885 Chr.1: 43164777-43164777 on Build GRCh38
rs767770848 Chr.1: 43164832-43164832 on Build GRCh38
rs971775355 Chr.1: 43164551-43164551 on Build GRCh38
rs11559311 Chr.1: 43164485-43164485 on Build GRCh38
rs748693560 Chr.1: 43164898-43164898 on Build GRCh38
rs1054552666 Chr.1: 43164829-43164829 on Build GRCh38
rs766933126 Chr.1: 43164721-43164721 on Build GRCh38
rs201723368 Chr.1: 43164663-43164663 on Build GRCh38
rs747263444 Chr.1: 43164495-43164495 on Build GRCh38
rs879568441 Chr.1: 43164494-43164644 on Build GRCh38
rs747979294 Chr.1: 43164507-43164507 on Build GRCh38
rs756316772 Chr.1: 43164728-43164728 on Build GRCh38
rs750770503 Chr.1: 43164712-43164712 on Build GRCh38
rs752791601 Chr.1: 43164833-43164833 on Build GRCh38
rs766587245 Chr.1: 43164817-43164817 on Build GRCh38
rs755112494 Chr.1: 43164725-43164725 on Build GRCh38
rs748434637 Chr.1: 43164461-43164461 on Build GRCh38
rs751184199 Chr.1: 43164397-43164397 on Build GRCh38
rs759148337 Chr.1: 43164545-43164545 on Build GRCh38
rs754167669 Chr.1: 43164470-43164470 on Build GRCh38
rs753873150 Chr.1: 43164830-43164830 on Build GRCh38
rs1001513304 Chr.1: 43164719-43164719 on Build GRCh38
rs751972108 Chr.1: 43164724-43164724 on Build GRCh38
rs11559312 Chr.1: 43164644-43164644 on Build GRCh38
rs766950805 Chr.1: 43164694-43164694 on Build GRCh38
rs181497889 Chr.1: 43164717-43164717 on Build GRCh38
rs778937681 Chr.1: 43164785-43164785 on Build GRCh38
rs760363408 Chr.1: 43164792-43164792 on Build GRCh38
rs200101008 Chr.1: 43164629-43164629 on Build GRCh38
rs150637152 Chr.1: 43164789-43164789 on Build GRCh38
rs186044867 Chr.1: 43164853-43164853 on Build GRCh38
rs760106882 Chr.1: 43164828-43164828 on Build GRCh38
rs759104670 Chr.1: 43164680-43164680 on Build GRCh38
rs765532570 Chr.1: 43164603-43164603 on Build GRCh38
rs371798157 Chr.1: 43164790-43164790 on Build GRCh38
rs768277562 Chr.1: 43164804-43164804 on Build GRCh38
rs750953145 Chr.1: 43164620-43164620 on Build GRCh38
rs543939580 Chr.1: 43164552-43164552 on Build GRCh38
rs781528619 Chr.1: 43164460-43164462 on Build GRCh38
rs528905830 Chr.1: 43164729-43164729 on Build GRCh38
rs774978698 Chr.1: 43164791-43164791 on Build GRCh38
rs773780597 Chr.1: 43164664-43164664 on Build GRCh38
rs564219282 Chr.1: 43164555-43164555 on Build GRCh38
rs61776916 Chr.1: 43164870-43164870 on Build GRCh38
rs753963463 Chr.1: 43164852-43164852 on Build GRCh38
rs770353070 Chr.1: 43164463-43164476 on Build GRCh38
rs367925917 Chr.1: 43164695-43164695 on Build GRCh38
rs371251837 Chr.1: 43164526-43164526 on Build GRCh38
rs777466628 Chr.1: 43164768-43164768 on Build GRCh38
rs922376152 Chr.1: 43164903-43164903 on Build GRCh38
rs189165159 Chr.1: 43164455-43164455 on Build GRCh38
rs749618599 Chr.1: 43164678-43164678 on Build GRCh38
rs1063390 Chr.1: 43164447-43164447 on Build GRCh38
rs369373223 Chr.1: 43164478-43164478 on Build GRCh38
rs769672241 Chr.1: 43164511-43164511 on Build GRCh38
rs745750983 Chr.1: 43164788-43164788 on Build GRCh38
rs908257273 Chr.1: 43164412-43164412 on Build GRCh38
rs756399600 Chr.1: 43164736-43164736 on Build GRCh38
rs763406535 Chr.1: 43164815-43164815 on Build GRCh38
rs932340877 Chr.1: 43164490-43164490 on Build GRCh38
rs199606135 Chr.1: 43164430-43164430 on Build GRCh38
rs767130724 Chr.1: 43164593-43164593 on Build GRCh38
rs746310555 Chr.1: 43164480-43164480 on Build GRCh38
rs768893864 Chr.1: 43164497-43164497 on Build GRCh38
rs764507117 Chr.1: 43164468-43164468 on Build GRCh38
rs780171747 Chr.1: 43164489-43164489 on Build GRCh38
rs747091326 Chr.1: 43164626-43164626 on Build GRCh38
rs776100286 Chr.1: 43164805-43164805 on Build GRCh38
rs12135134 Chr.1: 43164558-43164558 on Build GRCh38
rs377680618 Chr.1: 43164523-43164523 on Build GRCh38
rs757806675 Chr.1: 43164477-43164477 on Build GRCh38
rs376694289 Chr.1: 43164668-43164668 on Build GRCh38
rs904463829 Chr.1: 43164557-43164557 on Build GRCh38
rs752294175 Chr.1: 43164594-43164594 on Build GRCh38
rs760310247 Chr.1: 43164697-43164697 on Build GRCh38
rs891837300 Chr.1: 43164598-43164598 on Build GRCh38
rs563104383 Chr.1: 43164549-43164549 on Build GRCh38
rs752119924 Chr.1: 43164820-43164820 on Build GRCh38
rs376986557 Chr.1: 43164623-43164623 on Build GRCh38
rs756450746 Chr.1: 43164842-43164842 on Build GRCh38
rs765659667 Chr.1: 43164698-43164698 on Build GRCh38
rs1049401 Chr.1: 43164449-43164449 on Build GRCh38
rs749755523 Chr.1: 43164844-43164844 on Build GRCh38
rs375913443 Chr.1: 43164601-43164601 on Build GRCh38
rs778077010 Chr.1: 43164847-43164847 on Build GRCh38

View Full Product Details