Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40990797-40991394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001301237.1 NM_001905.3 NR_125440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4238161 c.556-9G>T p.? Chr.1: 40991156-40991156 on Build GRCh38
5928048 c.556-8T>C p.? Chr.1: 40991157-40991157 on Build GRCh38
535209 c.614G>A p.R205K Chr.1: 40991223-40991223 on Build GRCh38
5529382 c.584C>T p.S195L Chr.1: 40991172-40991172 on Build GRCh38
6637689 c.620G>C p.S207T Chr.1: 40991208-40991208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930152414 Chr.1: 40991037-40991037 on Build GRCh38
rs767110030 Chr.1: 40991141-40991142 on Build GRCh38
rs773267175 Chr.1: 40991134-40991134 on Build GRCh38
rs754783094 Chr.1: 40991324-40991324 on Build GRCh38
rs748665833 Chr.1: 40991137-40991138 on Build GRCh38
rs758999285 Chr.1: 40991161-40991161 on Build GRCh38
rs977606780 Chr.1: 40991035-40991035 on Build GRCh38
rs4660182 Chr.1: 40990881-40990881 on Build GRCh38
rs776745334 Chr.1: 40991117-40991117 on Build GRCh38
rs200737909 Chr.1: 40991156-40991156 on Build GRCh38
rs1056146453 Chr.1: 40990825-40990825 on Build GRCh38
rs572319596 Chr.1: 40991246-40991246 on Build GRCh38
rs1048535526 Chr.1: 40991039-40991039 on Build GRCh38
rs747327086 Chr.1: 40991130-40991133 on Build GRCh38
rs749108890 Chr.1: 40991275-40991275 on Build GRCh38
rs368774990 Chr.1: 40991150-40991151 on Build GRCh38
rs959581294 Chr.1: 40990874-40990874 on Build GRCh38
rs964417955 Chr.1: 40991353-40991353 on Build GRCh38
rs572312351 Chr.1: 40991149-40991150 on Build GRCh38
rs200861913 Chr.1: 40991151-40991151 on Build GRCh38
rs554721789 Chr.1: 40991331-40991331 on Build GRCh38
rs377619700 Chr.1: 40991151-40991151 on Build GRCh38
rs765659493 Chr.1: 40991214-40991214 on Build GRCh38
rs918874001 Chr.1: 40991349-40991349 on Build GRCh38
rs781456987 Chr.1: 40991252-40991252 on Build GRCh38
rs773076522 Chr.1: 40991274-40991274 on Build GRCh38
rs1034819319 Chr.1: 40991126-40991126 on Build GRCh38
rs1012612358 Chr.1: 40990850-40990850 on Build GRCh38
rs748379401 Chr.1: 40991258-40991258 on Build GRCh38
rs952764300 Chr.1: 40990856-40990856 on Build GRCh38
rs986729259 Chr.1: 40990953-40990953 on Build GRCh38
rs747941008 Chr.1: 40991122-40991122 on Build GRCh38
rs527444357 Chr.1: 40990916-40990916 on Build GRCh38
rs936428994 Chr.1: 40990966-40990966 on Build GRCh38
rs1001981673 Chr.1: 40990870-40990870 on Build GRCh38
rs985238561 Chr.1: 40991284-40991284 on Build GRCh38
rs11209331 Chr.1: 40991017-40991017 on Build GRCh38
rs568218457 Chr.1: 40991285-40991289 on Build GRCh38
rs1032156922 Chr.1: 40991218-40991218 on Build GRCh38
rs966538497 Chr.1: 40990900-40990900 on Build GRCh38
rs142312983 Chr.1: 40991242-40991242 on Build GRCh38
rs773676512 Chr.1: 40991137-40991139 on Build GRCh38
rs758848799 Chr.1: 40991227-40991227 on Build GRCh38
rs540402867 Chr.1: 40991137-40991137 on Build GRCh38
rs552306231 Chr.1: 40990883-40990883 on Build GRCh38
rs985551340 Chr.1: 40990984-40990984 on Build GRCh38
rs977961427 Chr.1: 40991203-40991203 on Build GRCh38
rs1056137038 Chr.1: 40991066-40991066 on Build GRCh38
rs779891480 Chr.1: 40991110-40991110 on Build GRCh38
rs188426095 Chr.1: 40991318-40991318 on Build GRCh38
rs943671195 Chr.1: 40991003-40991003 on Build GRCh38
rs977888390 Chr.1: 40990925-40990925 on Build GRCh38
rs34406683 Chr.1: 40991095-40991096 on Build GRCh38
rs955458121 Chr.1: 40990933-40990933 on Build GRCh38
rs895032626 Chr.1: 40990843-40990843 on Build GRCh38
rs767168383 Chr.1: 40991164-40991164 on Build GRCh38
rs910938319 Chr.1: 40991306-40991306 on Build GRCh38
rs750794414 Chr.1: 40991222-40991222 on Build GRCh38
rs1024875966 Chr.1: 40990894-40990894 on Build GRCh38
rs536533173 Chr.1: 40991104-40991104 on Build GRCh38
rs769757382 Chr.1: 40991124-40991124 on Build GRCh38
rs747075079 Chr.1: 40991249-40991249 on Build GRCh38
rs116700741 Chr.1: 40991149-40991149 on Build GRCh38
rs925060566 Chr.1: 40990950-40990950 on Build GRCh38
rs757606681 Chr.1: 40991196-40991196 on Build GRCh38
rs958054114 Chr.1: 40991253-40991253 on Build GRCh38
rs894843371 Chr.1: 40991087-40991087 on Build GRCh38
rs879695820 Chr.1: 40991158-40991158 on Build GRCh38
rs769709248 Chr.1: 40991263-40991263 on Build GRCh38
rs183893726 Chr.1: 40990973-40990973 on Build GRCh38
rs947979634 Chr.1: 40991092-40991092 on Build GRCh38
rs538323954 Chr.1: 40991042-40991042 on Build GRCh38
rs138824927 Chr.1: 40991011-40991011 on Build GRCh38
rs755056680 Chr.1: 40991251-40991251 on Build GRCh38
rs200909069 Chr.1: 40990932-40990932 on Build GRCh38
rs386366777 Chr.1: 40991143-40991144 on Build GRCh38
rs1001614378 Chr.1: 40991051-40991051 on Build GRCh38
rs533748476 Chr.1: 40991308-40991308 on Build GRCh38
rs150200292 Chr.1: 40990873-40990873 on Build GRCh38
rs377028455 Chr.1: 40991136-40991136 on Build GRCh38
rs770943617 Chr.1: 40991285-40991285 on Build GRCh38
rs116134285 Chr.1: 40991179-40991179 on Build GRCh38
rs111518708 Chr.1: 40991136-40991137 on Build GRCh38
rs910875190 Chr.1: 40990986-40990986 on Build GRCh38
rs964322692 Chr.1: 40990978-40990978 on Build GRCh38
rs141075178 Chr.1: 40991366-40991366 on Build GRCh38

View Full Product Details