Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84414400-84414926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE2B
Entrez Gene ID
58511
Gene Name
deoxyribonuclease II beta
Gene Aliases
DLAD
UniGene
Hs.129142
Gene Chromosome Location
Chr.1: 84398532-84415895 on Build GRCh38
Transcripts
NM_021233.2 NM_058248.1 XM_011541878.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344546 c.296A>G p.K99R Chr.1: 84414702-84414702 on Build GRCh38
2243639 c.980G>A p.R327Q Chr.1: 84414762-84414762 on Build GRCh38
3492947 c.924G>A p.W308* Chr.1: 84414706-84414706 on Build GRCh38
2243630 c.253T>G p.L85V Chr.1: 84414659-84414659 on Build GRCh38
232196 c.185G>A p.R62Q Chr.1: 84414591-84414591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889576489 Chr.1: 84414722-84414722 on Build GRCh38
rs751888980 Chr.1: 84414746-84414746 on Build GRCh38
rs772304277 Chr.1: 84414484-84414484 on Build GRCh38
rs762926399 Chr.1: 84414609-84414609 on Build GRCh38
rs994127903 Chr.1: 84414808-84414808 on Build GRCh38
rs771852488 Chr.1: 84414842-84414843 on Build GRCh38
rs201823459 Chr.1: 84414521-84414521 on Build GRCh38
rs777143916 Chr.1: 84414792-84414792 on Build GRCh38
rs573492273 Chr.1: 84414444-84414444 on Build GRCh38
rs772236685 Chr.1: 84414566-84414566 on Build GRCh38
rs773211478 Chr.1: 84414811-84414811 on Build GRCh38
rs773228559 Chr.1: 84414485-84414485 on Build GRCh38
rs376877314 Chr.1: 84414554-84414554 on Build GRCh38
rs760791945 Chr.1: 84414491-84414491 on Build GRCh38
rs767081210 Chr.1: 84414633-84414633 on Build GRCh38
rs200047881 Chr.1: 84414665-84414665 on Build GRCh38
rs1026763183 Chr.1: 84414543-84414543 on Build GRCh38
rs763825915 Chr.1: 84414516-84414516 on Build GRCh38
rs199602718 Chr.1: 84414869-84414869 on Build GRCh38
rs755606469 Chr.1: 84414645-84414645 on Build GRCh38
rs763553743 Chr.1: 84414843-84414843 on Build GRCh38
rs755623199 Chr.1: 84414531-84414531 on Build GRCh38
rs570370636 Chr.1: 84414740-84414740 on Build GRCh38
rs745523268 Chr.1: 84414502-84414502 on Build GRCh38
rs767186634 Chr.1: 84414525-84414525 on Build GRCh38
rs781335051 Chr.1: 84414761-84414761 on Build GRCh38
rs762766886 Chr.1: 84414506-84414506 on Build GRCh38
rs753207337 Chr.1: 84414739-84414739 on Build GRCh38
rs766038489 Chr.1: 84414641-84414641 on Build GRCh38
rs745936782 Chr.1: 84414685-84414685 on Build GRCh38
rs766122330 Chr.1: 84414721-84414721 on Build GRCh38
rs746783773 Chr.1: 84414815-84414815 on Build GRCh38
rs570910368 Chr.1: 84414863-84414863 on Build GRCh38
rs982564731 Chr.1: 84414581-84414581 on Build GRCh38
rs751936110 Chr.1: 84414861-84414861 on Build GRCh38
rs768578597 Chr.1: 84414621-84414621 on Build GRCh38
rs76042396 Chr.1: 84414744-84414744 on Build GRCh38
rs565736451 Chr.1: 84414805-84414805 on Build GRCh38
rs770718741 Chr.1: 84414819-84414819 on Build GRCh38
rs375823079 Chr.1: 84414591-84414591 on Build GRCh38
rs748371215 Chr.1: 84414482-84414482 on Build GRCh38
rs755522428 Chr.1: 84414547-84414547 on Build GRCh38
rs144779905 Chr.1: 84414590-84414590 on Build GRCh38
rs531415773 Chr.1: 84414631-84414631 on Build GRCh38
rs963663537 Chr.1: 84414458-84414458 on Build GRCh38
rs778741286 Chr.1: 84414778-84414778 on Build GRCh38
rs774840549 Chr.1: 84414838-84414838 on Build GRCh38
rs780225027 Chr.1: 84414893-84414893 on Build GRCh38
rs756271909 Chr.1: 84414880-84414880 on Build GRCh38
rs760446433 Chr.1: 84414719-84414719 on Build GRCh38
rs200661167 Chr.1: 84414662-84414662 on Build GRCh38
rs1003580118 Chr.1: 84414901-84414901 on Build GRCh38
rs545449983 Chr.1: 84414666-84414666 on Build GRCh38
rs145614783 Chr.1: 84414762-84414762 on Build GRCh38
rs771876558 Chr.1: 84414810-84414810 on Build GRCh38
rs771117957 Chr.1: 84414671-84414671 on Build GRCh38
rs779459080 Chr.1: 84414650-84414650 on Build GRCh38
rs749916394 Chr.1: 84414529-84414529 on Build GRCh38
rs182527874 Chr.1: 84414832-84414832 on Build GRCh38
rs557864256 Chr.1: 84414848-84414848 on Build GRCh38
rs368296260 Chr.1: 84414634-84414634 on Build GRCh38
rs3116432 Chr.1: 84414501-84414501 on Build GRCh38
rs1028340401 Chr.1: 84414428-84414428 on Build GRCh38
rs991843493 Chr.1: 84414898-84414898 on Build GRCh38
rs780742897 Chr.1: 84414541-84414541 on Build GRCh38
rs776351674 Chr.1: 84414829-84414829 on Build GRCh38
rs753453449 Chr.1: 84414723-84414723 on Build GRCh38
rs771812059 Chr.1: 84414540-84414540 on Build GRCh38
rs764754558 Chr.1: 84414833-84414833 on Build GRCh38
rs202202221 Chr.1: 84414767-84414767 on Build GRCh38
rs767758185 Chr.1: 84414867-84414867 on Build GRCh38
rs147325752 Chr.1: 84414828-84414828 on Build GRCh38
rs146223730 Chr.1: 84414735-84414735 on Build GRCh38
rs747184876 Chr.1: 84414667-84414667 on Build GRCh38
rs777991220 Chr.1: 84414571-84414571 on Build GRCh38
rs750598705 Chr.1: 84414873-84414873 on Build GRCh38
rs748539735 Chr.1: 84414565-84414565 on Build GRCh38
rs3768250 Chr.1: 84414697-84414697 on Build GRCh38
rs754350963 Chr.1: 84414544-84414544 on Build GRCh38
rs760288673 Chr.1: 84414637-84414637 on Build GRCh38
rs933675481 Chr.1: 84414699-84414699 on Build GRCh38
rs191605025 Chr.1: 84414698-84414698 on Build GRCh38
rs373884042 Chr.1: 84414747-84414747 on Build GRCh38
rs370855181 Chr.1: 84414676-84414676 on Build GRCh38
rs771855021 Chr.1: 84414716-84414716 on Build GRCh38
rs759054755 Chr.1: 84414727-84414727 on Build GRCh38
rs527804375 Chr.1: 84414642-84414642 on Build GRCh38
rs377602479 Chr.1: 84414495-84414495 on Build GRCh38
rs1026569429 Chr.1: 84414827-84414827 on Build GRCh38
rs200871122 Chr.1: 84414622-84414622 on Build GRCh38
rs900648170 Chr.1: 84414752-84414752 on Build GRCh38
rs775426791 Chr.1: 84414603-84414603 on Build GRCh38
rs761548011 Chr.1: 84414709-84414709 on Build GRCh38
rs756927629 Chr.1: 84414523-84414523 on Build GRCh38
rs374590812 Chr.1: 84414617-84414617 on Build GRCh38

View Full Product Details