Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150552571-150553154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL4
Entrez Gene ID
54507
Gene Name
ADAMTS like 4
Gene Aliases
ADAMTSL-4, ECTOL2, TSRC1
UniGene
Hs.516243
Gene Chromosome Location
Chr.1: 150549369-150560937 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_017001506.1

Gene Symbol
LOC100289061
Entrez Gene ID
100289061
Gene Name
uncharacterized LOC100289061
Gene Aliases
-
UniGene
Hs.677561
Gene Chromosome Location
Chr.1: 150548564-150556700 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_017001506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6576842 c.207G>T p.R69R Chr.1: 150553026-150553026 on Build GRCh38
4981025 c.318T>A p.T106T Chr.1: 150553137-150553137 on Build GRCh38
3473439 c.242G>A p.S81N Chr.1: 150553061-150553061 on Build GRCh38
5795545 c.293G>A p.R98Q Chr.1: 150553112-150553112 on Build GRCh38
4851362 c.164C>T p.S55F Chr.1: 150552983-150552983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762836841 Chr.1: 150552991-150552991 on Build GRCh38
rs778236726 Chr.1: 150552936-150552936 on Build GRCh38
rs781562228 Chr.1: 150552913-150552913 on Build GRCh38
rs761529518 Chr.1: 150552867-150552867 on Build GRCh38
rs1024865790 Chr.1: 150552822-150552822 on Build GRCh38
rs910153294 Chr.1: 150552774-150552774 on Build GRCh38
rs776567842 Chr.1: 150552854-150552854 on Build GRCh38
rs747590395 Chr.1: 150553039-150553039 on Build GRCh38
rs971608344 Chr.1: 150552875-150552875 on Build GRCh38
rs755040821 Chr.1: 150552907-150552907 on Build GRCh38
rs751940403 Chr.1: 150552927-150552927 on Build GRCh38
rs774759808 Chr.1: 150552682-150552682 on Build GRCh38
rs760309871 Chr.1: 150552930-150552930 on Build GRCh38
rs958813832 Chr.1: 150552613-150552613 on Build GRCh38
rs767121643 Chr.1: 150552979-150552979 on Build GRCh38
rs751772197 Chr.1: 150552998-150552998 on Build GRCh38
rs148393631 Chr.1: 150553111-150553111 on Build GRCh38
rs939895316 Chr.1: 150553037-150553037 on Build GRCh38
rs977889757 Chr.1: 150552881-150552881 on Build GRCh38
rs771686014 Chr.1: 150552597-150552597 on Build GRCh38
rs754709263 Chr.1: 150553091-150553091 on Build GRCh38
rs775692968 Chr.1: 150553059-150553059 on Build GRCh38
rs200120255 Chr.1: 150553129-150553129 on Build GRCh38
rs775513427 Chr.1: 150553130-150553130 on Build GRCh38
rs1015366132 Chr.1: 150552700-150552700 on Build GRCh38
rs587624924 Chr.1: 150552788-150552788 on Build GRCh38
rs750644116 Chr.1: 150552874-150552874 on Build GRCh38
rs958904143 Chr.1: 150552806-150552806 on Build GRCh38
rs995203831 Chr.1: 150552771-150552771 on Build GRCh38
rs745633055 Chr.1: 150553122-150553122 on Build GRCh38
rs749626359 Chr.1: 150552941-150552941 on Build GRCh38
rs587776927 Chr.1: 150552897-150552897 on Build GRCh38
rs587593757 Chr.1: 150553112-150553112 on Build GRCh38
rs754288272 Chr.1: 150553068-150553068 on Build GRCh38
rs749108522 Chr.1: 150553045-150553045 on Build GRCh38
rs1024281147 Chr.1: 150552999-150552999 on Build GRCh38
rs911666795 Chr.1: 150553112-150553112 on Build GRCh38
rs150631268 Chr.1: 150552981-150552981 on Build GRCh38
rs794726689 Chr.1: 150553058-150553058 on Build GRCh38
rs767591627 Chr.1: 150553002-150553002 on Build GRCh38
rs747697443 Chr.1: 150553098-150553098 on Build GRCh38
rs975518971 Chr.1: 150553096-150553096 on Build GRCh38
rs764670992 Chr.1: 150553065-150553065 on Build GRCh38
rs768650448 Chr.1: 150552644-150552644 on Build GRCh38
rs766712956 Chr.1: 150552995-150552995 on Build GRCh38
rs746529379 Chr.1: 150552950-150552950 on Build GRCh38
rs898140143 Chr.1: 150552759-150552759 on Build GRCh38
rs983065900 Chr.1: 150552726-150552726 on Build GRCh38
rs759875828 Chr.1: 150553055-150553055 on Build GRCh38
rs765034632 Chr.1: 150552871-150552871 on Build GRCh38
rs1024312192 Chr.1: 150552670-150552670 on Build GRCh38
rs763307395 Chr.1: 150552884-150552884 on Build GRCh38
rs146377111 Chr.1: 150553058-150553058 on Build GRCh38
rs144538837 Chr.1: 150553107-150553107 on Build GRCh38
rs754479372 Chr.1: 150553024-150553024 on Build GRCh38
rs587758041 Chr.1: 150552686-150552686 on Build GRCh38
rs587760979 Chr.1: 150552948-150552948 on Build GRCh38
rs963323596 Chr.1: 150553056-150553056 on Build GRCh38
rs773265662 Chr.1: 150552854-150552854 on Build GRCh38
rs371107461 Chr.1: 150552807-150552807 on Build GRCh38
rs760671345 Chr.1: 150553061-150553061 on Build GRCh38
rs374443010 Chr.1: 150553015-150553015 on Build GRCh38
rs764414687 Chr.1: 150552916-150552916 on Build GRCh38
rs760169235 Chr.1: 150552642-150552642 on Build GRCh38
rs750755864 Chr.1: 150553080-150553080 on Build GRCh38
rs1004344502 Chr.1: 150552697-150552697 on Build GRCh38
rs757553666 Chr.1: 150553016-150553016 on Build GRCh38
rs755521002 Chr.1: 150553099-150553099 on Build GRCh38
rs962890910 Chr.1: 150552792-150552792 on Build GRCh38
rs587688843 Chr.1: 150553115-150553115 on Build GRCh38
rs773108415 Chr.1: 150552990-150552990 on Build GRCh38
rs761977789 Chr.1: 150552879-150552880 on Build GRCh38
rs768099905 Chr.1: 150552960-150552960 on Build GRCh38
rs754048794 Chr.1: 150553009-150553009 on Build GRCh38
rs376796356 Chr.1: 150553020-150553020 on Build GRCh38
rs1014232801 Chr.1: 150552630-150552630 on Build GRCh38
rs759709764 Chr.1: 150553000-150553000 on Build GRCh38
rs757643403 Chr.1: 150553078-150553078 on Build GRCh38
rs147688134 Chr.1: 150553051-150553051 on Build GRCh38
rs775033921 Chr.1: 150552600-150552600 on Build GRCh38
rs769827413 Chr.1: 150553040-150553040 on Build GRCh38
rs202059899 Chr.1: 150552906-150552906 on Build GRCh38
rs761772078 Chr.1: 150552985-150552985 on Build GRCh38
rs766686599 Chr.1: 150552892-150552892 on Build GRCh38
rs770798771 Chr.1: 150553046-150553046 on Build GRCh38
rs587755814 Chr.1: 150553005-150553005 on Build GRCh38
rs779697639 Chr.1: 150553018-150553018 on Build GRCh38
rs199599791 Chr.1: 150553079-150553079 on Build GRCh38
rs777154231 Chr.1: 150552980-150552980 on Build GRCh38
rs773215849 Chr.1: 150553043-150553043 on Build GRCh38
rs587761331 Chr.1: 150553093-150553093 on Build GRCh38
rs111348494 Chr.1: 150552911-150552911 on Build GRCh38
rs41317513 Chr.1: 150552949-150552949 on Build GRCh38
rs921678535 Chr.1: 150552812-150552812 on Build GRCh38
rs780795465 Chr.1: 150553025-150553025 on Build GRCh38

View Full Product Details