Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110016289-110016805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL1
Entrez Gene ID
10768
Gene Name
adenosylhomocysteinase like 1
Gene Aliases
DCAL, IRBIT, PPP1R78, PRO0233, XPVKONA
UniGene
Hs.743973
Gene Chromosome Location
Chr.1: 109984741-110023742 on Build GRCh38
Transcripts
NM_001242673.1 NM_001242674.1 NM_001242675.1 NM_001242676.1 NM_006621.5 XM_011540535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4856757 c.823G>A p.A275T Chr.1: 110016384-110016384 on Build GRCh38
5689305 c.917A>T p.D306V Chr.1: 110016684-110016684 on Build GRCh38
893601 c.960T>C p.G320G Chr.1: 110016727-110016727 on Build GRCh38
6363870 c.848C>A p.T283N Chr.1: 110016409-110016409 on Build GRCh38
7170894 c.900C>T p.G300G Chr.1: 110016667-110016667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549162131 Chr.1: 110016481-110016481 on Build GRCh38
rs757610161 Chr.1: 110016721-110016721 on Build GRCh38
rs760583703 Chr.1: 110016636-110016636 on Build GRCh38
rs781468164 Chr.1: 110016730-110016730 on Build GRCh38
rs746411212 Chr.1: 110016490-110016490 on Build GRCh38
rs781695173 Chr.1: 110016485-110016485 on Build GRCh38
rs1012408771 Chr.1: 110016644-110016644 on Build GRCh38
rs757584904 Chr.1: 110016478-110016478 on Build GRCh38
rs775614920 Chr.1: 110016494-110016494 on Build GRCh38
rs756548235 Chr.1: 110016756-110016756 on Build GRCh38
rs151242532 Chr.1: 110016312-110016312 on Build GRCh38
rs138987880 Chr.1: 110016702-110016702 on Build GRCh38
rs978132021 Chr.1: 110016773-110016773 on Build GRCh38
rs774612499 Chr.1: 110016578-110016578 on Build GRCh38
rs534958536 Chr.1: 110016519-110016519 on Build GRCh38
rs780701110 Chr.1: 110016464-110016464 on Build GRCh38
rs445885 Chr.1: 110016750-110016750 on Build GRCh38
rs896913216 Chr.1: 110016566-110016566 on Build GRCh38
rs751987139 Chr.1: 110016623-110016642 on Build GRCh38
rs747340362 Chr.1: 110016474-110016474 on Build GRCh38
rs556651308 Chr.1: 110016776-110016776 on Build GRCh38
rs749376899 Chr.1: 110016495-110016495 on Build GRCh38
rs765980253 Chr.1: 110016642-110016642 on Build GRCh38
rs902068367 Chr.1: 110016751-110016751 on Build GRCh38
rs754547222 Chr.1: 110016659-110016659 on Build GRCh38
rs941052252 Chr.1: 110016514-110016514 on Build GRCh38
rs565732609 Chr.1: 110016492-110016492 on Build GRCh38
rs112525217 Chr.1: 110016621-110016621 on Build GRCh38
rs761529477 Chr.1: 110016332-110016332 on Build GRCh38
rs376789939 Chr.1: 110016738-110016738 on Build GRCh38
rs932479372 Chr.1: 110016463-110016463 on Build GRCh38
rs968682587 Chr.1: 110016769-110016769 on Build GRCh38
rs374677505 Chr.1: 110016589-110016589 on Build GRCh38
rs567763788 Chr.1: 110016424-110016424 on Build GRCh38
rs530098093 Chr.1: 110016454-110016454 on Build GRCh38
rs768811650 Chr.1: 110016505-110016505 on Build GRCh38
rs746383352 Chr.1: 110016755-110016755 on Build GRCh38
rs755777832 Chr.1: 110016757-110016758 on Build GRCh38
rs544261222 Chr.1: 110016649-110016649 on Build GRCh38
rs11544576 Chr.1: 110016368-110016368 on Build GRCh38
rs780554518 Chr.1: 110016766-110016766 on Build GRCh38
rs141465433 Chr.1: 110016383-110016383 on Build GRCh38
rs752322046 Chr.1: 110016715-110016715 on Build GRCh38
rs368265679 Chr.1: 110016503-110016503 on Build GRCh38
rs754337489 Chr.1: 110016628-110016628 on Build GRCh38
rs767260773 Chr.1: 110016630-110016630 on Build GRCh38
rs773175602 Chr.1: 110016635-110016635 on Build GRCh38
rs914315516 Chr.1: 110016548-110016548 on Build GRCh38
rs149672808 Chr.1: 110016347-110016347 on Build GRCh38
rs376546533 Chr.1: 110016770-110016770 on Build GRCh38
rs183814196 Chr.1: 110016563-110016563 on Build GRCh38
rs404275 Chr.1: 110016752-110016752 on Build GRCh38
rs375435451 Chr.1: 110016386-110016386 on Build GRCh38
rs187768149 Chr.1: 110016608-110016608 on Build GRCh38

View Full Product Details