Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232557589-232558152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E2
Entrez Gene ID
9470
Gene Name
eukaryotic translation initiation factor 4E family member 2
Gene Aliases
4E-LP, 4EHP, EIF4EL3, IF4e
UniGene
Hs.292026
Gene Chromosome Location
Chr.2: 232550587-232583645 on Build GRCh38
Transcripts
NM_001276336.1 NM_001276337.1 NM_001282958.1 NM_004846.3 XM_005246975.3 XM_011512206.2 XM_017005377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661647 c.205C>G p.P69A Chr.2: 232557953-232557953 on Build GRCh38
6157648 c.179A>C p.Y60S Chr.2: 232557927-232557927 on Build GRCh38
4764819 c.251A>G p.Q84R Chr.2: 232557999-232557999 on Build GRCh38
7167264 c.200G>C p.R67T Chr.2: 232557948-232557948 on Build GRCh38
4477096 c.211C>T p.R71C Chr.2: 232557959-232557959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188579107 Chr.2: 232557640-232557640 on Build GRCh38
rs192523391 Chr.2: 232558105-232558105 on Build GRCh38
rs765729211 Chr.2: 232557964-232557964 on Build GRCh38
rs779842924 Chr.2: 232558021-232558021 on Build GRCh38
rs911656781 Chr.2: 232557776-232557776 on Build GRCh38
rs34228509 Chr.2: 232557910-232557911 on Build GRCh38
rs751146807 Chr.2: 232557982-232557982 on Build GRCh38
rs1018630716 Chr.2: 232558108-232558108 on Build GRCh38
rs376444089 Chr.2: 232557865-232557865 on Build GRCh38
rs943833394 Chr.2: 232557796-232557796 on Build GRCh38
rs770687160 Chr.2: 232557635-232557635 on Build GRCh38
rs951550569 Chr.2: 232558083-232558083 on Build GRCh38
rs1023876088 Chr.2: 232557666-232557666 on Build GRCh38
rs540566064 Chr.2: 232557985-232557985 on Build GRCh38
rs554396964 Chr.2: 232558008-232558008 on Build GRCh38
rs1000827363 Chr.2: 232557829-232557829 on Build GRCh38
rs781237091 Chr.2: 232558000-232558000 on Build GRCh38
rs773350098 Chr.2: 232557943-232557943 on Build GRCh38
rs929674043 Chr.2: 232557700-232557700 on Build GRCh38
rs367941662 Chr.2: 232557926-232557926 on Build GRCh38
rs1039586192 Chr.2: 232557812-232557812 on Build GRCh38
rs200468379 Chr.2: 232557955-232557955 on Build GRCh38
rs918802292 Chr.2: 232557718-232557718 on Build GRCh38
rs758085264 Chr.2: 232557892-232557892 on Build GRCh38
rs149527191 Chr.2: 232557967-232557967 on Build GRCh38
rs958911844 Chr.2: 232558128-232558128 on Build GRCh38
rs1026325146 Chr.2: 232558068-232558068 on Build GRCh38
rs769552669 Chr.2: 232557911-232557911 on Build GRCh38
rs919552473 Chr.2: 232557937-232557937 on Build GRCh38
rs983188670 Chr.2: 232557735-232557735 on Build GRCh38
rs112617540 Chr.2: 232557668-232557668 on Build GRCh38
rs899739466 Chr.2: 232557813-232557813 on Build GRCh38
rs1039939878 Chr.2: 232558017-232558017 on Build GRCh38
rs566435158 Chr.2: 232557693-232557693 on Build GRCh38
rs754375724 Chr.2: 232557956-232557956 on Build GRCh38
rs776814967 Chr.2: 232558059-232558059 on Build GRCh38
rs1046856732 Chr.2: 232557717-232557717 on Build GRCh38
rs912300763 Chr.2: 232557986-232557986 on Build GRCh38
rs370882938 Chr.2: 232557902-232557902 on Build GRCh38
rs200177620 Chr.2: 232558005-232558005 on Build GRCh38
rs763222158 Chr.2: 232558064-232558064 on Build GRCh38
rs770885403 Chr.2: 232558048-232558048 on Build GRCh38
rs770904202 Chr.2: 232557901-232557901 on Build GRCh38
rs1034892639 Chr.2: 232558126-232558126 on Build GRCh38
rs1190439 Chr.2: 232557680-232557680 on Build GRCh38
rs761927224 Chr.2: 232557836-232557836 on Build GRCh38
rs778758674 Chr.2: 232557866-232557866 on Build GRCh38
rs921838653 Chr.2: 232557857-232557857 on Build GRCh38
rs760669411 Chr.2: 232557952-232557952 on Build GRCh38
rs564893561 Chr.2: 232558116-232558116 on Build GRCh38
rs368683481 Chr.2: 232558035-232558035 on Build GRCh38
rs374256886 Chr.2: 232558026-232558026 on Build GRCh38
rs760143860 Chr.2: 232558060-232558060 on Build GRCh38
rs767599592 Chr.2: 232557839-232557839 on Build GRCh38
rs944351609 Chr.2: 232557827-232557827 on Build GRCh38
rs371944603 Chr.2: 232558045-232558045 on Build GRCh38
rs374461889 Chr.2: 232558061-232558061 on Build GRCh38
rs759430772 Chr.2: 232557841-232557841 on Build GRCh38
rs766734353 Chr.2: 232557854-232557854 on Build GRCh38
rs760020998 Chr.2: 232557961-232557961 on Build GRCh38
rs754298867 Chr.2: 232557861-232557861 on Build GRCh38
rs773296546 Chr.2: 232558036-232558036 on Build GRCh38
rs758136547 Chr.2: 232557969-232557969 on Build GRCh38
rs772130461 Chr.2: 232557929-232557929 on Build GRCh38
rs777223942 Chr.2: 232557973-232557973 on Build GRCh38
rs761261423 Chr.2: 232557842-232557842 on Build GRCh38
rs763865175 Chr.2: 232558070-232558070 on Build GRCh38
rs896597825 Chr.2: 232557979-232557979 on Build GRCh38
rs745718108 Chr.2: 232557907-232557907 on Build GRCh38
rs538609623 Chr.2: 232557875-232557875 on Build GRCh38
rs541061215 Chr.2: 232557747-232557747 on Build GRCh38
rs951634122 Chr.2: 232557724-232557724 on Build GRCh38
rs199822591 Chr.2: 232558018-232558018 on Build GRCh38
rs774592191 Chr.2: 232557924-232557924 on Build GRCh38
rs148255880 Chr.2: 232557900-232557900 on Build GRCh38

View Full Product Details