Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25744846-25745356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPA
Entrez Gene ID
2551
Gene Name
GA binding protein transcription factor alpha subunit
Gene Aliases
E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07
UniGene
Hs.473470
Gene Chromosome Location
Chr.21: 25734947-25772460 on Build GRCh38
Transcripts
NM_001197297.1 NM_002040.3 XM_005260938.4 XM_011529520.2 XM_011529521.2 XM_017028313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5731342 c.79A>G p.I27V Chr.21: 25745211-25745211 on Build GRCh38
1632436 c.126A>G p.I42M Chr.21: 25745258-25745258 on Build GRCh38
725429 c.145G>C p.G49R Chr.21: 25745277-25745277 on Build GRCh38
725430 c.122C>G p.A41G Chr.21: 25745254-25745254 on Build GRCh38
281283 c.185C>A p.S62Y Chr.21: 25745317-25745317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749374327 Chr.21: 25745283-25745283 on Build GRCh38
rs771262795 Chr.21: 25745177-25745177 on Build GRCh38
rs35594642 Chr.21: 25745162-25745163 on Build GRCh38
rs201420702 Chr.21: 25745279-25745279 on Build GRCh38
rs774781678 Chr.21: 25745179-25745179 on Build GRCh38
rs959709149 Chr.21: 25745027-25745027 on Build GRCh38
rs552186514 Chr.21: 25744961-25744961 on Build GRCh38
rs190464308 Chr.21: 25745075-25745075 on Build GRCh38
rs753178683 Chr.21: 25745258-25745258 on Build GRCh38
rs749462335 Chr.21: 25745170-25745170 on Build GRCh38
rs764053336 Chr.21: 25744938-25744938 on Build GRCh38
rs1030411225 Chr.21: 25744918-25744918 on Build GRCh38
rs775449712 Chr.21: 25745213-25745213 on Build GRCh38
rs1009769835 Chr.21: 25744924-25744924 on Build GRCh38
rs765162436 Chr.21: 25745085-25745085 on Build GRCh38
rs1009555151 Chr.21: 25745214-25745214 on Build GRCh38
rs71327962 Chr.21: 25745069-25745072 on Build GRCh38
rs562336545 Chr.21: 25745025-25745025 on Build GRCh38
rs761373243 Chr.21: 25745229-25745229 on Build GRCh38
rs532284455 Chr.21: 25744940-25744940 on Build GRCh38
rs531354418 Chr.21: 25745029-25745029 on Build GRCh38
rs150628153 Chr.21: 25745134-25745134 on Build GRCh38
rs1021173106 Chr.21: 25744966-25744966 on Build GRCh38
rs1054107800 Chr.21: 25745088-25745088 on Build GRCh38
rs751862299 Chr.21: 25744952-25744955 on Build GRCh38
rs926454969 Chr.21: 25745076-25745076 on Build GRCh38
rs566091237 Chr.21: 25745148-25745148 on Build GRCh38
rs954801923 Chr.21: 25744932-25744932 on Build GRCh38
rs759344351 Chr.21: 25745194-25745194 on Build GRCh38
rs200493501 Chr.21: 25745230-25745230 on Build GRCh38
rs757716220 Chr.21: 25745110-25745110 on Build GRCh38
rs771813200 Chr.21: 25745212-25745212 on Build GRCh38
rs1051685005 Chr.21: 25744896-25744896 on Build GRCh38
rs143783879 Chr.21: 25745225-25745225 on Build GRCh38
rs544231877 Chr.21: 25745012-25745012 on Build GRCh38
rs753363596 Chr.21: 25745228-25745228 on Build GRCh38
rs369928917 Chr.21: 25744913-25744913 on Build GRCh38
rs61734758 Chr.21: 25745231-25745231 on Build GRCh38
rs547885721 Chr.21: 25745061-25745061 on Build GRCh38
rs373506995 Chr.21: 25745130-25745130 on Build GRCh38
rs756590811 Chr.21: 25745266-25745266 on Build GRCh38
rs950564091 Chr.21: 25745055-25745055 on Build GRCh38
rs936561287 Chr.21: 25745081-25745081 on Build GRCh38
rs891334684 Chr.21: 25744902-25744902 on Build GRCh38
rs1009607701 Chr.21: 25744917-25744917 on Build GRCh38
rs903825455 Chr.21: 25744969-25744969 on Build GRCh38
rs1007710364 Chr.21: 25744960-25744960 on Build GRCh38
rs1008066178 Chr.21: 25745327-25745327 on Build GRCh38
rs1018204616 Chr.21: 25744970-25744970 on Build GRCh38
rs963630541 Chr.21: 25744993-25744993 on Build GRCh38
rs1025225856 Chr.21: 25745052-25745052 on Build GRCh38
rs1000840282 Chr.21: 25745007-25745007 on Build GRCh38
rs1032309036 Chr.21: 25744875-25744875 on Build GRCh38
rs781437569 Chr.21: 25745237-25745237 on Build GRCh38
rs371865875 Chr.21: 25745036-25745036 on Build GRCh38
rs771174570 Chr.21: 25745313-25745313 on Build GRCh38

View Full Product Details