Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29320789-29321338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357228 c.11G>A p.S4N Chr.21: 29321291-29321291 on Build GRCh38
5792508 c.38C>G p.S13C Chr.21: 29321318-29321318 on Build GRCh38
6609522 c.50G>A p.S17N Chr.21: 29321330-29321330 on Build GRCh38
385355 c.46C>T p.H16Y Chr.21: 29321326-29321326 on Build GRCh38
6046967 c.45G>A p.V15V Chr.21: 29321325-29321325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989155874 Chr.21: 29320959-29320959 on Build GRCh38
rs374133191 Chr.21: 29321275-29321275 on Build GRCh38
rs1002002126 Chr.21: 29321213-29321213 on Build GRCh38
rs771621322 Chr.21: 29321285-29321285 on Build GRCh38
rs563072943 Chr.21: 29321153-29321153 on Build GRCh38
rs928011753 Chr.21: 29321193-29321193 on Build GRCh38
rs781710744 Chr.21: 29321297-29321297 on Build GRCh38
rs953845292 Chr.21: 29320818-29320818 on Build GRCh38
rs991123214 Chr.21: 29321209-29321209 on Build GRCh38
rs567518461 Chr.21: 29321026-29321026 on Build GRCh38
rs532237493 Chr.21: 29320846-29320846 on Build GRCh38
rs747863102 Chr.21: 29321258-29321258 on Build GRCh38
rs141543969 Chr.21: 29321291-29321291 on Build GRCh38
rs773597979 Chr.21: 29321309-29321309 on Build GRCh38
rs981109136 Chr.21: 29320875-29320875 on Build GRCh38
rs909965499 Chr.21: 29320840-29320840 on Build GRCh38
rs1020612695 Chr.21: 29321287-29321287 on Build GRCh38
rs897944831 Chr.21: 29321170-29321170 on Build GRCh38
rs568848960 Chr.21: 29320861-29320861 on Build GRCh38
rs998140937 Chr.21: 29321050-29321050 on Build GRCh38
rs1006190364 Chr.21: 29320826-29320826 on Build GRCh38
rs867579600 Chr.21: 29320918-29320918 on Build GRCh38
rs1024984339 Chr.21: 29321206-29321206 on Build GRCh38
rs752987492 Chr.21: 29321207-29321207 on Build GRCh38
rs531092587 Chr.21: 29320985-29320985 on Build GRCh38
rs578048152 Chr.21: 29321082-29321082 on Build GRCh38
rs543689666 Chr.21: 29320977-29320977 on Build GRCh38
rs755725197 Chr.21: 29321275-29321275 on Build GRCh38
rs558228761 Chr.21: 29321111-29321112 on Build GRCh38
rs1037701851 Chr.21: 29321160-29321160 on Build GRCh38
rs983913328 Chr.21: 29321095-29321095 on Build GRCh38
rs778789040 Chr.21: 29321133-29321133 on Build GRCh38
rs770992854 Chr.21: 29321273-29321273 on Build GRCh38
rs1017036489 Chr.21: 29321106-29321106 on Build GRCh38
rs182161842 Chr.21: 29321086-29321086 on Build GRCh38
rs777733150 Chr.21: 29321261-29321261 on Build GRCh38
rs749193862 Chr.21: 29321270-29321270 on Build GRCh38
rs926526436 Chr.21: 29320894-29320894 on Build GRCh38
rs762065180 Chr.21: 29321300-29321300 on Build GRCh38
rs867166575 Chr.21: 29320828-29320828 on Build GRCh38
rs889130970 Chr.21: 29320820-29320820 on Build GRCh38
rs368391255 Chr.21: 29321252-29321252 on Build GRCh38
rs192163096 Chr.21: 29321016-29321016 on Build GRCh38
rs1055767813 Chr.21: 29320893-29320893 on Build GRCh38
rs946881148 Chr.21: 29320957-29320957 on Build GRCh38
rs889337685 Chr.21: 29321124-29321124 on Build GRCh38
rs367633782 Chr.21: 29321301-29321301 on Build GRCh38
rs745439507 Chr.21: 29321276-29321276 on Build GRCh38
rs781536847 Chr.21: 29321250-29321250 on Build GRCh38
rs758653110 Chr.21: 29321212-29321212 on Build GRCh38
rs1024751723 Chr.21: 29320863-29320863 on Build GRCh38
rs536489990 Chr.21: 29321066-29321066 on Build GRCh38
rs993560247 Chr.21: 29321195-29321195 on Build GRCh38
rs753640646 Chr.21: 29320871-29320871 on Build GRCh38
rs1050677409 Chr.21: 29320811-29320811 on Build GRCh38
rs763385315 Chr.21: 29321311-29321311 on Build GRCh38
rs745600599 Chr.21: 29321258-29321258 on Build GRCh38
rs973160920 Chr.21: 29321146-29321146 on Build GRCh38
rs961423626 Chr.21: 29321113-29321113 on Build GRCh38
rs913673069 Chr.21: 29320960-29320960 on Build GRCh38
rs561920729 Chr.21: 29321038-29321038 on Build GRCh38
rs551858577 Chr.21: 29320847-29320847 on Build GRCh38
rs116727521 Chr.21: 29321260-29321260 on Build GRCh38

View Full Product Details