Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39820866-39821305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTHD1
Entrez Gene ID
150350
Gene Name
ENTH domain containing 1
Gene Aliases
CACNA1I, dJ370M22.3
UniGene
Hs.474869
Gene Chromosome Location
Chr.22: 39743044-39893884 on Build GRCh38
Transcripts
NM_152512.3 XM_006724149.3 XM_006724150.3 XM_006724151.1 XM_006724153.3 XM_011529927.2 XM_011529928.2 XM_011529931.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286025 c.748C>T p.P250S Chr.22: 39821077-39821077 on Build GRCh38
6324415 c.777C>T p.V259V Chr.22: 39821048-39821048 on Build GRCh38
4137475 c.832+6T>C p.? Chr.22: 39820987-39820987 on Build GRCh38
3554760 c.729G>A p.L243L Chr.22: 39821096-39821096 on Build GRCh38
5614884 c.829G>A p.A277T Chr.22: 39820996-39820996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757583484 Chr.22: 39821014-39821016 on Build GRCh38
rs767366391 Chr.22: 39820963-39820963 on Build GRCh38
rs755953008 Chr.22: 39821040-39821040 on Build GRCh38
rs1054855894 Chr.22: 39821193-39821193 on Build GRCh38
rs964217014 Chr.22: 39820996-39820996 on Build GRCh38
rs746063676 Chr.22: 39820978-39820978 on Build GRCh38
rs115492335 Chr.22: 39821109-39821109 on Build GRCh38
rs760611896 Chr.22: 39821158-39821158 on Build GRCh38
rs1037473950 Chr.22: 39821282-39821282 on Build GRCh38
rs757415348 Chr.22: 39821045-39821045 on Build GRCh38
rs776974725 Chr.22: 39821015-39821015 on Build GRCh38
rs765236440 Chr.22: 39820972-39820972 on Build GRCh38
rs759198692 Chr.22: 39821030-39821030 on Build GRCh38
rs142690162 Chr.22: 39821073-39821073 on Build GRCh38
rs762593608 Chr.22: 39821016-39821016 on Build GRCh38
rs981097598 Chr.22: 39821191-39821191 on Build GRCh38
rs769007786 Chr.22: 39821093-39821093 on Build GRCh38
rs759286537 Chr.22: 39821123-39821123 on Build GRCh38
rs749436618 Chr.22: 39820972-39820972 on Build GRCh38
rs376997525 Chr.22: 39820920-39820920 on Build GRCh38
rs746091066 Chr.22: 39821067-39821067 on Build GRCh38
rs146804198 Chr.22: 39821159-39821159 on Build GRCh38
rs753827617 Chr.22: 39821161-39821161 on Build GRCh38
rs949409291 Chr.22: 39821171-39821171 on Build GRCh38
rs561435749 Chr.22: 39821273-39821273 on Build GRCh38
rs775761712 Chr.22: 39821000-39821000 on Build GRCh38
rs761588854 Chr.22: 39821162-39821162 on Build GRCh38
rs941880378 Chr.22: 39821276-39821276 on Build GRCh38
rs753713322 Chr.22: 39821043-39821043 on Build GRCh38
rs925038789 Chr.22: 39821092-39821092 on Build GRCh38
rs778800311 Chr.22: 39821047-39821047 on Build GRCh38
rs774070537 Chr.22: 39821020-39821020 on Build GRCh38
rs559566853 Chr.22: 39820939-39820939 on Build GRCh38
rs762921359 Chr.22: 39821128-39821128 on Build GRCh38
rs957032500 Chr.22: 39821062-39821062 on Build GRCh38
rs756075838 Chr.22: 39820969-39820969 on Build GRCh38
rs577847629 Chr.22: 39821001-39821001 on Build GRCh38
rs895085927 Chr.22: 39821194-39821194 on Build GRCh38
rs752686988 Chr.22: 39820966-39820966 on Build GRCh38
rs780415643 Chr.22: 39821086-39821086 on Build GRCh38
rs770483413 Chr.22: 39821112-39821112 on Build GRCh38
rs764130053 Chr.22: 39821042-39821042 on Build GRCh38
rs545116511 Chr.22: 39821037-39821037 on Build GRCh38
rs143615095 Chr.22: 39821134-39821134 on Build GRCh38
rs747420037 Chr.22: 39821087-39821087 on Build GRCh38
rs773694952 Chr.22: 39821122-39821122 on Build GRCh38
rs926506529 Chr.22: 39821206-39821206 on Build GRCh38
rs138393426 Chr.22: 39821018-39821018 on Build GRCh38
rs767452165 Chr.22: 39821033-39821033 on Build GRCh38
rs372102762 Chr.22: 39820973-39820973 on Build GRCh38
rs767161415 Chr.22: 39821266-39821266 on Build GRCh38
rs769100029 Chr.22: 39821014-39821014 on Build GRCh38
rs775254057 Chr.22: 39821153-39821153 on Build GRCh38
rs754882247 Chr.22: 39820950-39820950 on Build GRCh38
rs374855607 Chr.22: 39820985-39820985 on Build GRCh38
rs758575050 Chr.22: 39821078-39821078 on Build GRCh38
rs758933589 Chr.22: 39820960-39820960 on Build GRCh38
rs34385292 Chr.22: 39821160-39821161 on Build GRCh38
rs765950707 Chr.22: 39821277-39821277 on Build GRCh38
rs201637698 Chr.22: 39821127-39821127 on Build GRCh38

View Full Product Details