Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39086324-39086711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3G
Entrez Gene ID
60489
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3G
Gene Aliases
A3G, ARCD, ARP-9, ARP9, CEM-15, CEM15, MDS019, bK150C2.7, dJ494G10.1
UniGene
Hs.660143
Gene Chromosome Location
Chr.22: 39077005-39087743 on Build GRCh38
Transcripts
NM_021822.3 XM_006724290.1 XM_017028903.1 XM_017028904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5605536 c.952C>T p.Q318* Chr.22: 39086495-39086495 on Build GRCh38
3912812 c.794T>G p.V265G Chr.22: 39086337-39086337 on Build GRCh38
7260576 c.924C>T p.C308C Chr.22: 39086467-39086467 on Build GRCh38
1183259 c.1018T>C p.Y340H Chr.22: 39086561-39086561 on Build GRCh38
7117708 c.894C>A p.F298L Chr.22: 39086437-39086437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750840279 Chr.22: 39086602-39086602 on Build GRCh38
rs142051350 Chr.22: 39086531-39086531 on Build GRCh38
rs370218151 Chr.22: 39086536-39086536 on Build GRCh38
rs546381147 Chr.22: 39086576-39086576 on Build GRCh38
rs747795976 Chr.22: 39086451-39086452 on Build GRCh38
rs17537574 Chr.22: 39086622-39086622 on Build GRCh38
rs760845966 Chr.22: 39086586-39086586 on Build GRCh38
rs778235663 Chr.22: 39086421-39086421 on Build GRCh38
rs199664843 Chr.22: 39086354-39086354 on Build GRCh38
rs910569055 Chr.22: 39086630-39086630 on Build GRCh38
rs375368638 Chr.22: 39086542-39086542 on Build GRCh38
rs947635898 Chr.22: 39086515-39086515 on Build GRCh38
rs1043433751 Chr.22: 39086649-39086649 on Build GRCh38
rs924919246 Chr.22: 39086668-39086668 on Build GRCh38
rs565345332 Chr.22: 39086621-39086621 on Build GRCh38
rs780812644 Chr.22: 39086615-39086615 on Build GRCh38
rs976609076 Chr.22: 39086617-39086617 on Build GRCh38
rs111358546 Chr.22: 39086648-39086648 on Build GRCh38
rs952376367 Chr.22: 39086580-39086580 on Build GRCh38
rs776436131 Chr.22: 39086357-39086357 on Build GRCh38
rs145107817 Chr.22: 39086510-39086510 on Build GRCh38
rs984421224 Chr.22: 39086650-39086650 on Build GRCh38
rs550655085 Chr.22: 39086675-39086675 on Build GRCh38
rs746526466 Chr.22: 39086540-39086540 on Build GRCh38
rs754179641 Chr.22: 39086592-39086592 on Build GRCh38
rs778841760 Chr.22: 39086558-39086558 on Build GRCh38
rs150838967 Chr.22: 39086541-39086541 on Build GRCh38
rs751626392 Chr.22: 39086394-39086394 on Build GRCh38
rs963671655 Chr.22: 39086692-39086692 on Build GRCh38
rs143828753 Chr.22: 39086380-39086380 on Build GRCh38
rs773097056 Chr.22: 39086560-39086560 on Build GRCh38
rs753026054 Chr.22: 39086407-39086407 on Build GRCh38
rs371126678 Chr.22: 39086501-39086501 on Build GRCh38
rs757630232 Chr.22: 39086599-39086599 on Build GRCh38
rs368035373 Chr.22: 39086564-39086564 on Build GRCh38
rs373206979 Chr.22: 39086468-39086468 on Build GRCh38
rs149026423 Chr.22: 39086373-39086373 on Build GRCh38
rs375494335 Chr.22: 39086519-39086519 on Build GRCh38
rs771345487 Chr.22: 39086428-39086428 on Build GRCh38
rs779575301 Chr.22: 39086429-39086429 on Build GRCh38
rs372449819 Chr.22: 39086618-39086618 on Build GRCh38
rs777340258 Chr.22: 39086452-39086452 on Build GRCh38
rs1022591962 Chr.22: 39086570-39086570 on Build GRCh38
rs545130640 Chr.22: 39086404-39086404 on Build GRCh38
rs757574695 Chr.22: 39086443-39086443 on Build GRCh38
rs751640043 Chr.22: 39086513-39086513 on Build GRCh38
rs768418669 Chr.22: 39086381-39086382 on Build GRCh38
rs5757471 Chr.22: 39086590-39086590 on Build GRCh38
rs918473281 Chr.22: 39086387-39086387 on Build GRCh38
rs755728234 Chr.22: 39086411-39086411 on Build GRCh38
rs749734529 Chr.22: 39086426-39086426 on Build GRCh38
rs868327835 Chr.22: 39086362-39086362 on Build GRCh38
rs138907659 Chr.22: 39086530-39086530 on Build GRCh38
rs961570517 Chr.22: 39086613-39086613 on Build GRCh38
rs910552728 Chr.22: 39086495-39086495 on Build GRCh38
rs776823029 Chr.22: 39086647-39086647 on Build GRCh38
rs139265278 Chr.22: 39086552-39086552 on Build GRCh38
rs367837968 Chr.22: 39086527-39086527 on Build GRCh38
rs775549186 Chr.22: 39086584-39086584 on Build GRCh38
rs542681958 Chr.22: 39086481-39086481 on Build GRCh38
rs138470593 Chr.22: 39086403-39086403 on Build GRCh38
rs770873704 Chr.22: 39086567-39086567 on Build GRCh38
rs528806696 Chr.22: 39086575-39086575 on Build GRCh38
rs779336410 Chr.22: 39086539-39086539 on Build GRCh38
rs771626944 Chr.22: 39086615-39086615 on Build GRCh38
rs923440157 Chr.22: 39086516-39086516 on Build GRCh38
rs765762449 Chr.22: 39086600-39086600 on Build GRCh38
rs772204741 Chr.22: 39086624-39086624 on Build GRCh38
rs371999625 Chr.22: 39086566-39086566 on Build GRCh38
rs183180481 Chr.22: 39086456-39086456 on Build GRCh38
rs761742463 Chr.22: 39086365-39086365 on Build GRCh38
rs17496046 Chr.22: 39086366-39086366 on Build GRCh38
rs757732607 Chr.22: 39086538-39086538 on Build GRCh38
rs575305230 Chr.22: 39086467-39086467 on Build GRCh38
rs758949719 Chr.22: 39086606-39086606 on Build GRCh38
rs766418500 Chr.22: 39086391-39086391 on Build GRCh38
rs767773347 Chr.22: 39086520-39086520 on Build GRCh38
rs375546536 Chr.22: 39086572-39086572 on Build GRCh38
rs142987758 Chr.22: 39086455-39086455 on Build GRCh38
rs760308553 Chr.22: 39086348-39086348 on Build GRCh38
rs769604992 Chr.22: 39086559-39086559 on Build GRCh38
rs746521588 Chr.22: 39086437-39086437 on Build GRCh38

View Full Product Details