Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38063635-38063951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEC1
Entrez Gene ID
9940
Gene Name
deleted in lung and esophageal cancer 1
Gene Aliases
CFAP81, DLC-1, DLC1, F56
UniGene
Hs.714499
Gene Chromosome Location
Chr.3: 38038595-38122741 on Build GRCh38
Transcripts
NM_001321153.1 NM_007335.3 NM_007337.3 XM_006713438.3 XM_006713439.3 XM_006713440.3 XM_011534317.2 XM_011534318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002665 c.1129C>G p.P377A Chr.3: 38063875-38063875 on Build GRCh38
7101378 c.1102G>T p.D368Y Chr.3: 38063848-38063848 on Build GRCh38
7101377 c.1102G>T p.D368Y Chr.3: 38063848-38063848 on Build GRCh38
4002664 c.1129C>G p.P377A Chr.3: 38063875-38063875 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761086025 Chr.3: 38063738-38063738 on Build GRCh38
rs878983556 Chr.3: 38063766-38063766 on Build GRCh38
rs759741718 Chr.3: 38063887-38063887 on Build GRCh38
rs1031043257 Chr.3: 38063732-38063732 on Build GRCh38
rs377480209 Chr.3: 38063681-38063681 on Build GRCh38
rs140010429 Chr.3: 38063925-38063925 on Build GRCh38
rs933661052 Chr.3: 38063694-38063694 on Build GRCh38
rs552816446 Chr.3: 38063742-38063742 on Build GRCh38
rs370671078 Chr.3: 38063804-38063804 on Build GRCh38
rs531896839 Chr.3: 38063912-38063912 on Build GRCh38
rs956572737 Chr.3: 38063748-38063748 on Build GRCh38
rs763399339 Chr.3: 38063841-38063841 on Build GRCh38
rs751804734 Chr.3: 38063879-38063879 on Build GRCh38
rs947971865 Chr.3: 38063840-38063840 on Build GRCh38
rs988565798 Chr.3: 38063662-38063662 on Build GRCh38
rs878860824 Chr.3: 38063767-38063767 on Build GRCh38
rs538593183 Chr.3: 38063783-38063783 on Build GRCh38
rs770059469 Chr.3: 38063818-38063818 on Build GRCh38
rs1030357817 Chr.3: 38063678-38063678 on Build GRCh38
rs200640048 Chr.3: 38063829-38063829 on Build GRCh38
rs372927373 Chr.3: 38063822-38063822 on Build GRCh38
rs991991658 Chr.3: 38063779-38063779 on Build GRCh38
rs773813909 Chr.3: 38063683-38063683 on Build GRCh38
rs1039020623 Chr.3: 38063808-38063808 on Build GRCh38
rs898673675 Chr.3: 38063825-38063826 on Build GRCh38
rs766549926 Chr.3: 38063822-38063823 on Build GRCh38
rs1018247276 Chr.3: 38063707-38063707 on Build GRCh38
rs117013868 Chr.3: 38063875-38063875 on Build GRCh38
rs111637593 Chr.3: 38063720-38063720 on Build GRCh38
rs36022978 Chr.3: 38063818-38063818 on Build GRCh38
rs145773322 Chr.3: 38063706-38063706 on Build GRCh38
rs979448507 Chr.3: 38063729-38063729 on Build GRCh38
rs372015250 Chr.3: 38063891-38063891 on Build GRCh38

View Full Product Details