Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32725315-32725739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT10
Entrez Gene ID
25904
Gene Name
CCR4-NOT transcription complex subunit 10
Gene Aliases
-
UniGene
Hs.444851
Gene Chromosome Location
Chr.3: 32685145-32773875 on Build GRCh38
Transcripts
NM_001256741.1 NM_001256742.1 NM_015442.2 NR_046352.1 XM_006713084.3 XM_006713085.3 XM_011533566.2 XM_011533567.2 XM_017006109.1 XM_017006110.1 XM_017006111.1 XM_017006112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059649 c.878G>A p.C293Y Chr.3: 32725465-32725465 on Build GRCh38
4323609 c.1055G>T p.R352I Chr.3: 32725462-32725462 on Build GRCh38
3767397 c.863-1G>T p.? Chr.3: 32725449-32725449 on Build GRCh38
3767398 c.1043-1G>T p.? Chr.3: 32725449-32725449 on Build GRCh38
3823670 c.961C>G p.Q321E Chr.3: 32725548-32725548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780407052 Chr.3: 32725467-32725467 on Build GRCh38
rs994251894 Chr.3: 32725582-32725582 on Build GRCh38
rs878862934 Chr.3: 32725629-32725629 on Build GRCh38
rs368383242 Chr.3: 32725618-32725618 on Build GRCh38
rs766685444 Chr.3: 32725579-32725579 on Build GRCh38
rs749469656 Chr.3: 32725537-32725537 on Build GRCh38
rs754843838 Chr.3: 32725439-32725440 on Build GRCh38
rs778149139 Chr.3: 32725524-32725524 on Build GRCh38
rs150438661 Chr.3: 32725502-32725502 on Build GRCh38
rs763544063 Chr.3: 32725420-32725420 on Build GRCh38
rs781760762 Chr.3: 32725608-32725608 on Build GRCh38
rs979616017 Chr.3: 32725401-32725401 on Build GRCh38
rs944341695 Chr.3: 32725580-32725580 on Build GRCh38
rs749504496 Chr.3: 32725621-32725621 on Build GRCh38
rs750851789 Chr.3: 32725462-32725462 on Build GRCh38
rs911382142 Chr.3: 32725387-32725387 on Build GRCh38
rs758693045 Chr.3: 32725465-32725465 on Build GRCh38
rs760617200 Chr.3: 32725414-32725414 on Build GRCh38
rs765303681 Chr.3: 32725677-32725677 on Build GRCh38
rs979555335 Chr.3: 32725714-32725714 on Build GRCh38
rs762613815 Chr.3: 32725577-32725577 on Build GRCh38
rs761305787 Chr.3: 32725558-32725558 on Build GRCh38
rs925665898 Chr.3: 32725407-32725407 on Build GRCh38
rs748214308 Chr.3: 32725520-32725520 on Build GRCh38
rs372368232 Chr.3: 32725552-32725552 on Build GRCh38
rs761504002 Chr.3: 32725439-32725439 on Build GRCh38
rs188844579 Chr.3: 32725400-32725400 on Build GRCh38
rs779766447 Chr.3: 32725634-32725634 on Build GRCh38
rs563068800 Chr.3: 32725607-32725607 on Build GRCh38
rs1038174436 Chr.3: 32725560-32725560 on Build GRCh38
rs374522809 Chr.3: 32725671-32725671 on Build GRCh38
rs150956266 Chr.3: 32725384-32725384 on Build GRCh38
rs1027475331 Chr.3: 32725545-32725545 on Build GRCh38
rs768620008 Chr.3: 32725555-32725555 on Build GRCh38
rs141044437 Chr.3: 32725650-32725650 on Build GRCh38
rs759652990 Chr.3: 32725522-32725522 on Build GRCh38
rs751920279 Chr.3: 32725604-32725604 on Build GRCh38
rs771836452 Chr.3: 32725405-32725405 on Build GRCh38
rs765304468 Chr.3: 32725700-32725700 on Build GRCh38
rs771157793 Chr.3: 32725626-32725626 on Build GRCh38
rs894439988 Chr.3: 32725423-32725423 on Build GRCh38
rs767243169 Chr.3: 32725344-32725344 on Build GRCh38
rs753316569 Chr.3: 32725425-32725425 on Build GRCh38
rs76520124 Chr.3: 32725368-32725368 on Build GRCh38
rs868462187 Chr.3: 32725419-32725419 on Build GRCh38
rs771786675 Chr.3: 32725542-32725542 on Build GRCh38
rs753190187 Chr.3: 32725615-32725615 on Build GRCh38
rs756277100 Chr.3: 32725617-32725617 on Build GRCh38
rs750086206 Chr.3: 32725565-32725565 on Build GRCh38
rs746808851 Chr.3: 32725635-32725635 on Build GRCh38
rs768650184 Chr.3: 32725641-32725641 on Build GRCh38
rs764658347 Chr.3: 32725564-32725564 on Build GRCh38
rs775320677 Chr.3: 32725544-32725544 on Build GRCh38
rs533096801 Chr.3: 32725658-32725658 on Build GRCh38
rs755468651 Chr.3: 32725478-32725478 on Build GRCh38
rs200963829 Chr.3: 32725556-32725556 on Build GRCh38
rs1025592160 Chr.3: 32725597-32725597 on Build GRCh38
rs1046082404 Chr.3: 32725501-32725501 on Build GRCh38
rs6550155 Chr.3: 32725413-32725413 on Build GRCh38
rs150996180 Chr.3: 32725468-32725468 on Build GRCh38
rs764858731 Chr.3: 32725442-32725442 on Build GRCh38

View Full Product Details